Simplifying controls for oncology with AcroMetrix products
As oncology testing expands, laboratories are challenged with finding highly characterized, standardized and reproducible quality control materials to support validation or verification of new assays. AcroMetrix control products for oncology testing aid laboratories in meeting quality and regulatory requirements, while supporting the goal of standardization in molecular testing by establishing traceable, consistent, accurate, and meaningful test results. 
AcroMetrix products for oncology testing
Solid tumor controls
Highly multiplexed quality control to assess performance of next-generation sequencing (NGS) assays that detect somatic mutations.
Conceptual image of a cancer cell and DNA molecule
Solid tumor validation panels
NGS validation ladder featuring 555 hotspots representative across various cancers at varying tumor burden levels. For use in analytical validation of various oncology NGS assays.
Conceptual image of cancer cells
Liquid biopsy
Platform-agnostic reference material to help standardize ctDNA research endeavors.
Conceptual image of liquid biopsy showing DNA in a tube of blood
Order AcroMetrix oncology quality control products
Quality Controls
Verification & validation panels
CE marked in accordance with Requirements of European Regulation (EU) 2017/746. 
Not all products are CE marked or have 510(k) clearance for sales in the U.S.
Availability of products in each country depends on local regulatory marketing authorization status.
IVD = In-Vitro Diagnostic
RUO = Research Use Only