Cryo-transmission electron microscope for high-fidelity atomic resolution cryo-EM
The 300 kV Thermo Scientific Krios 5 Cryo-Transmission Electron Microscope (Cryo-TEM) has been designed for high-performance single particle analysis (SPA), cellular cryo-electron tomography (cryo-ET) and microcrystal electron diffraction (MicroED). The Krios 5 Cryo-TEM is capable of quickly producing high-quality structures thanks to a unique focus on high throughput with high data fidelity, increased automation of routine alignments, improved optical precision, and AI-enabled software, which make it a highly productive and “ready-to-use” system for all expertise levels across multiple scientific fields.
Experience up to 25% faster throughput relative to previous Krios Cryo-TEM generations with high data fidelity for single particle analysis. Extended aberration-free image shift (AFIS) accelerates data collection, while AI-powered Thermo Scientific Smart EPU Software plugins automate routine alignments. Thermo Scientific Tomography 5 Software provides high image quality via automatic 8K Electron Event Representation (EER) decoding and on-the-fly motion correction for crisp tomograms, unlocking the full potential of the Thermo Scientific Falcon 4i Direct Electron Detector beyond the Nyquist limit. Easily transfer Autogrid cassettes between Autoloaders contamination-free with the Thermo Scientific Vacuum Capsule. Designed for sustainability, the Krios 5 Cryo-TEM features energy-saving Eco modes and environmental footprint traceability with a My Green Lab ACT Ecolabel.
Krios 5 Cryo-TEM benefits for cell and structural biology research
High-data-fidelity atomic-resolution cryo-EM
The Krios 5 Cryo-TEM offers up to 25% faster throughput with high data fidelity, extended aberration-free image shift (AFIS) and magnification calibration accuracy within 1% for single particle analysis. Combined with the optional Thermo Scientific Selectris X Imaging Filter and E-CFEG, the Krios 5 Cryo-TEM can deliver enhanced image contrast for rapid, atomic-resolution imaging.
Faster single particle analysis with AI-automation
AI-assistance with Smart EPU Software enables automated grid screening, quality monitoring, and data management. Smart Plugins in the Smart EPU Software use AI-based image analysis to automate grid area selection and dynamically adjust settings based on real-time analysis.
Accelerated, contamination-free cryo-electron tomography workflow
Tomography 5 Software provides automatic 8K Electron Event Representation (EER) decoding and on-the-fly motion correction for crisp tomograms by unlocking the full data potential beyond the Nyquist limit. Manage tomograms and experimental parameters with the Discovery Viewer in Thermo Scientific CryoFlow Software.  Easily transfer Autogrid cassettes between Autoloaders contamination-free with the Vacuum Capsule.
Energy-saving Eco modes and My Green Lab ACT Ecolabel
The Krios 5 Cryo-TEM features energy-saving Eco modes with fast “wake up” scheduling, plus environmental impact certification with the My Green Lab® ACT® Ecolabel
Learn more about cryo-electron microscopy with the Krios 5 Cryo-TEM
Unlock data beyond the Nyquist frequency and accelerate your cryo-ET workflow
Tomography 5 Software provides 8k electron-event representation decoding and on-the-fly motion correction, automatically capturing data beyond the Nyquist frequency for crisp tomograms and the high image quality required for sub-tomogram averaging. 
Chlamydomonas reinhardtii 80S ribosome protein structure determined with Krios Cryo TEM
AI-assistance with SmartEPU Software
AI-powered SmartEPU plugins automate data acquisition by analyzing images in real-time. They can be used to select optimal grid squares, adjust focus, classify foil holes, predict ice thickness, and skip poor-quality areas, significantly enhancing data collection efficiency in single particle analysis.
Preview of Smart EPU Software, used to select foil holes for cryo EM imaging
Automation of daily alignments
Routine alignments, including beam and aperture centering as well as coma and astigmatism correction, are fully automated. Magnification calibration within 1% and a diagnostics dashboard ensure optimal maintenance.
Preview of optics alignment in Thermo Scientific Microscope Companion Software for cryo EM
Low-power Eco modes
The low-power Eco modes of the Krios 5 Cryo-TEM deactivate specific parts of the instrument when it is not in use, reducing power consumption up to 80%. The software can be programmed for early wake-up, ensuring the system is ready when needed.
Front facing image of Krios 5 Cryo-TEM in Eco mode
Designed-in sustainability with the ACT Ecolabel
In addition to the energy-saving Eco modes of the Krios 5 Cryo-TEM, the My Green Lab ACT Ecolabel transparently documents the instrument’s environmental footprint, including manufacturing, energy and water use, as well as end-of-life disposal. At Thermo Fisher Scientific, our commitment to sustainability supports our mission to make the world healthier, cleaner, and safer.
ACT label logo
Krios 5 Cryo-TEM imaging capabilities
Upgrade options for previous-generation Krios Cryo-TEMs
Electron source: cold field emission gun (E-CFEG)
E-CFEG is a low-energy spread cold field emission gun for higher contrast at truly atomic resolution. Low energy spread (≤0.3 eV) enables higher contrast at high resolutions (≤2.0 Å); Improved contrast at high spatial frequencies to reach higher resolutions in less time.
Comparison of field emission gun contrast on Krios Cryo TEM for atomic resolution cryo EM Comparison of a Thermo Scientific high-brightness field emission gun and the cold field emission gun on the Krios Cryo-TEM.
Krios Cryo-TEM accessories and software
Increase your lab’s productivity and performance with innovative upgrades:
Falcon 4i Direct Electron Detector
Cryo-EM research in BSL-3 containment facilities
Work on contagious pathogens frequently requires specialized biosafety containment facilities. Until recently, cryo-EM equipment was not commonly found in these laboratories. At Thermo Fisher Scientific, we have developed products and services designed for these higher BSL facilities, so that our customers can accelerate their critical pathogen research with cryo-electron microscopy.
Cryo-EM in Biosafety Facilities for Virology Research
Krios Cryo-TEM technical specifications
Krios Cryo TEM Datasheet
Selectris Imaging Filters
Falcon 4i Direct Electron Detector
Vacuum Capsule datasheet
Smart EPU Software datasheet
Tomography 5 Software datasheet