Talos (Scanning) Transmission Electron Microscope capabilities
Thermo Scientific Talos Scanning Transmission Electron Microscopes ((S)TEMs) incorporate streamlined capabilities with high-throughput and flexibility for research, characterization, analysis, and metrology. These systems make scanning transmission electron microscopy (STEM) accessible to users of all experience levels by empowering access to precise, high-resolution insights into sample characteristics. From imaging and analysis with intuitive software to spectroscopy and diffraction, these user-friendly instruments let you explore nanoscale applications across diverse fields.
Whether you are investigating novel materials to discover the maximum potential of their physical and chemical properties, performing fundamental materials research, quality control, failure analysis, or conducting biological imaging, Talos (S)TEMs can help you acquire key insights down to the atomic scale.
Scanning transmission electron microscopy
Scanning Transmission Electron Microscopy STEM is a pivotal technology for materials science, life sciences, and semiconductor fields, unlocking detailed insights into the atomic structure and composition of samples that are not achievable with other techniques. By rastering a focused electron beam over a thin sample, STEM generates several distinct signals that reveal critical structural and compositional details. Primary signals include Bright-Field (BF), which captures electrons passing directly through the sample to highlight mass and thickness differences, and Annular Dark-Field (ADF), High-Angle Annular Dark-Field (HAADF) which enhances contrast based on atomic number (Z-contrast) by detecting scattered electrons, while Electron Energy Loss Spectroscopy (EELS) provides chemical and electronic information by measuring electron energy loss, and X-ray Energy Dispersive Spectroscopy (EDS) detects characteristic X-rays for elemental analysis. Together, these imaging modes make STEM instrumental in advancing our understanding of nanoscale function and performance across a variety of fields.
Panther STEM Detector and AutoSTEM Software
The Thermo Scientific Panther STEM Detector, part of the Talos portfolio, stands out as a superior solution in the scanning transmission electron microscopy landscape. Fine-tuned for excellence with improved detectors and amplifiers, the Panther STEM Detector excels in multi-signal acquisition and mechanical alignment accuracy. This innovative system offers high throughput, user-friendly operation, and incredible flexibility in signal processing. Its ultra-high electron sensitivity for low-dose applications, coupled with up to 16 segments, sets it apart from competitors and makes it a preferred choice for materials science researchers. Going one step further, Thermo Scientific AutoSTEM Software enhances the Panther STEM Detector, enabling automated focus and astigmatism correction for consistently high-quality images. The embedded piezo-enhanced stage and the seamless ability to switch from HRTEM to HRSTEM further contribute to its efficiency and adaptability.
Panther STEM Detector and AutoSTEM Software Autofocus and astigmatism for fast, high-resolution STEM imaging.
HRSTEM with the Talos (S)TEM X-TWIN Lens
The Talos (S)TEM X-TWIN lens provides highly advanced scanning transmission electron microscopy resolution and analytical capabilities. The large X-TWIN pole piece gap, that provides high flexibility for a wide range of applications, combined with a reproducibly performing electron column opens new opportunities for high-resolution 3D and in situ dynamic observations. The X-TWIN is available on the Thermo Scientific Talos F200X, F200S, F200i, and F200E (S)TEMs. The 20–200 kV Talos product line is equipped with a fast 4k resolution Thermo Scientific Ceta Camera, which provides, in combination with drift compensated frame integration (DCFI), large field-of-view, drift-compensated imaging with high sensitivity, high speed, and precise sample navigation on a 64-bit platform. Talos (S)TEMs also include the workflow-based Thermo Scientific Velox Software for intuitive acquisition and analysis. All these features combine to make the Talos product line incredibly productive for all facets of STEM.
HRSTEM with the Talos TEM X-TWIN Lens High-resolution DCFI scanning transmission electron microscopy image of cross-section of WOx nanowire. Sample courtesy of Vojtech Kundrat and Prof. Reshef Tenne, Weizmann Institute of Science.
Integrated differential phase contrast transmission electron microscopy for the imaging of all elements
The iDPC scanning transmission electron microscopy method on Talos (S)TEMs exposes low-Z elements with bright contrast and dark background, placing considerably less dependence on defocus and/or thickness. In addition, it has been shown that iDPC-STEM images have a higher signal to noise ratio compared to ABF-STEM images. A higher signal to noise ratio provides the possibility of low-dose imaging, which is crucial for beam-sensitive materials and charge-prone samples.
iDPC scanning transmission electron microscopy method iDPC image of MIL 101 metal organic framework (MOF).
Spectroscopy with the Talos Scanning Transmission Electron Microscope
For the exploration of the analytical imaging frontier, broad-spectrum analytical imaging exemplifies electron microscopy's proven flexibility and agility through its wide array of analytical options. With foundational techniques such as energy dispersive X-ray spectroscopy (EDS) for atomic-scale elemental and phase characterization to electron energy loss spectroscopy (EELS) for insights into electronic and optical properties, this approach underscores the adaptability of STEM for diverse research needs. The Talos platform allows for a symmetrical detector design and the advanced X-CFEG for the high flexibility to enhance both energy dispersive X-ray spectroscopy and electron energy loss spectroscopy analyses. This configuration not only improves the precision of material characterization, but also showcases the instrument's ability to adapt and excel in various analytical scenarios for complex materials science challenges.
Elemental microanalysis with energy dispersive X-ray spectroscopy
Energy dispersive X-ray spectroscopy (EDS) remains a foundational pillar in the world of microanalysis, enabling detailed elemental and phase characterization down to the atomic scale. By capitalizing on the principle of characteristic X-ray emission from electron-sample interactions, EDS produces a comprehensive spectrum that reveals the detailed elemental composition of a specimen. This technique is a mainstay for scientists, technicians, and semiconductor engineers whereby offering both qualitative and quantitative insights into the intricate structure of samples.
In the evolving landscape of advanced materials and semiconductor device characterization, the Talos (S)TEM emerges as a synergistic tool, enhancing the capabilities of EDS. With its integration of the established Thermo Scientific ChemiSTEM Technology, especially with the ultra-high-brightness offered by the X-CFEG, the Talos (S)TEM optimizes the signal-to-noise ratio and facilitates the detection of trace elements with heightened sensitivity. This combination helps ensure that users are equipped with state-of-the-art tools for meticulous materials analysis.
Elemental microanalysis with energy dispersive X-ray spectroscopy Cadmium sulfide sample, mostly used for optical applications, analyzed with side-entry EDS. Sample courtesy Prof. Li Haidong, Qingdao University.
Electron energy-loss spectroscopy with the extreme-brightness cold field emission gun
Electron energy loss spectroscopy (EELS) is a cornerstone technique in advanced electron microscopy, providing deep insights into elemental composition, chemical bonding, oxidation states, and electronic properties of both valence and conduction bands. The Talos (S)TEM, equipped with the X-CFEG, stands out in the realm of electron energy loss spectroscopy. The X-CFEG offers excellent electron beam brightness and stability, which is crucial for EELS, to ensure high-energy-resolution imaging and top performance analysis.
As the challenges in materials science and semiconductor development grow in parallel with the study of increasingly complex structures, the demand for precise characterization from cutting-edge instrumentation becomes evident. The Talos (S)TEM, with its advanced features including the X-CFEG, offers superb precision and versatility to help meet these demands and solidify the Talos (S)TEM's position in modern materials research.
Electron energy-loss spectroscopy Atomically resolved STO STEM and EELS spectrum images with an energy resolution of <0.40 eV with 220 pA. Results were shot on a Talos F200 (S)TEM with X-CFEG in only two minutes.
Electron energy-loss spectroscopy The X-CFEG is superior to other guns with respect to brightness, source size, and energy spread. Here a Cobalt oxide STEM + EELS spectrum is shown with an energy resolution of <0.26 eV (red). The same sample was analyzed on a X-FEG with 0.7 eV energy resolution (green). The X-CFEG spectrum fine structure (red) is resolved better, revealing a shoulder on the Co L3 which is Co3. This shoulder is not visible on the green spectrum. Results were shot on a Talos F200 with X-CFEG (red) and X-FEG (green), respectively, and Panther STEM with Gatan Continuum 1086 at 200kv.
Scanning transmission electron microscopy data acquisition and analysis with intuitive software
Data acquisition is a critical process in scientific research and engineering applications, allowing the collection of valuable information for various purposes. The Talos (S)TEM is a state-of-the-art platform that offers exceptional imaging and analytical capabilities as well as automation that enables users to obtain high-quality data effectively. Featuring an intuitive user interface that is easily leveraged by users of all experience levels, the Talos (S)TEM provides a doorway to valuable insights of structure, composition, and properties of materials, paving the way for breakthroughs in various scientific disciplines.
Velox Software for fast materials analysis data acquisition
Integrated Thermo Scientific Velox Software makes data acquisition fast and easy while supporting analysis of multimodal data. Several automation packages are available on the Talos (S)TEM to help you get better insights into the chemical and structural bonding of materials. Seeing the bigger picture helps in understanding and reveals details that are required to develop new materials.
Velox Software for fast materials analysis data acquisition Simple, clean, and easy-to-use graphical user interface of Velox Software.
High-resolution 3D energy dispersive X-ray spectroscopy tomography
In the advanced realm of STEM, the incorporation of 3D energy dispersive X-ray spectroscopy tomography offers an unprecedented depth of material characterization. This synergy allows for high-resolution, three-dimensional elemental mapping, transcending the constraints of traditional 2D EDS analyses within STEM studies. The acquisition of volumetric data not only visualizes elemental distributions with nanoscale precision but also reveals intricate interfaces, compositional gradients, and inherent material heterogeneities. As we navigate the challenges of materials science, nanotechnology, semiconductors, and intricate biology, the integration of 3D energy dispersive X-ray spectroscopy tomography in STEM stands as a testament to the next frontier in nanoscale material analysis.
The geometries and positioning of the Super-X and Dual-X energy dispersive X-ray spectroscopy systems are ideal for providing 3D chemical composition information at high lateral resolutions, an essential application for any analytical facility. We offer a fully automated and comprehensive energy dispersive X-ray spectroscopy tomography application package including the tomography holder, the symmetrical Super-X/Dual-X Detector, the software to acquire and analyze tilt series automatically, and our Thermo Scientific Inspect3D Reconstruction Software and Avizo Visualization Software.
Thermo Scientific Automated Energy Dispersive X-ray Spectroscopy Tomography Software enables fast and easy setup with flexible conditions for optimizing throughput and data quality. You can set up tilt range and scheme, increment steps, and choose which detector(s) to use. The auto-functions in the Tomography Software allow for fully automated acquisition, which in turn allows unattended data collection, and is particularly useful for overnight experiments.
Maps Software for multi-scale materials analysis
Modern materials science requires multimodal, statistically meaningful data to obtain sample properties across platforms on multiple length scales. This has become increasingly important for research on nanoparticles and catalysis samples as well as for precipitates in metals. Thermo Scientific Maps Software for STEM and EDS automates navigation across multiple instruments and length scales to characterize large volumes quickly and reproducibly, providing accurate contextual analysis on relevant areas.
Maps Software for multi-scale materials analysis
Automated Particle Analysis Workflow for sample and particle analysis
The Thermo Scientific Automated Particle Workflow (APW) is an ideal solution for nanoparticle characterization, combining large-area and high-resolution imaging acquisition. It streamlines the workflow from sample to particle analysis by automating navigation, data acquisition, particle segmentation, and statistical analysis of morphological as well as chemical information. Automation controls navigation, focus, drift correction, tiling, stitching, and storing, all of which can be performed overnight and/or unattended. Process improvement can be completed on a much shorter schedule. Statistically relevant data sets on complex nanoparticles can be compiled in a single day with high-resolution, automated STEM.
Automated Particle Analysis Workflow Automated Particle Workflow example: Pt-Rh particle analysis: size, area, perimeter, shape factor, contacts, etc. Sample courtesy of Prof. B Gorman and Prof. R. Richards, Colorado School of Mines.
STEM imaging for life sciences research
Thermo Scientific Talos Scanning Transmission Electron Microscopes can be used for 2D or 3D visualization of biological samples from cells to tissues to macromolecular assemblies. STEM techniques can be performed at both ambient and cryogenic temperatures. Common techniques include:
  • Biological particle characterization—Negative stain TEM or cryo-TEM to assess critical quality attributes such as particle size, distribution, morphology, empty/full classification, or particle aggregation
  • Single particle analysis—Cryo-electron microscopy technique enabling structural characterization 
  • 3D STEM—Obtain structural information and spatial arrangements of intracellular compartments within a cell or for pleiomorphic samples such as enveloped viruses or lipid nanoparticles
  • Cryo-tomography—Cryo-electron microscopy technique delivering both structural and spatial arrangement insights
  • Microcrystal electron diffraction (MicroED) —Cryo-electron microscopy technique for structural determination of small molecules and proteins from nanocrystals
For Research Use Only. Not for use in diagnostic procedures.