48ab - Appl/Samples/Products/Resources/Contact
Electron Energy Loss Spectroscopy
Over the last three decades, electron energy loss spectroscopy (EELS) has become a formidable characterization technique. Used in combination with a transmission electron microscope (TEM), it can provide atomic-level chemical and structural characterization. This includes information about elemental composition, chemical bonding, oxidation states, valence and conduction band electronic properties, and surface properties. It can also be used in conjunction with other techniques, such as electron tomography, to provide advanced 3D mapping of elements and oxidation states.
As materials characterization becomes increasingly more demanding, there is a growing need for highly localized characterization of increasingly complex structures with ever smaller features. Fortunately, improvements in EELS and S/TEM instrumentation have kept pace with these demands, with advances including spherical aberration correction of the S/TEM lenses, as well as the use of monochromators to improve energy resolution, providing sub-angstrom spatial resolution and a higher probe current.
EELS TEM
With our X-FEG/Mono, X-FEG/UltiMono and X-CFEG guns on the Thermo Scientific Spectra and X-FEG on the Thermo Scientific Talos S/TEM platforms, Thermo Fisher Scientific has all your EELS applications covered. These systems allow you to perform ultra-high resolution EELS analysis to obtain valuable structural, elemental, and chemical information at the atomic scale.
Resources
Video Player is loading.
Current Time 0:00
Duration 2:18
Loaded: 7.20%
Stream Type LIVE
Remaining Time 2:18
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • default, selected
    OptiMono+ exciting an X-FEG/UltiMono from the monochromator off state (with 1 eV energy resolution) to the monochromator fully excited state (<30 meV) at 60 kV.
    Video Player is loading.
    Current Time 0:00
    Duration 0:30
    Loaded: 33.17%
    Stream Type LIVE
    Remaining Time 0:30
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      Localized positions of plasmon excitations along a gold nanowire as a function of their excitation energy (between 0.18 – 1.2 eV), investigated with an electron probe of <0.2 eV energy resolution.
      Video Player is loading.
      Current Time 0:00
      Duration 2:18
      Loaded: 7.20%
      Stream Type LIVE
      Remaining Time 2:18
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • default, selected
        OptiMono+ exciting an X-FEG/UltiMono from the monochromator off state (with 1 eV energy resolution) to the monochromator fully excited state (<30 meV) at 60 kV.
        Video Player is loading.
        Current Time 0:00
        Duration 0:30
        Loaded: 33.17%
        Stream Type LIVE
        Remaining Time 0:30
        1x
          • Chapters
          • descriptions off, selected
          • captions off, selected
          • en (Main), selected
          Localized positions of plasmon excitations along a gold nanowire as a function of their excitation energy (between 0.18 – 1.2 eV), investigated with an electron probe of <0.2 eV energy resolution.
          Applications
          Fundamental Materials Research_R&amp;D_Thumb_274x180_144DPI
          Fundamental Materials Research
          Novel materials are investigated at increasingly smaller scales for maximum control of their physical and chemical properties. Electron microscopy provides researchers with key insight into a wide variety of material characteristics at the micro- to nano-scale.
          Quality control and failure analysis using electron microscopy
          Quality control and failure analysis
          Quality control and assurance are essential in modern industry. We offer a range of EM and spectroscopy tools for multi-scale and multi-modal analysis of defects, allowing you to make reliable and informed decisions for process control and improvement.
          Samples
          Battery Research
          Battery development is enabled by multi-scale analysis with microCT, SEM and TEM, Raman spectroscopy, XPS, and digital 3D visualization and analysis. Learn how this approach provides the structural and chemical information needed to build better batteries.
          Learn more ›
          Polymers Research
          Polymer microstructure dictates the material’s bulk characteristics and performance. Electron microscopy enables comprehensive microscale analysis of polymer morphology and composition for R&D and quality control applications.
          Learn more ›
          Metals Research and Characterization
          Effective production of metals requires precise control of inclusions and precipitates. Our automated tools can perform a variety of tasks critical for metal analysis including; nanoparticle counting, EDS chemical analysis and TEM sample preparation.
          Learn more ›
          Fibers and Filters
          The diameter, morphology and density of synthetic fibers are key parameters that determine the lifetime and functionality of a filter. Scanning electron microscopy (SEM) is the ideal technique for quickly and easily investigating these features.
          Learn more ›
          Forensics
          Micro-traces of crime scene evidence can be analyzed and compared using electron microscopy as part of a forensic investigation. Compatible samples include glass and paint fragments, tool marks, drugs, explosives, and GSR (gunshot residue).
          Learn more ›
          Geological Research
          Geoscience relies on consistent and accurate multi-scale observation of features within rock samples. SEM-EDS, combined with automation software, enables direct, large-scale analysis of texture and mineral composition for petrology and mineralogy research.
          Learn more ›
          Catalysis Research
          Catalysts are critical for a majority of modern industrial processes. Their efficiency depends on the microscopic composition and morphology of the catalytic particles; EM with EDS is ideally suited for studying these properties.
          Learn more ›
          2D Materials
          Novel materials research is increasingly interested in the structure of low-dimensional materials. Scanning transmission electron microscopy with probe correction and monochromation allows for high-resolution two-dimensional materials imaging.
          Learn more ›
          Automotive Materials Testing
          Every component in a modern vehicle is designed for safety, efficiency, and performance. Detailed characterization of automotive materials with electron microscopy and spectroscopy informs critical process decisions, product improvements, and new materials.
          Learn more ›
          Style Sheet for Komodo Tabs
          Products
          Style Sheet for Instrument Cards Original
          Iliad (S)TEM
          • Advanced integration of EELS and (S)TEM optics
          • Electrostatic beam blanker
          • High energy resolution electron source
          Spectra Ultra
          • New imaging and spectroscopy capabilities on the most beam sensitive materials
          • A leap forward in EDX detection with Ultra-X
          • Column designed to maintain sample integrity.
          Spectra 200
          • High-resolution and contrast imaging for accelerating voltages from 30-200 kV
          • Symmetric S-TWIN/X-TWIN objective lens with wide-gap pole piece design of 5.4 mm
          • Sub-Angstrom STEM imaging resolution from 60 kV-200 kV
          Spectra 300
          • Highest-resolution structural and chemical information at the atomic level
          • Flexible high-tension range from 30-300 kV
          • Three lens condenser system
          Thermo Scientific Talos L120C transmission electron microscope (TEM)
          Talos L120C TEM
          • High versatility and stability
          • 4k x 4k Ceta CMOS camera for speed and large FOV
          • TEM magnification range of 25X to 650kX
          • EDS and STEM options for compositional analyses
          Talos F200X TEM
          • High-resolution, EDS cleanliness, and quality in 2D as well as 3D
          • X-FEG and X-CFEG available for the highest brightness and energy resolution
          • High accuracy and repeatable results with integrated Thermo Scientific Velox Software
          Talos F200i TEM
          • Compact design with X-TWIN objective lens
          • Available with S-FEG, X-FEG, and X-CFEG
          • Flexible and fast EDS options for comprehensive elemental analysis
          Talos F200S TEM
          • Intuitive and easy-to-use automation software
          • Available with Super-X EDS for rapid quantitative chemical analysis
          • High-throughput with simultaneous multi-signal acquisition
          Thermo Scientific Talos F200C transmission electron microscope (TEM)
          Talos F200C TEM
          • High-contrast and high-quality TEM and STEM imaging
          • 4k x 4k Ceta CMOS camera options for large FOV and high read-out speeds
          • Large pole piece gap and multiple in situ options
          Elemental Mapping
          • Fast and reliable information on the distribution of elements within the sample or the selected line
          • Easily exported and reported results
          Style Sheet to change H2 style to p with em-h2-header class
          Contact us
          Style Sheet for Support and Service footer
          Style Sheet for Fonts
          Style Sheet for Cards
          Electron microscopy services for
          the materials science
          To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.