Industrial water analysis: From point of entry to waste water
Maintaining a sustainable water program is essential for all water consuming industries. Our Discrete analyzers facilitate routine testing and monitoring, helping prevent corrosion and scaling, maintain compliance with wastewater permits, and monitor ultrapure water. Trust us to safeguard your processes and optimize performance with precise water quality analysis.
Industrial Water Analysis Infographic
Gallery and Gallery Plus Aqua Master Discrete Analyzers
Gallery and Gallery Plus Aqua Master discrete analyzers streamline water and nutrient analyses. Powered by custom-designed software, these  automated systems perform high-throughput simultaneous multiparameter water and nutrient analyses that meet local regulations.
Learn more
The ECM is an integrated part of the Gallery Aqua Master system and supports automated pH and conductivity measurements. It can measure both pH and conductivity simultaneously alongside the photometric testing. This module can be used for analysis of raw water, ground water, sea water, rainwater, municipal water, drinking water, sewage water, and wastewater. It is not suitable for measurement in pure water matrices such as DI water or steam condensate.
For laboratories that perform routine process water and wastewater analysis, Gallery and Gallery Plus Aqua Master systems with ECM module provide increased throughput while lowering the cost per analysis.
Ready-to-use reagent kits for water analysis
Save time by eliminating reagent preparation with the Thermo Scientific Gallery system reagents for environmental testing, such as water (clean water, wastewater, soil and sludge digests, effluents, and saline) and industrial (process water) analysis. With over 40 ready-to-use Gallery system reagents for environmental analysis, the kits include 1 to 3 reagents in 20 mL barcoded vials, allowing the necessary identification data, such as lot and expiration date, to be read automatically by the discrete analyzer's barcode reader. Automated dilutions make it possible to apply methods for a wide range of analyte concentrations and the test applications are based on recommended standard water and wastewater methods used in most countries. Save time and reduce errors by simplifying your analysis with the Gallery system reagents for environmental analysis.
Gallery and Gallery Plus Aqua Master discrete analyzers are easy to use, automated systems that allow laboratories to simplify their testing with the dual benefits of time and cost savings. All necessary analysis steps are automated, providing true walk-away time for the operator. Parallel determination of several analytes along with pH and conductivity from a single sample as well as the presence of several automated features ensures analytical efficiency.
Video Player is loading.
Current Time 0:00
Duration 2:27
Loaded: 0%
Stream Type LIVE
Remaining Time 2:27
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    List of analytes
    Corrosive anionsCorrosion inhibitorsCorrosion indicatorsBasic water analysis parametersEnvironmental parametersScaling parametersOrganic acidsNutrients
    Fluoride
    Chloride
    Sulfide
    Sulfate
    Sulfite
    Thiosulfate
    Ammonia
    Nitrite
    Phosphate
    Zinc
    Molybdenum
    Total iron
    Chromium (VI)
    Zinc
    Copper
    pH
    Conductivity
    Alkalinity
    Total hardness
    Total Kjedal Nitrogen (TKN)
    Total phosphorus
    Total phenol
    Total-Oxidizable Nitrogen (TON)
    Free and total cyanide
    Sulfide
    Aluminum
    Manganese
    Boron
    Silica
    Calcium
    Magnesium
    Formic acid
    Acetic acid
    Oxalic acid
    Glycolic acid
    Ammonia
    Total phosphorous
    TKN
    TON
    Ortho phosphate
    Environmental & industrial applications compendium
    Discover how Gallery Discrete Analyzers offer simultaneous, multi-parameter nutrient analysis for drinking water, wastewater, sewage water, soil and fertilizer and plant extract samples.
    Download compendium
    Solution for routine and comprehensive water analysis
    The combined power of DISC-IC improves productivity and efficiency in water testing labs, while it consolidates your steam and water chemistry analytical needs.
    Download brochure
    Streamlined water and nutrient analysis
    Learn how Gallery Aqua Master analyzers not only improve the reliability and sensitivity of results compared to manual techniques for water analysis, but also increase laboratory productivity.
    Watch webinar
    Automate your wet chemistry analysis
    This blog series provide educational resource for the latest applications, technical information on automated wet chemical analysis using discrete analyzers.
    Read blog
    Gallery Aqua Master systems brochure
    Combining rugged hardware, dedicated software, and simple operation, Gallery and Gallery Plus Aqua Master discrete analyzers offer high-throughput water and nutrient analysis to meet local regulations.
    Download brochure
    Looking for more automated wet chemical analysis resources?
    Visit our filterable and searchable resources library for product information, application notes, case studies, videos and more about our discrete analyzers.
    Access more resources ›
    *
    *
    *
    *
    *
    *
    *
    *
     Please confirm you would like to receive marketing and promotional email messages about Thermo Fisher Scientific™ products and services.
    This site is protected by reCAPTCHA and the Google Privacy Policy and Terms of Service apply.
    Style Sheet for Global Design System
    Style Sheet for Komodo Tabs
    CMD Wide-format style fixes
    CMD SchemaApp code