Save time by eliminating reagent preparation with the Thermo Scientific Gallery system reagents for environmental testing, such as water (clean water, wastewater, soil and sludge digests, effluents, and saline) and industrial (process water) analysis. With over 40 ready-to-use gallery system reagents for environmental analysis, the kits include 1 to 3 reagents in 20 mL barcoded vials, allowing the necessary identification data, such as lot and expiration date, to be read automatically by the discrete analyzer's barcode reader. Automated dilutions make it possible to apply methods for a wide range of analyte concentrations and the test applications are based on recommended standard water and wastewater methods used in most countries. Save time and reduce errors by simplifying your analysis with the Gallery system reagents for environmental analysis.
Benefits
The unique low volume cuvette design guarantees smallest reagent consumption, 2-240 μL, minimizes reagent waste, and as a result reduces reagent costs. Optimized kit sizes and on-board stability further minimize the amount of waste produced and increase cost efficiency.
Each vessel is bar coded with all the necessary data including reagent identification, volume, lot, and expiration date. Gallery’s built in barcode reader for sample and reagents Barcode reader improves the traceability and continuously monitors the reagent consumptions and provides real-time reagent information
Thermo Scientific Gallery and Gallery Plus discrete analyzers are easy to use, automated systems that allow laboratories to simplify their testing with the dual benefits of time and cost savings. All necessary analysis steps, startup, calibration, sample dilution are automated, providing true walk-away time for the operator.
Thermo Scientific system reagents are ready to go, saving the technician’s time and reducing errors. Ensuring confidence in the quality of results, the methods used for analysis in the Thermo Scientific discrete analyzers are well known enzymatic and colorimetric chemistries optimized to the result levels established in international reference methods, EPA, ASTM, DIN, ISO, OIV, AOAC, and EBC.
Ready to use reagents are easy to work with eliminating extra preparation steps and saving time and money. Gallery discrete analyzers assure fast, reliable and repeatable results by utilizing specific enzyme reactions, and robust robotics to streamline laboratory analysis.
The unique low volume cuvette design guarantees smallest reagent consumption, 2-240 μL, minimizes reagent waste, and as a result reduces reagent costs. Optimized kit sizes and on-board stability further minimize the amount of waste produced and increase cost efficiency.
Each vessel is bar coded with all the necessary data including reagent identification, volume, lot, and expiration date. Gallery’s built in barcode reader for sample and reagents Barcode reader improves the traceability and continuously monitors the reagent consumptions and provides real-time reagent information
Thermo Scientific Gallery and Gallery Plus discrete analyzers are easy to use, automated systems that allow laboratories to simplify their testing with the dual benefits of time and cost savings. All necessary analysis steps, startup, calibration, sample dilution are automated, providing true walk-away time for the operator.
Thermo Scientific system reagents are ready to go, saving the technician’s time and reducing errors. Ensuring confidence in the quality of results, the methods used for analysis in the Thermo Scientific discrete analyzers are well known enzymatic and colorimetric chemistries optimized to the result levels established in international reference methods, EPA, ASTM, DIN, ISO, OIV, AOAC, and EBC.
Ready to use reagents are easy to work with eliminating extra preparation steps and saving time and money. Gallery discrete analyzers assure fast, reliable and repeatable results by utilizing specific enzyme reactions, and robust robotics to streamline laboratory analysis.
Reagents for Food & Beverage Analysis
Gallery System Reagents for food and beverage analysis are ready-to-use, eliminate extra preparation steps, and save valuable laboratory time. Designed for the Gallery and Thermo Scientific Arena discrete analyzer, the volume-optimized system reagents minimize reagent waste, while continuous monitoring of volume, lot, and expiration dates provides real-time reagent information.
Learn more
Thermo Scientific Gallery discrete analyzers together with ready to use system reagents makes the overall discrete analysis simple, accurate and reliable.
  Each vessel is bar coded with all the necessary data including reagent identification, volume, lot, and expiration date.
  Continuous monitoring gives real-time reagent information.
  Volume optimized system kits minimize waste.
  Required reagent volumes can be up to 15 times smaller when compared to manual methods.
  A wide range of calibration standards ensure traceable results.
Ordering information
Catalog numberDescriptionAnalyteUnit sizeInterference filter (nm) main/ side (alternative)Test range
mg/L
Request
a quote
984187TON (nitrate + nitrite) by enzymatic reduction system reagents - R1 (275 tests)Nitrate (TON Enz)Each540/700 (for Gallery) 570 (for Aquakem)0 - 25 (for Gallery) 0 - 50 (for Aquakem)Request
a quote
984350TON (nitrate + nitrite) by vanadium reduction system reagents - R1 (360 tests)Nitrate (TON Vanadium)Pack of 45400.4-50Request
a quote
984351TON (nitrate + nitrite) by vanadium reduction system reagents - R2 (500 tests)Nitrate (TON Vanadium)Pack of 45400.4-50Request
a quote
984357Chromium (VI) system reagents (480 tests)Hexavalent ChromiumPack of 4540/ 8800.1-1.0Request
a quote
984358Magnesium system reagents (350 tests)Magnesium (Mg)Pack of 85100-400Request
a quote
984361Calcium (Ca) system reagents (350 tests)CalciumPack of 36600-1000Request
a quote
984362Ammonia system reagents - R1 (2000 tests)AmmoniaEach6601.0 - 10.0 (for Gallery)Request
a quote
984363Ammonia system reagents - R2 (1300 tests)AmmoniaPack of 46601.0 - 10.0 (for Gallery)Request
a quote
984364Chloride system reagents - R1 (500 tests)ChloridePack of 4480100 - 500 (for Gallery) 100 - 1000 (for Aquakem)Request
a quote
984365Chloride system reagents - R1 (2500 tests)ChloridePack of 20480100 - 500 (for Gallery) 100 - 1000 (for Aquakem)Request
a quote
984366Phosphate system reagents - R1 (2300 tests)PhosphatePack of 48801.0-10.0Request
a quote
984367Phosphate system reagents - R1 (11700 tests)PhosphatePack of 208801.0-10.0Request
a quote
984368Phosphate system reagents - R2 (3000 tests)PhosphatePack of 48801.0-10.0Request
a quote
984369TON (nitrate + nitrite) by hydrazine reduction system reagents - R1 (1300 tests)Nitrate (TON Hyd)Each5400 - 25 (for Gallery) 0 - 50 (for Aquakem)Request
a quote
984370TON (nitrate + nitrite) by hydrazine reduction system reagents - R2 (750 tests)Nitrate (TON Hyd)Pack of 45400 - 25 (for Gallery) 0 - 50 (for Aquakem)Request
a quote
984371Nitrite system reagents (1000 tests)NitritePack of 45400 - 25 (for Gallery) 0 - 50 (for Aquakem)Request
a quote
984372Nitrite system reagents (5000 tests)NitritePack of 205400 - 25 (for Gallery) 0 - 50 (for Aquakem)Request
a quote
984620Total Hardness system reagents - R1 (1100 tests)Total HardnessPack of 4620/ 8800-500Request
a quote
984621Total Hardness system reagents - R2 (975 tests)Total HardnessPack of 4620/ 8800-500Request
a quote
984622Total Hardness system reagents - R3 (4300 tests)Total HardnessPack of 4620/ 8800-500Request
a quote
984623Alkalinity system reagents - R1 (975 tests)AlkalinityPack of 4600/ 8800-400Request
a quote
984624Alkalinity system reagents - R2 (780 tests)AlkalinityPack of 4600/ 8800-400Request
a quote
984625Silica system reagents - R1 (970 tests)SilicaPack of 4700/ 4200-80Request
a quote
984626Silica system reagents - R2 (1950 tests)SilicaPack of 4700/ 4200-80Request
a quote
984627Silica system reagents - R3 (1950 tests)SilicaPack of 4700/ 4200-80Request
a quote
984629Fluoride system reagents - R1 (2200 tests)FluoridePack of 4600/ 8800 - 5.0 (for Gallery) 0 - 2.0 (for Aquakem)Request
a quote
984630Fluoride system reagents - R2 (4800 tests)FluoridePack of 4600/ 8800 - 5.0 (for Gallery) 0 - 2.0 (for Aquakem)Request
a quote
984631Fluoride system reagents - R3 (2200 tests)FluoridePack of 4600/ 8800 - 5.0 (for Gallery) 0 - 2.0 (for Aquakem)Request
a quote
984648Sulphate system reagents (1100 tests)SulphatePack of 4420100-500Request
a quote
984649Sulphate system reagents (5100 tests)SulphatePack of 6420100-500Request
a quote
984652TON (nitrate + nitrite) by hydrazine reduction system reagents - R2 (3400 tests) - for Aquakem onlyNitrate (TON Hyd)Pack of 65400 - 25 (for Gallery) 0 - 50 (for Aquakem)Request
a quote
Discrete Analyzer Market Applications
Simplified wine analysis
Thermo Scientific Gallery discrete analyzers make wine analysis simple, accurate and reliable. Our ready-to-use system reagents are specifically developed for rapid and cost-effective analysis of juice and wine.
High-throughput environmental analysis
Learn how our discrete analyzers help you comply with the U.S. EPA Clean Water Act (CWA) by automating photometric and electrochemical analysis to provide fast, reproducible results.
Resources
Brochure for automated wet chemical analysis
The brochure presents how the Thermo Scientific Gallery discrete analyzers can help growing laboratories perform food and beverage analysis, and environmental water analysis by automating multiparameter wet chemical analysis.
Environmental and industrial analysis webinars
Browse our searchable database of on-demand webinars about discrete industrial analyzers for environmental and industrial analysis to find methods and applications to accelerate the work in your lab.
Automated chloride analysis method for drinking water samples
A novel discrete analyzer-based automated method for drinking water chloride testing for compliance measurements that follows the United States Environmental Protection Agency (U.S. EPA) Safe Drinking Water Act (SDWA).
Search automated wet chemistry analysis resources
TypeTitle
Manuals & protocols Ammonia (DIC)
Manuals & protocols Chloride
Manuals & protocols Phosphate
Manuals & protocols Product Insert – Environmental – Sulphate
Manuals & protocols TON (Total Oxidized Nitrogen) as N and Nitrate by calculation (TON-Nitrite)
Manuals & protocols TON as N, Total Oxidized Nitrogen (Vanadium Chloride Reduction)
Manuals & protocols Total Oxidized Nitrogen (Enzymatic)
Product literature Application Note: Automated Total Oxidized Nitrogen Method Using Vanadium as Reductant with Correlation to Cadmium and Hydrazine Reductant Methods in Sea, Natural, and Waste Waters
Product literature Application Note: Determination of Water Pollutants Using Photometric Analysis
Product literature Application Note: Enzymatic Analysis of Urea from Swimming Pool Waters
Product literature Case Study: Fast and Better Analysis of Complex Samples in a Geochemical Laboratory
Product literature eBook: Fast and Accurate Gallery Discrete Industrial Analyzers – Automated Nutrient Analysis and Water Quality Monitoring
Product literature Specification Sheet: Discrete Industrial Analyzer System Reagents for Water/Environmental Samples
Material safety data sheets (MSDS)984187
Material safety data sheets (MSDS)984187_NADH
Material safety data sheets (MSDS)984350
Material safety data sheets (MSDS)984351
Material safety data sheets (MSDS)984357
Material safety data sheets (MSDS)984361
Material safety data sheets (MSDS)984362
Material safety data sheets (MSDS)984363
Material safety data sheets (MSDS)984364_984365
Material safety data sheets (MSDS)984366_984367
Material safety data sheets (MSDS)984368_R2
Material safety data sheets (MSDS)984369
Contact us
*
*
*
*
*
*
*
*
 Please confirm you would like to receive marketing and promotional email messages about Thermo Fisher Scientific™ products and services.
This site is protected by reCAPTCHA and the Google Privacy Policy and Terms of Service apply.
Style Sheet for Komodo Tabs
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
CMD Wide-format style fixes
CMD SchemaApp code