More Filters
System
Description
Dionex AminoPac PA10 Column
Copy link
Application: Separation of free amino acids and carbohydrates
Eluent: Hydroxide,Acetate
Dionex CarboPac PA300-4μm Column
Copy link
Application: Separation of oligosaccharides,Separation of neutral and charged glycans
Column features: High-pressure IC
Eluent: Hydroxide,Acetate
Dionex CarboPac PA210-Fast-4μm Column
Copy link
Application: Analysis of mono-, di-, tri-, tetra-, and pentasaccharides,Quantification of nonderivatized sugars
Column features: Fast analysis,High-pressure IC
Eluent: Hydroxide
Dionex CarboPac PA200 Column
Copy link
Application: Separation of neutral and charged oligosaccharides,Analysis of carbohydrates released from glycoproteins
Eluent: Hydroxide,Acetate
Application: Determination of oligosaccharides, linear polysaccharides and galactooligosaccharides
Column features: Dual EGC mode
Eluent: Hydroxide and MSA
Dionex CarboPac PA100 Column
Copy link
Application: Separation of oligosaccharides from therapeutic glycoproteins,Separation of neutral and charged oligosaccharides
Eluent: Hydroxide,Acetate
Dionex CarboPac PA20-Fast-4μm Column
Copy link
Application: Analysis of mono- and disaccharides in foods, beverages, glycoprotein therapeutics and mammalian glycoproteins
Column features: Fast analysis,High-pressure IC
Eluent: Hydroxide,Acetate
Application: Analysis of sialic acids
Column features: Fast analysis
Eluent: Hydroxide,Acetate
Dionex CarboPac PA20 Column
Copy link
Application: Analysis of mono- and disaccharides in foods, beverages, glycoprotein therapeutics and mammalian glycoproteins
Eluent: Hydroxide,Acetate
Application: Isocratic and gradient separations of mono- and disaccharides, and sialic acids
Column features: Dual EGC mode
Eluent: Hydroxide and MSA