More Filters
System
Description
Dionex IonPac AS32-Fast-4μm Column
Copy link
Application: Analysis of polythionates, persulfate, polysulfonated aromatics, and aromatic dyes,Analysis of perchlorate in matrices containing high chloride, carbonate and sulfate
Column features: Fast analysis,High-pressure IC
Eluent: Hydroxide
Dionex IonPac AS31 Column
Copy link
Application: Analysis of haloacetic acids, dalapon, and bromate in drinking water prior to MS detection,US EPA 557
Eluent: Hydroxide
Dionex IonPac AS30 Column
Copy link
Application: Analysis of trace oxyhalides and inorganic anions in drinking water preserved with ethylenediamine (EDA)
Eluent: Hydroxide
Dionex IonPac AS28-Fast-4μm Column
Copy link
Application: Analysis of trace inorganic anions and low molecular weight organic acids in high-purity water
Column features: Fast analysis,High-pressure IC
Eluent: Hydroxide
Dionex IonPac AS27 Column
Copy link
Application: Analysis of trace bromate in drinking water preserved with ethylenediamine (EDA)
Eluent: Hydroxide
Dionex IonPac AS25 Column
Copy link
Application: Analysis of multivalent anions and polarizable anions in complex samples
Eluent: Hydroxide
Dionex IonPac AS24 Column
Copy link
Application: Separation of haloacetic acids and bromate in drinking water prior to MS detection,US EPA 557
Eluent: Hydroxide
Dionex IonPac AS21 Column
Copy link
Application: Analysis of trace perchlorate prior to MS detection,US EPA 331.0
Eluent: Hydroxide
Dionex IonPac AS20 Column
Copy link
Application: Analysis of trace perchlorate in drinking water in the presence of high concentrations of chloride, carbonate and sulfate,US EPA 314.1
Eluent: Hydroxide
Dionex IonPac AS19-4μm Column
Copy link
Application: Analysis of inorganic anions, bromate and oxyhalides,Trace bromate and inorganic anions in water,US EPA 300.0 and 300.1
Column features: High-pressure IC
Eluent: Hydroxide
Dionex IonPac AS19 Column
Copy link
Application: Analysis of inorganic anions, bromate and oxyhalides,Trace bromate and inorganic anions in water,US EPA 300.0 and 300.1
Eluent: Hydroxide
Dionex IonPac AS18-Fast-4μm Column
Copy link
Application: Analysis of inorganic anions and organic acids,US EPA 300.0 (A) and 300.1
Column features: Fast analysis,High-pressure IC
Eluent: Hydroxide
Dionex IonPac AS18-Fast Column
Copy link
Application: Analysis of inorganic anions and organic acids,US EPA 300.0 (A) and 300.1
Column features: Fast analysis
Eluent: Hydroxide
Dionex IonPac AS18 Column
Copy link
Application: Analysis of inorganic anions and organic acids,US EPA 300.0 (A)
Eluent: Hydroxide
Dionex IonPac AS17-C Column
Copy link
Application: Analysis of inorganic anions in high-purity water,US EPA 300.0 (A)
Eluent: Hydroxide
Dionex IonPac AS16-4μm Column
Copy link
Application: Analysis of trace perchlorate and polarizable anions,2D-IC analysis of trace perchlorate
Column features: High-pressure IC
Eluent: Hydroxide
Dionex IonPac AS16 Column
Copy link
Application: Analysis of trace perchlorate and polarizable anions,2D-IC analysis of trace perchlorate
Eluent: Hydroxide
Dionex IonPac AS15 Column
Copy link
Application: Analysis of trace inorganic anions and low molecular weight organic acids in high-purity water
Eluent: Hydroxide
Dionex IonPac AS11-HC-4μm Column
Copy link
Application: Separation of inorganic anions and organic acids in high ionic strength samples
Column features: High-pressure IC
Eluent: Hydroxide
Dionex IonPac AS11-HC Column
Copy link
Application: Separation of inorganic anions and organic acids in high ionic strength samples,Separation of mono- and divalent organic acids
Eluent: Hydroxide
Dionex IonPac AS11 Column
Copy link
Application: Analysis of inorganic anions and organic acids
Eluent: Hydroxide
Dionex IonPac AS10 Column
Copy link
Application: Trace-level analysis of inorganic anions and organic acids in high-ionic strength matrices
Eluent: Hydroxide
Dionex IonPac Fast Anion IIIA Column
Copy link
Application: Rapid analysis of phosphoric and citric acids
Column features: Fast analysis
Eluent: Hydroxide