Search Thermo Fisher Scientific
Thermo Scientific flow computers incorporate the latest measurement standards and calculations for hydrocarbon measurement. Use this comparison chart to see which flow computer is right for you.
Still have questions? We're here to help!
AutoXP Gas and Liquid Flow Computer | AutoPILOT PRO Multi-Run Hydrocarbon Flow Computer | AutoEXEC 32-Run Expandable Gas and Liquid Flow Computer | |
| ![]() | ![]() | ![]() |
Upstream | |||
Run Capacity | 1-2 Runs | 1-6 Runs | up to 32 Runs |
Plunger Capability | Yes | Yes | Yes |
API 21.1 Compliant | Yes | Yes | Yes |
Field Mountable | Yes | Yes | No |
Midstream | |||
API 21.1 & 21.2 Compliant | Yes | Yes | Yes |
Expandable IO | No | Yes | Yes |
GC Connectivity | Yes | Yes | Yes |
Downstream | |||
Full Station Control | No | Yes | Yes |
Rack Mount | No | No | Yes |
USM Diagnostics | No | Yes | Yes |
General | |||
Bluetooth | Yes | No | No |
Intrinsically Safe | No | Yes | Yes* |
Proving Loop Control | No | No | Yes |
Custom Reporting | Yes | Yes | Yes |
Flow Calculation Speed | 1x Sec | 1x Sec | 10x Sec |
*The AutoEXEC provides intrinsically safe connections for the AutoMITTER PRO when the AutoMITTER PRO is connected via the AutoMITTER safety interface board. |