Refinery Water Process Analysis
Protect your refinery equipment and the environment
Industrial water processes keep the refinery operational. They include the use, treatment, and analysis of scrubber solutions and boiler/cooling water. Scrubber solutions include aqueous alkanolamine solutions used to neutralize acidic gases (e.g., carbon dioxide and hydrogen sulfide) and remove them from the process stream in order to prevent equipment corrosion and release into the atmosphere. In a similar fashion, boiler/cooling water are treated with amines to neutralize any acidic buildup. 
Let one of our experts help you tailor a solution specific to your application and workflow needs.
Contact us
Petroleum and natural gas testing workflows
Select from a choice of available products according to the process, matrix, application and analyte of interest. 
Process Monitoring
Wastewater Monitoring
Industrial Water
Amines, Anions, Organic acids
IC, GC
Cations, Anions, Ammonium,
Alkai & Alkaline earth metals
IC, ICP-OES (cations only)
Metals (e.g. Zn, Cd)
ICP-OES
Organic acids
IC, GC
 BTEX*, PCB**
ASE, GC
S
ICP-OES
Scrubber Solutions
Organic acids, Sulfur species, Amines,
Alkanolamines, Polyphosphates, Polyphosphonates
IC
Cations, Anions
IC, ICP-OES (cations only)
*benzene, toluene, ethyl benzene, xylenes
**polychorlinatedbiphenyl
Solutions to resolve analytical challenges in industrial and waste water processing infographic
Reliable water analysis is critical to eliminating corrosion and scaling that can damage valuable industrial components and to ensure regulatory compliance. Download the infographic to learn about our solutions to the top three analytical challenges in industrial water and waste water processing.
Learn more