Web Gauging Platforms for Thickness and Basis Weight Measurement
Leading through innovation
Manufacturing process control and repeatability is key to ensuring your products have superior quality, meet industry standards and are attractive to consumers. Thermo Scientific web gauging thickness and basis weight measurement platforms provide efficient production of uniform, reliable, functional products. Our technologies include infrared, x-ray, nuclear and optical measurements, plus an advanced controls portfolio and an intuitive operator interface, to give a unique insight into your process. Everything you need to manage your production.
Web gauging platforms
LInspector Measurement and Control System
LInspector Measurement and Control System
The Thermo Scientific LInspector measurement and control system is our newest solution and is specifically designed for lithium-ion battery applications. It addresses the need for accurate electrode coating weight measurement, multilayer thickness measurement of separator films and electrode calendering thickness measurement. The system consists of one or multiple scanners, sensors and a control system tailored to the application and helps manufacturers improve the safety and efficiency of lithium-ion batteries and to deliver high quality, reliable products.
21PlusHD measurement and control system
21PlusHD
The Thermo Scientific 21PlusHD measurement and control system for continuous web processing supports a comprehensive portfolio that offers over forty specialized application packages. The system consists of one or multiple scanners, sensors and a control system tailored to your application, ensuring a fast return on investment, through improved product quality process efficiency and raw materials savings.
IPlus! measurement and control system
IPlus!
The Thermo Scientific IPlus! measurement and control system is our reliable and robust value based system. Its advanced sensor technology and powerful software, packaged in an intuitive easy-to-use system provides users with high performance and a low cost of ownership. The system provides data to improve product quality and uniformity, process efficiency and raw material savings.
Video: Non-contact basis weight and thickness measurement gauges
Video Player is loading.
Current Time 0:00
Duration 3:40
Loaded: 0%
Stream Type LIVE
Remaining Time 3:40
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Learn how Thermo Scientific web thickness and basis weight systems ensure the performance of flat-sheet materials in the plastics, lithium-ion battery, and textiles industries.
    What You Need to Know About Web Gauging Systems eBook
    Optimize your process with the right system
    Do you know how to choose a gauging platform, sensor, and scanner that will work best for your application? Download the free eBook, What You Need to Know About Web Gauging Systems.
    Download eBook