Unstoppable confidence in quantitative performance
Delivering attogram-level sensitivity and best-in-class selected reaction monitoring (SRM) performance, the TSQ 9610 GC‑MS/MS system consistently meets required detection limits as you run samples. Choose the rugged Thermo Scientific ExtractaBrite ion source or the ultra-sensitive, ultra-robust Thermo Scientific Advanced Electron Ionization (AEI) ion source, depending on your sensitivity needs.
The XLXR electron multiplier detector which extends dynamic range twofold compared to the previous design. Extended dynamic range capability enables labs to combine methods, including calibration curve development, allowing analysis of low- and high-concentration compounds in a single run.
Unstoppable confidence in uptime for full-time productivity
The TSQ 9610 GC-MS/MS system is specifically designed for complex sample matrices in high-throughput applications. Using NeverVent technology with the Vacuum Probe Interlock (VPI) and V-Lock source plug, planned maintenance such as changing column and ion source cleaning can be performed without venting the mass spectrometer, increasing uptime. NeverVent technology is available on both the Thermo Scientific ExtractaBrite and the Thermo Scientific NeverVent Advanced Electron Ionization (NV-AEI) sources.
Unstoppable confidence in your investment
Rapid instrument deployment reduces time to results and revenue generation. That’s why the TSQ 9610 Triple Quadrupole GC-MS/MS System includes a comprehensive set of automated tools to provide users with a seamless experience when transitioning from other platforms, developing methods, and analyzing samples. This built-in intelligence reduces operator training needs and time to full productivity for rapid ROI.
Unstoppable confidence in sample throughput
Maximizing sample throughput is essential to producing results on time. The TSQ 9610 Triple Quadrupole GC-MS/MS System incorporates user-centric innovations that extend service intervals, reduce maintenance, and allow methods consolidation to boost sample throughput while maintaining certainty in quality of results. 
The right choice for any workflow
Intuitive Thermo Scientific Chromeleon Chromatography Data System (CDS) supports quantitative workflows. To jump-start your method development, download ready-to-use Thermo Scientific eWorkflows from the Thermo Scientific Appslab Library of Analytical Applications .
Make targeted and untargeted analyses in food, environmental and forensic toxicology applications simple and fast with Thermo Scientific TraceFinder software. For the analysis of dioxins in food, a full eWorkflow is available for easy implementation in any lab.
Resolve issues before they become problems with SmartStatus instrument monitoring
Monitor key aspects of your GC-MS system in detail. The fully customizable Thermo Scientific SmartStatus instrument monitoring within Chromeleon CDS software enables you to quickly see when instruments are ready to run samples, identify exactly when consumable replacement is required or system maintenance is needed.
Watch video overviews
Data-driven GC/GC-MS instrument monitoring with SmartStatus intelligent software
The perfect fit
Build your next GC-MS system and receive a quote
Build my GC-MS
The perfect fit
TSQ 9610 Triple Quadrupole GC-MS/MS
For high-throughput analytical laboratories, the TSQ 9610 GC-MS/MS system provides sensitive, specific quantitation of target compounds.
Learn more
Partnering in sustainability – ACT label 
Reduce your lab’s environmental impact with the TSQ 9610 Triple Quadrupole GC-MS, recipient of the ACT label from My Green Lab. The ACT label, the standard for determining the environmental impact of laboratory products, helps labs make informed, sustainable purchasing decisions. 
Download ACT label
Meeting the New EC Regulations for Dioxin Detection
and Confirmation Using GC-MS/MS
Esteban Abad Holgado, Scientist, Head of the Dioxins Laboratory, Consejo Superior de Investigaciones Cientificas (IDAEA-CSIC), Spain
Cristian Cojocariu, Senior GC Triple Quadrupole Mass Spectrometry Specialist, Thermo Fisher Scientific
Watch on-demand 
Making the GC-MS Triple Transition with Ease
Paul Silcock, Marketing Manager, Triple Quadrupole GC-MS, Thermo Fisher Scientific
Cristian Cojocariu, Senior Applications Specialist, Triple Quadrupole GC-MS, Thermo Fisher Scientific
GC-MS/MS Based Multi-Residue Determination of Pesticides
Zareen Khan, Research Associate, ICAR-National Research Centre for Grapes, National Referral Laboratory, Pune, India
Dr. Shidhar Jayaram Gawade, Application Manager, GC/GC-MS, Thermo Fisher Scientific, India Pvt, Ltd
Watch on-demand
Validation of a Routine Multi-Residue Pesticide Method Using QuEChERS and GC-MS/MS Workflow Solutions
David Steiniger, Senior Application Scientist, Thermo Fisher Scientific
Laszlo Hollosi, Food Safety Application Chemist, Thermo Fisher Scientific, GmbH}
Watch on-demand
Meeting the New EC Regulations for Dioxin Detection
and Confirmation Using GC-MS/MS
Esteban Abad Holgado, Scientist, Head of the Dioxins Laboratory, Consejo Superior de Investigaciones Cientificas (IDAEA-CSIC), Spain
Cristian Cojocariu, Senior GC Triple Quadrupole Mass Spectrometry Specialist, Thermo Fisher Scientific
Watch on-demand 
Making the GC-MS Triple Transition with Ease
Paul Silcock, Marketing Manager, Triple Quadrupole GC-MS, Thermo Fisher Scientific
Cristian Cojocariu, Senior Applications Specialist, Triple Quadrupole GC-MS, Thermo Fisher Scientific
GC-MS/MS Based Multi-Residue Determination of Pesticides
Zareen Khan, Research Associate, ICAR-National Research Centre for Grapes, National Referral Laboratory, Pune, India
Dr. Shidhar Jayaram Gawade, Application Manager, GC/GC-MS, Thermo Fisher Scientific, India Pvt, Ltd
Watch on-demand
Validation of a Routine Multi-Residue Pesticide Method Using QuEChERS and GC-MS/MS Workflow Solutions
David Steiniger, Senior Application Scientist, Thermo Fisher Scientific
Laszlo Hollosi, Food Safety Application Chemist, Thermo Fisher Scientific, GmbH}
Watch on-demand
NOW Foods: Reducing helium gas consumption with HeSaver-H2Safer technology
NOW Foods is a high-throughput laboratory for food testing. Helium shortage is impacting their costs and generates concerns for delayed delivery
Additional resources
Thermo Scientific EXTREVA ASE Accelerated Solvent Extractor
Our new EXTREVA ASE Accelerated Solvent Extractor offers walkaway automation of your sample preparation workflow, from sample extraction to in-cell cleanup to evaporation. It’s the perfect partner for your GC-MS analyses.
Learn more ›
Interested in the analysis of nitrosamines in pharmaceutics?
Download the latest applications notes, register for the latest webinars, and learn more about the analysis of nitrosamines and their precursors.
Learn more
* Required field
*
*
*
*
*
*
*
*
 Please confirm you would like to receive marketing and promotional email messages about Thermo Fisher Scientific products and services.
This site is protected by reCAPTCHA and the Google Privacy Policy and Terms of Service apply.
CMD Wide-format style fixes
Style Sheet for Global Design System
Style Sheet for Komodo Tabs