IC-MS elucidates highly polar compounds
Ion chromatography (IC), when coupled to mass spectrometry (MS), offers an alternative to the analysis of ionic or highly polar compounds using LC-MS.  
Ionic properties
IC-MS has a broader range than HPLC-HILIC; it is also more robust and reproducible than HILIC.
Separation and detection of 21 polar metabolites by capillary IC-HRAM analysis
The high resolution that is inherent with IC has led to the differentiation of many isobaric and isomeric polar metabolites via IC-MS, including sugar phosphate isomers.
Metabolite IC Separations Graph
IC-MS metabolomics
IC can be easily integrated with mass spectrometry because analytes are often provided in the ionic form needed for MS detection. By adding suppressor technology, standard IC eluents and methods can be used. Also, the suppressed background is composed of pure deionized water, allowing easy and advantageous switching between anion and cation feed into the mass spectrometer.
Just as with LC-MS, electrospray ionization (ESI) and atmospheric pressure chemical ionization (APCI) are the two ionization techniques most commonly used with IC-MS. ESI is ideal for semipolar and polar compounds, whereas APCI is more suitable for neutral or less polar compounds. With IC-MS, ESI is generally preferred and used.
In both ESI and APCI, anions are formed by a loss of H+, while cations are formed by a gain of H+, Na+, or some other ion like NH4+ or K+. Formed ions do not undergo fragmentation, such as occurs in GC-MS with electron impact (EI) ionization. In IC, ion suppression is reduced due to the desalting step prior to MS, thus enabling more compounds to be detected and accurately quantified.
IC-MS may be used in both untargeted metabolomics and in targeted metabolomics workflows.
Metabolite identification
IC-MS workflows are similar to those of LC-MS and, unlike GC-MS workflows, offer no spectral libraries for compound identification. However, because molecular ions are usually present in IC-MS, they can be used as part of the compound identification strategy. To begin with, ions can be used to search a database of metabolites such as the METLIN database. With recent advances in HRAM MS systems, it is now possible to calculate the empirical formula of a target compound from the molecular ion.
To better elucidate unknown metabolites, the molecular ion can be fragmented by MS/MS, and MS/MS spectral libraries can be compared and matched. Quite often, this does not lead to a confident identification, and further MS (MSn) analysis is needed. Following MSn, manual de novo interpretation/structural elucidation can be performed as part of the identification process.
Related resources
Orthogonal separation techniques
LC-MS Metabolomics
LC-MS offers the broadest coverage of compounds due to its ability to work with different column chemistries such as reversed phase and hydrophobic interaction liquid chromatography. Common LC-MS compounds include lipids, polyamines, alcohols etc.
Featured videos
Share
Style Sheet for Global Design System
CMD SchemaApp code