Peptone performance to match your vaccine production
With over 100 years of culture media manufacturing experience, from raw materials to final product and powered by our peptones, we offer the expertise, quality assurance and scale to ensure successful microbial vaccine production.
Our Thermo Scientific Oxoid peptones are available in both animal-origin and animal-origin-free options, offering a range of AN/TN ratios to suit the specific needs of your process.
Talk to our team of experts
Request a no-obligation consultation on choosing the right peptones for veterinary vaccine fermentation.
Contact us box
Supports quality media design
  • Balanced nutrient composition
  • Batch-to-batch consistency, available in sizes from 500g up to 25kg and in single batch sizes up to 2.5T
  • Highly soluble
  • BSE and mycoplasma free
Multi-method suitability
  • Batch culture
  • Fed-batch culture
  • Continuous culture
  • Cell culture
Animal origin media
Our animal origin (AO) media are available in beef, pork and casein formulations, and sourced from OIE* designated BSE Negligible countries. 
Featured products
Proteose Peptone
Bacteriological Peptone
Animal origin free media
We also offer animal origin free (AOF) media in soy and yeast formulations, to eliminate the risk of BSE contamination. 
Featured products
Vegetable Peptone No. 1
Vegetable Peptone Broth
GMO-free Soya Peptone
Animal health and vet vaccine trends
The landscape of animal health is undergoing significant changes, especially in the context of bacterial infections and vaccine development. Take a look at our latest article Animal Health Trends & Vet Vaccines in 2025 to explore key trends in animal health, focusing on bacterial infections, the benefits of vaccination versus treatment, the role of peptones in vaccine fermentation, and how to evaluate your peptone mix for optimal efficacy.
Boost consistency and yields in your veterinary vaccine production
Peptones for veterinary vaccine fermentation
Optimize your peptone mix, elevate your vaccine development. Your peptone mix could be the key to unlocking higher vaccine yields and ensuring batch consistency. Schedule a consultation with our experts.
Interested in learning more about selecting the right peptones for your screening processes? 
*Sourced from Office International des Epizooties (World Organisation for Animal Health)-designated BSE negligible countries 
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
1x1 image pixel for data collection