Peptide Mapping
Biopharmaceutical peptide mapping
Peptide mapping is a critical workflow in biotherapeutic protein characterization and is essential for elucidating the primary amino acid structure of proteins. For recombinant protein pharmaceuticals, such as monoclonal antibodies (mAbs) and antibody-drug conjugates (ADCs), peptide mapping is used for proof of identity, primary structural characterization and quality assurance/quality control (QA/QC). Global regulatory agencies, including US Food and Drug Administration (US FDA) and European Medicines Agency (EMA), look to harmonized guidelines from the International Council for Harmonisation (ICH). ICH Q6B  covers the test procedures and acceptance criteria for biologic drug products, and specifies the use of peptide mapping as a critical quality test procedure for drug characterization used to confirm desired product structure for lot release purposes.
Comprehensive peptide maps, simplified
In order to generate a peptide map, the therapeutic protein must first be digested into its constituent peptides via a chemical or enzymatic reaction. Robust separation and identification of the resultant peptides then provides insight into a protein’s full sequence information; displaying each amino acid component and the surrounding amino acid microenvironment, including disulfide linkage information. Structural characterization at this level highlights post translational modifications (PTMs) such as site-specific glycosylation, amino acid substitutions (sequence variants) and/or truncations which may result from erroneous transcription of complementary DNA. Within a bioproduction environment, peptide mapping is necessary for manufacturing process monitoring and QC. It facilitates product comparability testing, which is necessary to identify any product-related impurities, such as deamidation and/or oxidation following any formulation, manufacturing process or storage change.
Due to its complexity and inherent variability, peptide mapping is generally performed in a comparative manner; for example, biosimilars would be compared to a reference or control substance, such as the innovator biologic, in a side-by-side experiment. An in-depth analysis is then required to identify minor and even isobaric differences in protein primary structure. The modern biopharmaceutical and protein research laboratory is tasked with providing high quality analytical results, often in high-throughput, regulated environments. Some technologies currently employed for biopharmaceutical peptide mapping are subject to high levels of irreproducibility, poor sensitivity, and high levels of time-consuming manual work—with protracted methodologies that are not amenable to automation and often require 24 hours to achieve full protein digestion.
This variability impacts data confidence. Moreover, it increases potential for introduction of non-product related artifacts during manual sample handling.
The most recent advances in protein sample preparation chemistries, ultra-high performance liquid chromatography (UHPLC), mass spectrometry (MS) hardware and intuitive software facilitate the generation of comprehensive, confident peptide maps.
Peptide mapping products and workflows ›
Featured webinar series: biopharmaceutical peptide mapping
Take the Smart Route to Protein Digestion: How to Increase Reproducibility Whilst Reducing Prep Time
Learn how to increase confidence in biopharma peptide mapping. In this series, four new technologies will be demonstrated that dramatically improve upon established mapping workflows.
Video Player is loading.
Current Time 0:00
Duration 43:36
Loaded: 0.38%
Stream Type LIVE
Remaining Time 43:36
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
      1. Now Playing
        Up NextTake the Smart Route to Protein Digestion: How to Increase Reproducibility Whilst Reducing Prep Time
        Learn how to increase confidence in biopharma peptide mapping. In this series, four new technologies will be demonstrated that dramatically improve upon established mapping workflows.
      2. Now Playing
        Up NextPeptide Separations with Pinpoint Precision
        Learn how to increase confidence in biopharma peptide mapping. In this series, four new technologies will be demonstrated that dramatically improve upon established mapping workflows.
      3. Now Playing
        Up NextDon't Miss a Thing on Your Peptide Mapping Journey
        Learn how to increase confidence in biopharma peptide mapping. In this series, four new technologies will be demonstrated that dramatically improve upon established mapping workflows.
      4. Now Playing
        Up NextEasily Navigate Biotherapeutic Modifications
        Learn how to increase confidence in biopharma peptide mapping. In this series, four new technologies will be demonstrated that dramatically improve upon established mapping workflows.
    Featured peptide mapping learning content
    Application note
    Combining the Thermo Scientific™ SMART Digest™ magnetic kits with the Thermo Scientific™ KingFisher™ Duo Prime purification system provides an automated approach to protein digest method optimization.
     Download the brochure
    Application note
    Combining the Thermo Scientific™ SMART Digest™ magnetic kits with the Thermo Scientific™ KingFisher™ Duo Prime purification system provides an automated approach to protein digest method optimization.
     Download the application note
    Application note
    Two classic in-solution digestion approaches were compared to the recently developed SMART Digest kit method to quantify the extent of post-translational and chemical modifications of a therapeutic recombinant mAb, rituximab.
    Peptide mapping videos
    BioPharMoore Episode 2: SMARTening up on Protein Digestion
    Join Dr Rowan Moore as she investigates alternatives to traditional in-solution protein digestion protocols for peptide mapping of biopharmaceuticals.
    Video Player is loading.
    Current Time 0:00
    Duration 5:07
    Loaded: 3.20%
    Stream Type LIVE
    Remaining Time 5:07
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
        1. Now Playing
          Up NextBioPharMoore Episode 2: SMARTening up on Protein Digestion
          Join Dr Rowan Moore as she investigates alternatives to traditional in-solution protein digestion protocols for peptide mapping of biopharmaceuticals.
        2. Now Playing
          Up NextA Journey in Biotherapeutic Peptide Mapping with Alexion Pharmaceutical
          Learn about how the latest chromatography, mass spectrometry and software are accelerating confident decision making at Alexion Pharmaceuticals, Inc.
        3. Now Playing
          Up NextHigh throughput Peptide Mapping by UHPLC-MS
          Stephane Houel, Biopharma Product Marketing Specialist, describes a high throughput peptide mapping workflow using the Thermo Scientific Vanquish UHPLC system and Thermo Scientific Q Exactive HF mass spectrometer.
        4. Now Playing
          Up NextGet Smart with Protein Digestion
          Optimize your protein digestion protocol; discover the Thermo Scientific SMART Digest Kits for fast, simple and reproducible peptide mapping.
        5. Now Playing
          Up NextThermo Scientific SMART Digest Kits
          Get faster, easier and more reproducible results from your samples in just three easy steps.
        6. Now Playing
          Up NextHigh-precision, automated peptide mapping of proteins
          NIBRT Application note overview detailing development of a robust and reproducible, high-precision, automated, peptide mapping workflow for LC and LC-MS
      Search all peptide mapping information resources
      TypeTitle
      AppsLabAppsLab Peptide Mapping eWorkflows
      Application NotePeptide Mapping of Challenging Monoclonal Antibodies
      Application BriefHigh Resolution, High-Precision Peptide Mapping of a Monoclonal Antibody Digest
      Application NoteComparison of Alternative Approaches to Trypsin Protein Digestion for Reproducible and Efficient Peptide Mapping Analysis of Monoclonal Antibodies
      Application NoteHigh-throughput Peptide Mapping of Trastuzumab using a Tandem LC-MS Workflow
      Application NoteAutomated Biopharmaceutical Protein Digestion Optimization for Peptide Mapping Method Development
      Application NoteHigh-precision, automated peptide mapping of proteins
      Application NoteFast Digestion Method Optimization Using Innovative Trypsin Technology
      Application NoteHigh-Throughput Peptide Mapping with the Vanquish UHPLC System and the Q Exactive HF Mass Spectrometer
      Application NoteImpact on Tryptic Digestion when Detergents and Chaotropes are Essential: A Case Study with Ribonuclease A
      Application NoteImprovement in Speed and Reproducibility of Protein Digestion and Peptide Quantitation, Utilizing Novel Sample Preparation Technology in a Full Solution Workflow
      Application NoteIncreased Efficiency of Biomolecule Identification by Optimization of Trypsin Digestion Buffers
      Application NoteIncreased Long-term Stability of Peptide Mapping Using the Vanquish UHPLC System
      Application NoteLC-UV-MS Peptide Mapping Development for Easy Transfer to LC-UV QA/QC
      Application NoteIncreasing Sensitivity in HCP Analysis: from Simplified Sample Preparation to Reliable Results
      Application NoteReliable Results in Peptide Mapping Using the Vanquish Flex UHPLC System
      Application NoteSelecting Buffers to Remove Uncertainty in Tryptic Digestion
      Application NoteHILIC – an alternative separation technique for glycopeptides
      Application NoteInvestigating Process-Related Post-Translational Modifications in NISTmAb RM 8671 Using High-Throughput Peptide Mapping Analysis
      ArticleDevelopments in Peptide Mapping Technology for the Biopharmaceutical Industry
      ArticleColumn Temperature Control in Peptide Mapping. Maximizing Peak Capacity by Keeping Your Column Hot the Right Way.
      ArticleHigh-Throughput, High-Resolution Peptide Maps
      BlogAdmiring the Architecture of the Building Blocks of Life
      BlogAhoy! Do I Need Satellite Navigation for my Peptide Map?
      BlogFinding HOPE Using Peptide Mapping?
      BlogNot All Who Wander Are Lost
      BlogWhat’s faster? Digesting a protein, or reading this blog?
      BlogBioPharMoore: Unravelling Biopharmaceutical Protein Digestion
      BrochurePeptide Mapping Workflows: Upgrade Your Maps
      BrochureVanquish Flex UHPLC System: Built for BioPharma
      BrochureBioPharma Finder Software - Brochure
      BrochureQ Exactive BioPharma
      eBookPutting Peptides on the Map
      eBookSimpler, Faster and More Reproducible Protein Digestion
      Online ToolSMART Digest Kits - Digest Time Calculator
      PosterDifferentiate Minor Difference of Protein Structure in Biosimilar and Reference Products Using High-Resolution Orbitrap LC-MS/MS
      Poster NoteCharacterization of Monoclonal Antibodies and ADCs using a Benchtop Orbitrap Mass Spectrometer
      Poster NoteComprehensive Sequence and Post-translational Modifications Analysis of Monoclonal Antibody by Flash Digest and LC-High Resolution MS
      Poster NoteDegradation Profiling of a Monoclonal Antibody Using Multiple Fragmentation Techniques and a Novel Peptide Mapping Software
      Poster NoteDifferentiate Minor Difference of Protein Structure in Biosimilar and Reference Products Using High-Resolution Orbitrap LC-MS/MS
      Poster NoteDisulfide Bond Analysis on Orbitrap Mass Spectrometry
      Poster NoteRapid Peptide Mapping via Automated Integration of On-line Digestion, Separation and Mass Spectrometry for the Analysis of Therapeutic Proteins
      Poster NoteComplete Characterization of a Cysteine linked Antibody-Drug Conjugate Performed on a Hybrid Quadrupole-Orbitrap Mass Spectrometer with High Mass Range
      Poster NoteOrbitrap Based Mass Spectrometric Characterization of Antibody Drug Conjugates Engineered through Antibody Glycans
      Poster NoteComplete Characterization of a Cysteinelinked Antibody-Drug Conjugate Performed on a Hybrid Quadrupole-Orbitrap Mass Spectrometer with High Mass Range
      Application NoteRobust and reproducible peptide mapping and intact mass analysis workflows on a single instrument platform
      Poster NoteCharacterization of Monoclonal Antibodies and ADCs using a Benchtop Orbitrap Mass Spectrometer
      Poster NoteFull Characterization of Heterogeneous Antibody Samples Under Denaturing and Native Conditions on a Hybrid Quadrupole-Orbitrap Mass Spectrometer
      Poster VideoConvenient High pH Peptide Fractionation
      Poster VideoHigh throughput Peptide Mapping by UHPLC-MS
      Poster VideoImproved Protein Identification using SubCellular Localization
      Poster VideoOrbitrap Based Mass Spectrometric Antibody Drug Conjugate Characterization
      PresentationNew, rapid and reproducible peptide mapping workflows
      Product SpotlightSMART Digest Kit Q&A
      Product SpotlightSMART Digest Soluble Trypsin Kit Smarter protein digestion
      Technical NoteHigh Resolution Peptide Mapping for Biopharmaceutical Analysis
      Video
      Video
      Video
      Video
      Vlog
      Web PageBuilt For Biopharma Hypersite - Peptide Mapping
      Web PagePeptide Mapping Webpage
      WebinarImproved Characterization of Biotherapeutics with Better Peptide Mapping
      Webinar
      Web PageBioPharma Finder Software - Mass Spectrometry - Orbitrap
      Web PageBioPharma Finder Software - Product page