Simple, rapid, accurate quantification and qualification of DNA, RNA, and protein
All Thermo Scientific NanoDrop microvolume spectrophotometers include our innovative technology which allows you to quantify DNA, RNA, and protein from just 1–2 µL of sample. With no need for cuvettes or capillaries, results are delivered in seconds, streamlining your workflow and enhancing your research capabilities.
The NanoDrop Ultra offers exceptional accuracy for advanced applications, while the NanoDrop Eight maximizes throughput by measuring eight samples simultaneously. The NanoDrop Lite Plus provides a compact, user-friendly solution for routine measurements. Trusted by researchers worldwide and cited in over 55,000 scientific publications, NanoDrop spectrophotometers are indispensable tools in any lab, ensuring reliable and rapid results every time.
Gain the most comprehensive information about the concentration and quality of your DNA or RNA samples with the innovative NanoDrop Ultra FL and NanoDrop UltraC FL Spectrophotometers, which uniquely combine microvolume UV-Vis absorbance and fluorescence measurements into a single, versatile platform.
NanoDrop Microvolume Spectrophotometer and Fluorometer capabilities
Advanced sample intelligence
With full-spectrum analysis, Acclaro Sample Intelligence software uses advanced chemometric algorithms to identify and correct for contaminants and differentiate DNA from RNA.
Regulatory compliance support
Optional security tools for selected NanoDrop microvolume spectrophotometers help your lab stay compliant with federal data regulations.
Keep your instrument up to date
Download the latest software update to take advantage of the full functionality of your NanoDrop instrument.
Protein quantification
NanoDrop spectrophotometers support protein sample quantification for direct A280 and A205 measurements as well as colorimetric assays such as Bradford, BCA, Lowry, and Pierce 660.
RT-qPCR workflows
Nucleic acid quantification is critical in both steps of the RT-qPCR process. Acclaro software identifies contaminants and provides corrected nucleic acid concentrations to help ensure qPCR success.
For Research Use Only. Not for use in diagnostic procedures.