Meet your analytical challenges with our complete line of ultraviolet-visible (UV-Vis) Spectrophotometers
Our proven designs and user-friendly software help you quantify, assess purity, and more. Rely on industry-tested solutions for the education, pharmaceutical, chemical, beverage, and basic research communities.
UV-Vis Spectrophotometer features
Experience accuracy, ease-of-use, and reliability in an affordable package. From spectrophotometers for life scientists, to routine QA/QC methods, to sampling accessories for high-end materials research, we have the ideal UV-Vis spectrophotometer for your laboratory or classroom. Features common to Thermo Scientific SPECTRONIC 200, Evolution, and GENESYS spectrophotometers:
Fast, accurate
Go from samples to answers quickly and easily. Thermo Scientific UV-Vis and Vis instruments are designed to streamline measurements with consistent, high-quality results, time after time. Exchange samples with ease in large, accessible sample compartments.
Our instruments and software enable fast, accurate results
Reliable
Built on the foundation of over 60 years of proven spectroscopy experience, the next-generation Thermo Scientific UV-Vis and Vis spectrophotometers combine exceptional performance with contemporary technology and design.
Scientists using a GENESYS spectrophotometer in a laboratory
Intuitive
Each instrument features easy-to-use, easy-to-learn onboard software or full-featured control and analysis software that enables you to start taking measurements immediately. 
Insight Pro Software screens
Flexible
Choose what’s right for you—span just the visible colorimetric portion (340-780 nm) or across the full UV-Vis spectrum (190-1100 nm). Select among single beam, dual beam, or double beam optics, fixed or variable pathlength, and basic or advanced capabilities. Then expand its versatility with all the accessories you need.
Scientist using an Evolution spectrophotometer in a laboratory
21 CFR Part 11 compliance software
Evolution UV-Vis Spectrophotometers meet all USP and EP performance characteristics and enable 21 CFR Part 11 compliance. Thermo Scientific Insight Pro Security Software helps your lab stay compliant. It includes all the benefits of Insight Pro Software with added security for ensuring the integrity of scientific data required by pharmaceutical, biotech companies, and academics working on FDA-regulated studies.
Insight Pro Security Software
Streamline your basic, complex, and advanced routines and workflows. Validate according to current USP, EP, or JP standards
Meet 21 CFR Part 11 compliance: 
  • Control user account access, apply digital signatures, implement controls, view electronic audit trails for reliable electronic documentation
  • Database storage ensures electronic records are properly maintained for FDA submissions
Validate your system—Insight Pro Software includes methods to run IQ/OQ verification testing:
  • Evaluate instrument performance within manufacturer’s specifications
  • Meet current pharmacopoeia requirements
  • For general instrument qualification, Qualification Filter Kit for UV-Vis contains some of the standards used for pharmacopeia PV testing
  • USP and EP standard sets available to meet all pharmacopoeia requirements for qualification in the UV as well as visible regions