SPECTRONIC 200 Spectrophotometer for UV-Vis measurements
Support your teaching goals. The Thermo Scientific SPECTRONIC 200 Spectrophotometer is the standard for routine measurements in instructional and analytical laboratories. Designed for simplicity and speed, the SPECTRONIC 200 is a rugged, simple-to-use spectrophotometer that provides quick, easy measurements and years of trouble-free performance. On-board software, controlled from an intuitive navigation pad using a full-color screen, delivers capabilities beyond expectations.
Educational spectrophotometer
With multiple sample compartments, the SPECTRONIC 200 Educator model* helps get the most out of anyone’s teaching budget.
The SPECTRONIC 200 Educator model* provides the greatest value for teaching labs, with much of the same hardware and software features as the standard model at a more affordable price. What’s different? The Educator model does not include the programable analyzer, offers local language support only in English, Spanish, and Thai, and truncates data to two decimal places rather than three.
* Note that the SPECTRONIC 200 Educator is available to distributors in the United States and Canada only.
SPECTRONIC 200 Spectrophotometer features
  • Smart technology – live display and onboard software to program standard methods enables total control without a computer
  • Simple, rugged, and fast – easy to set up, use, maintain, and clean
  • Bright, color LCD – adjusts to the perfect angle for any technician to use the navigation pad and intuitive menus
SPECTRONIC 200 Spectrophotometer specifications
Optical Design
Single Beam
Spectral Bandwidth
≤4 nm
Light Source
Tungsten-halogen
Detector
2048 element CCD
Wavelength
Range
340 nm to 1000 nm
Accuracy
±2 nm
Repeatability
±1 nm
Data interval
1 nm
Photometric
Range
-0.3 A to 2.5 A
Readout
ABS, %T, Concentration1
Accuracy
±0.01 A at 0.3 A
±0.05 A at 1.0 A
(SPECTRONIC standard filters measured at 590 nm)
Repeatability
±0.3 %T at 50 %T
Stray Light
<0.2 %T (with SPECTRONIC standard SRM 400 filter)
Display
Variable angle 320 × 240 pixel color graphical LCD 7 × 5 cm, 8.6 cm diagonal (2.75” × 2”, 3.4” diagonal)
Keypad
Sealed tactile rubber
Standard Features
Sample compartment
Lifts out for cleaning
Cuvette racks and compartments
2 included with dedicated compartments
Included cuvettes
1 cm plastic (quantity 10)
Standard Interfaces
USB-B for connection to a remote computer
USB-A for connection to a printer or USB memory device
Languages
On-board software in English, Spanish, French, German, Italian, Korean, Japanese, Chinese and Thai2
Power Requirements
100–240 V, 50–60 Hz (selected automatically)
Dimensions (W × D × H)
39 cm × 30 cm × 16 cm (15.3” × 11.8” × 3.6”)
Weight
5.2 kg (11.4 lbs)
Warranty
1 year
1 Standard model reports data to three decimal places. Educator model reports data to two decimal places
2 Educator model only features English, Spanish, and Thai
SPECTRONIC 200 Spectrophotometer ordering information
3 Available in North America from authorized dealers only
For Research Use Only. Not for use in diagnostic procedures.