Dynabeads T reg cells
Isolated cells that are bead- and antibody-free
  • Isolate pure CD4+CD25+ cells with high expression of Foxp3
  • No antibody bound to CD25 after cell isolation
  • Simple cell isolation, no columns
Order now
Dynabeads Regulatory CD4+CD25+ T Cell Kit is an easy-to-use, flow-compatible kit for positive isolation of human regulatory T cells (Tregs). This tube-based technology gives you highly functional Tregs with up to 97% purity (CD4+CD25+ expression); on average over 80% of the isolated CD25+ cells express the transcription factor Foxp3.
Regulatory T cells (Tregs) are vital in maintaining self-tolerance and immune cell homeostasis. Tregs regulate the immune responses responsible for autoimmune diseases and suppress other cell responses.
With this simple three-step magnetic isolation procedure (Figure 1) cells are left both bead- and antibody-free after isolation, and can be used in any cell-based downstream experiment. Due to the Dynabeads polymer coating and the fact that the beads are completely removed after isolation, your cells are exposed to minimal stress and there will be no foreign structures from the isolation method left with your cells after isolation.
Isolating Tregs from PBMC in three steps with Dynabeads
Figure 1. Isolating Tregs from PBMC in three steps with Dynabeads Regulatory CD4+CD25+ T Cell Kit. First, untouched CD4+ T cells are negatively isolated using Ab mix and Depletion Dynabeads (step 1). Next CD25+ cells are positively isolated using Dynabeads CD25 (step 2), and cells are released from beads using the DETACHaBEAD Kit (step 3).
Designed to retain immunosuppressive potential of isolated cells
Regulatory CD4+CD25+ T cells are a specialized subpopulation of T cells that act to maintain homeostasis within the immune system. The characterization of this cell population has firmly established their existence and their critical role in keeping the balance of the immune response. Evidence from experimental mouse and human models have demonstrated that the immunosuppressive potential of these cells can be utilized in the treatment of various diseases such as autoimmunity, infectious diseases and cancer.
Dynabeads Regulatory CD4+CD25+ T Cell Kit is designed to maximize:
  • Suppressive capacity of isolated Tregs
  • Expansion of isolated Tregs
  • Suppressive capacity of expanded Tregs
Process for isolating regulatory T cells with presence of CD4± CD25+
Figure 2. Presence of CD4± CD25+ cells during the three step isolation process.A. In the PBMC sample, 2.5% of the cells were CD4+CD25+. B. After negative isolation of CD4+ cells, 7% of the CD4+ cells expressed CD25. C. After isolation of CD25+ cells using Dynabeads CD25, 1% of the low expressing CD25+ cells remained in the CD4+CD25- fraction. D. The isolated CD4+CD25+ cells were 97% pure.
Tube-based vs. Column-based Treg isolation using Dynabeads
Figure 3. Tube-based vs. Column-based Treg isolation. Higher number of Foxp3+ cells using Dynabeads. Tregs were isolated from two different donors using Dynabeads Regulatory CD4+CD25+ T Cell Kit (left) or a column-based method (right). Treg purity was identified by expression of CD25 and Foxp3 after isolation.
Order the regulatory CD4+/CD25+ T Cell kit
Catalog # Name Size Price (USD) Qty
11363D Dynabeads™ Regulatory CD4+/CD25+ T Cell Kit Each
1,180.65
Online Exclusive
1,328.00
Save 147.35 (11%)
Support
Cell Analysis Support Center
Find tips, troubleshooting help, and resources for your cell analysis workflow.
Services & Support
For Research Use Only. Not for use in diagnostic procedures.
Stylesheet for Classic Wide Template adjustments