Proven Magnetic Separation Technology for Tumor Cells
Dynabeads are used within a variety of applications within the field of cancer research, ranging from detecting circulating tumor cells to expanding T cells in immunotherapy.
On this page
Circulating tumor cell detection
Isolate and detect minimal residual cancer cells with Dynabeads. These easy-to-use products quickly provide clear, sensitive and accurate results. The tube-based method is extremely gentle to cells, ensuring high cell viability and yield.
  • Up to 5 log enrichment of tumor cells
  • Detect 1 tumor cell per ml of blood
  • Obtain viable cells for analysis by applications including staining, FISH, PCR or RT-PCR.
You can enrich tumour cells using Dynabeads coated with antibodies against specific cell markers. Add Dynabeads to the starting sample, which then bind to the target cells. Separate the bead-bound cells with a magnet (DynaMag). The product you choose will depend on your chosen downstream application.
Tumor cell detection is a two part procedure:
  • Enrich tumor cells directly from whole blood, bone marrow or MNC samples
  • Analyze enriched tumor cells for characterization and confirmation
Enriched tumor cells for molecular analysis:
Use Dynabeads Epithelial Enrich to positively isolate epithelial tumor cells from whole blood or bone marrow samples. After enrichment, obtain pure mRNA from the cells with Dynabeads mRNA DIRECT Kit.
Enriched tumor cells for cellular analysis
Option 1:
Use CELLection Epithelial Enrich to enrich tumor cells, then remove the beads from the cells. The bead-free cells can be used for any downstream application.
Dynabeads FlowComp Flexi, CELLection Pan Mouse IgG and CELLection Biotin Binder can also be used in combination with your own tumor-specific antibodies.
Option 2:
Deplete leucocytes from your sample with Dynabeads CD45 to leave untouched tumor cells that can be used for any downstream application.
Human and mouse T cell expansion
Ready-to-use Dynabeads CD3/CD28 T Cell Expander provides a reliable way to repeatedly stimulate T cell clones without the need for autologous or MHC-matched antigen-presenting cells and antigen. Achieve full activation and expansion of T cells by providing simultaneous TCR/CD3 and CD28 signals to T cells in culture. Expanded T cells can be used in immunotherapy research.
Bridge the gap from mouse to human models with the equivalent product for expanding mouse cells, Dynabeads Mouse CD3/CD28 T Cell Expander.
Isolation and expansion
Isolate a high yield and purity of human CD4+CD25+ cells with Dynabeads Regulatory CD4+CD25+ T Cell Kit.
Expand human regulatory T cells with Dynabeads Human Treg Expander. Expanded cells retain suppressive phenotype and capacity.
Immunotherapy
Dynabeads ClinExVivo CD3/CD28 (formerly Xcyte™ Dynabeads) offers a unique and proprietary technology for activation and expansion of autologous T cells to fight cancer and infectious disease. T cells expanded with Dynabeads ClinExVivo CD3/CD28 are currently being tested in clinical trials in patients with chronic lymphocytic leukemia, non-Hodgkin's lymphoma, and multiple myeloma. Read more.
Angiogenesis
Isolate endothelial cells directly from tissue digests with Dynabeads CD31. For rapid and consistent results in protein or gene expression analysis, lyse the endothelial cells while they are still attached to the beads and directly process for further molecular analysis.
Hero banner dark gradient overlay
For Research Use Only. Not for use in diagnostic procedures.
Stylesheet for Classic Wide Template adjustments