microscopic cell image showing multicolor fluorescence
ProLong antifade mountants for fixed cells suppress photobleaching and preserve the signals of your fluorescently labeled target molecules for long-term storage and analysis. ProLong reagents cure within 12–48 hours (depending on sample thickness), forming a semi-rigid gel with a refractive index ranging from 1.47–1.52. Mounted samples can be saved for months with appropriate storage.
Key benefits of ProLong antifade mountants for fixed cells include:
  • Inhibit photobleaching across the spectrum, even after prolonged storage
  • Low background across the spectrum
  • Hard-setting mountants
  • Available with or without DAPI
  • Ready-to-use benchtop formulations
  • Choice of refractive index based on your needs: 1.52 for ProLong Glass reagent and 1.47 for ProLong Diamond and ProLong Gold reagents
Choose the antifade reagent that matches your experiment
Performance highlights of ProLong antifade reagents for fixed cells
ProLong Gold antifade mountant performance
ProLong Gold antifade mountant has become a widely used antifade reagent for our Invitrogen high-performance fluorescent dyes and stains because it preserves signal across the entire visible spectrum and causes little or no quenching of the initial signal. ProLong Gold is room-temperature stable—keep it on the bench, ready to use in your next imaging experiment.
ProLong Diamond antifade mountant performance
ProLong Diamond antifade mountant delivers all the performance of ProLong Gold reagent with Alexa Fluor dyes and provides additional protection for traditional dyes and fluorescent proteins. ProLong Diamond reagent is a superior antifade and mountant, offering low background and photobleaching protection across the visible and near-infrared spectrum. Keep ProLong Diamond reagent refrigerated for the best performance.
ProLong Glass antifade mountant performance
ProLong Glass antifade mountant delivers all the performance of ProLong Diamond mountant and offers the best clarity of the ProLong product series—better resolution and improved focal depth for brighter, sharper images. ProLong Glass reagent is a superior antifade reagent with the finest clarity, offering same-as-glass refractive index of 1.52, best focal depth, and low background and photobleaching protection across the visible and near infrared spectrum. Keep ProLong Glass reagent refrigerated for best performance.
 Download the scientific poster
multipanel image series showing fluorescence stained cell following repeated illumination
A 60-second time-lapse showing the enhanced resistance to photobleaching afforded by ProLong antifade mountants. Fixed HeLa cells were labeled with fluorescein phalloidin and mounted in ProLong Glass reagent, ProLong Diamond reagent, ProLong Gold reagent, or 50% PBS/glycerol. Images were acquired at 12-second intervals using a 20x objective with continuous illumination from a standard 100-watt Hg-arc lamp.
FITC photobleach curves obtained from samples illuminated on a widefield or a confocal microscope. HeLa cells were labeled with mouse anti-tubulin primary antibody, detected with fluorescein (FITC)-labeled goat anti-mouse antibody, and mounted with PBS + 50% glycerol or various commercially available antifade mountants. (A) Photobleach curves were collected by illuminating the samples for 2 minutes using a 100‑watt Hg-arc lamp, imaged using a 20x air objective, then acquired using a 12-bit monochrome camera. The data plotted is the mean florescence intensity of three fields of view over time. (B) Photobleach curves were collected using a confocal microscope with a 20x oil-immersion objective scanning regions of interest fifty times with a pixel dwell time of 1.6 microseconds. Excitation source power intensity was set such that ProLong Diamond retained 50% of initial signal intensity at the end of the final scan. Detector gain was held constant for all mounting media. Plotted data is the mean fluorescence intensity from fifteen regions of interest across mounted samples as number of scans.
Find the right ProLong antifade reagent for fixed cells
ProLong Gold Antifade Mountant
ProLong Diamond Antifade MountantProLong Glass Antifade Mountant
Alexa Fluor dyes
Yes
Yes
Yes
Traditional dyes and fluorescent proteins
No
Yes
Yes
Refractive index (after curing)
1.47
1.47
1.52
Storage temperatureRoom temperature2–8°C2–8°C
 Ordering informationOrdering informationOrdering information
Enhanced signal preservation across the spectrum
ProLong Glass and ProLong Diamond reagents are superior antifades and mountants, offering low background and protection against photobleaching across the visible and near-infrared spectrum in addition to exhibiting less initial quenching of the fluorescent signal. It provides extra protection for traditional dyes such as FITC and fluorescent proteins such as GFP. In a comparison of commercial hard-setting antifade mountants in a standardized experiment, ProLong antifade mountants outperformed other products at preserving signals and preventing photobleaching across the spectrum.
Fluorescent dye
Ex/Em (nm)
Resistance to photobleaching*
ProLong Glass ProLong Diamond ProLong GoldVECTASHIELD
DAPI
345/455
+++
+++
++
++
Hoechst350/461++++++++++++
GFP
488/510
++
++
Not recommended
+
BODIPY FL
505/513
Not tested
+++
+
+
Fluorescein
494/518
+++
+++
+
++
Alexa Fluor 488
495/519
+++
++
++
++
Alexa Fluor Plus 488495/519+++++++
Alexa Fluor 555
555/565
+++
+++
+++
+++
Alexa Fluor Plus 555555/565++++++++++
Cy3
550/570
++
++
++
++
Alexa Fluor 546
556/575
++++
+++
++
Tetramethylrhodamine
555/580
++
+++
+
+
TagRFP555/584Not tested+Not recommended+
Alexa Fluor 568
578/603
+++
+++
+
+
mCherry575/610++++++++++++
Texas Red
595/615
+++
+++
+++
++
Alexa Fluor 594
590/617
+++
+++
+++
++
TO-PRO-3
642/661
+++
+++
+
+
Alexa Fluor 647
652/668
+++
+++
+++
Not recommended
Alexa Fluor Plus 647652/668+++++++++Not recommended
Cy5
650/670
+++
+++
+++
++
Key: +++ = above 80%; ++ = 65—80%, + = 50—65%, Not recommended = below 50%. Percentages express remaining signal intensity.
* Photobleaching resistance was quantified by measuring remaining signal intensity on a Zeiss™ LSM 710 confocal microscope. HeLa or U2OS cells were stained and mounted using standard Immunocytochemistry (ICC) protocols. Five regions within three fields of view were scanned 15 times with 1.58 μs dwell time per pixel. Excitation wavelength and intensity were optimized by fluorophore. On an epi-fluorescence microscope using 100-watt Hg-arc lamp, this amount of light/photon exposure will be equal to 60–90 seconds.
VECTASHIELD is a trademark of Vector Laboratories, Inc., Burlingame, CA. Hoechst is a trademark of Hoechst, GmbH. Cy is a trademark of GE Healthcare UK, Ltd.
Get the sharpest, brightest images
Offering additional benefits for the ProLong reagent line, ProLong Glass antifade mountant features a refractive index (RI) of approximately 1.52, which is similar to that of cover glass, immersion oil, and oil objective optics. This superior optical clarity means ProLong Glass reagent delivers up to 75% improvement in axial resolution and 3–4 times more imageable focal depth compared to mountants with refractive index of 1.47, producing sharp images for monolayers of cells, tissue slices, or 3D cell cultures up to 150 μm thickness/focal depth.
confocal image series of samples mounted in different antifade mountants
ProLong Glass antifade mountant improved the depth of imaging by 40 percent, allowing nuclei to be imaged 70 μm into the tissue section. FFPE pig brain sections were processed and stained with DAPI nuclear stain overnight.  Stained samples were mounted with different commercial, hard-setting antifade media according to each mounting medium's recommended protocols.  Tissue sections were imaged on a Zeiss™ LSM 710 confocal microscope using a Plan-Apochromat 63x/1.4 NA Oil objective, sampling at a rate of 83 nm in the x and y dimensions and 100 nm in the z dimension, with a pixel size of 0.07 μm. Z‑projections were generated using Zeiss™ Zen software.
graph of refractive index vs time for a series of commercial mountants
ProLong Glass Antifade Mountant cures to a final RI of ~1.52. The refractive indices of various ProLong antifade mountants were measured using a Thermo Scientific ABBE-3L refractometer by applying 300 µL of mountant and spreading to cover the measuring prism surface.
graph of resolution as a function of focal depth for samples in two mountants
ProLong Glass, with RI of 1.52, maintains a higher axial resolution than mountants with RI of 1.47. Lateral and axial resolution of 170 μm microspheres in two mountants, plotted as a point spread function (PSF) of images acquired at various focal depths. (Error bars: SD with n ≥ 3).
Axial and lateral confocal images of A549 spheroid mounted with ProLong Glass Antifade Mountant
Axial and lateral confocal images of A549 spheroid mounted with ProLong Glass Antifade Mountant.
ProLong Glass Antifade Mountants
Catalog # Name Size Price (USD) Qty
P36980 ProLong™ Glass Antifade Mountant, 5 x 2 mL Each
332.65
Online Exclusive
358.00
Save 25.35 (7%)
P36982 ProLong™ Glass Antifade Mountant, 2 mL Each
96.65
Online Exclusive
103.00
Save 6.35 (6%)
P36984 ProLong™ Glass Antifade Mountant, 10 mL Each
296.65
Online Exclusive
323.00
Save 26.35 (8%)
P36981 ProLong™ Glass Antifade Mountant with NucBlue™ Stain, 5 x 2 mL Each
344.65
Online Exclusive
377.00
Save 32.35 (9%)
P36983 ProLong™ Glass Antifade Mountant with NucBlue™ Stain, 2 mL Each
107.65
Online Exclusive
116.00
Save 8.35 (7%)
P36985 ProLong™ Glass Antifade Mountant with NucBlue™ Stain, 10 mL Each
304.65
Online Exclusive
333.00
Save 28.35 (9%)
ProLong Diamond Antifade Mountants
Catalog # Name Size Price (USD) Qty
P36961 ProLong™ Diamond Antifade Mountant, 5 x 2 mL Each
298.65
Online Exclusive
322.00
Save 23.35 (7%)
P36965 ProLong™ Diamond Antifade Mountant, 2 mL Each
88.65
Online Exclusive
95.00
Save 6.35 (7%)
P36970 ProLong™ Diamond Antifade Mountant, 10 mL Each
268.65
Online Exclusive
291.00
Save 22.35 (8%)
P36962 ProLong™ Diamond Antifade Mountant with DAPI, 5 x 2 mL Each
312.65
Online Exclusive
337.00
Save 24.35 (7%)
P36966 ProLong™ Diamond Antifade Mountant with DAPI, 2 mL Each
95.65
Online Exclusive
103.00
Save 7.35 (7%)
P36971 ProLong™ Diamond Antifade Mountant with DAPI, 10 mL Each
287.65
Online Exclusive
309.00
Save 21.35 (7%)
ProLong Gold Antifade Mountants
Catalog # Name Size Price (USD) Qty
P36934 ProLong™ Gold Antifade Mountant, 5 x 2 mL Each
296.65
Online Exclusive
323.00
Save 26.35 (8%)
P36930 ProLong™ Gold Antifade Mountant, 10 mL Each
277.65
Online Exclusive
302.00
Save 24.35 (8%)
P36935 ProLong™ Gold Antifade Mountant with DNA Stains DAPI, 5 x 2 mL Each
307.65
Online Exclusive
332.00
Save 24.35 (7%)
P36931 ProLong™ Gold Antifade Mountant with DNA Stains DAPI, 10 mL Each
290.65
Online Exclusive
313.00
Save 22.35 (7%)
Stylesheet for Classic Wide Template adjustments
Hero banner dark gradient overlay
For Research Use Only. Not for use in diagnostic procedures.