Search Thermo Fisher Scientific
![]() | ![]() | ![]() | ![]() | ![]() | ![]() | ![]() |
3 | 349 | - | 358 | 763 | (in buffer) 5 | flow cytometry |
NovaFluor UV 765 dye has a unique emission spectrum when excited by the 349 nm laser, which fits in a spectral space that is currently not accessible by other spectral dyes. Adding spectrally unique dyes may allow for building out larger panels with minimal loss of resolution. NovaFluor UV 765 dye should generally be paired with moderately to highly expressed antigens in multicolor panels. The macromolecule-based NovaFluor UV 765 dye produces highly stable fluorescence, and stained samples retain their fluorescence intensity and spectral signature when stored at 4°C. Use NovaFluor dyes with CellBlox Plus Blocking Buffer to reduce background and to block non-specific binding of NovaFluor labels, PE and APC tandems observed with macrophages and monocytes.
NovaFluor dyes, including NovaFluor UV 765 dye, are built using Phiton technology and are compatible with both spectral flow cytometry and traditional flow cytometry. NovaFluor dyes exhibit narrow emission spectra and minimal cross-laser excitation, which helps reduce spectral spillover for better marker resolution. In addition, their unique spectral signatures can provide the opportunity to detect additional markers in flow cytometry panels by opening up previously unusable channels.
Protocols that fit your needs in flow cytometry ranging from sample preparation to numerous cell stimulation conditions, staining, immunophenotyping, and data analysis strategies.
A handy reference poster featuring the broad range of our available dyes and labeling reagents.
Protocols that fit your needs in imaging ranging from sample and assay preparation to staining, labeling, and data analysis strategies.
A tool for selecting the optimal fluorescent dyes for your Invitrogen EVOS cell imaging systems.
Cy™ is a trademark or registered trademark of GE Healthcare.
Not for resale. Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.
Brilliant Ultra Violet™ and Brilliant Violet™ are trademarks or registered trademarks of Becton, Dickinson and Company or its affiliates, and are used under license.
For Research Use Only. Not for use in diagnostic procedures.