Alexa Fluor 700 excitation shown as dotted line and emission shown as solid red histogram
1633/640720/30696719(in buffer) 1
flow cytometry
Invitrogen Alexa Fluor 700 dye is a dim, near-infrared dye that can be excited with 633-640 nm red lasers. Highly expressed antigens can be labeled with Alexa Fluor 700 in flow cytometry panels allowing for brighter dyes to be used with less-abundant antigens. It fits in a panel between APC and APC-eFluor 780.
We offer Alexa Fluor 700 dye conjugated to a variety of antibodies for cellular labeling and detection. In addition, reactive dye forms and protein labeling kits are available to enable you to generate your own antibody conjugates or probes.
Protein Labeling Reagents Selection Guide (NHS ester, maleimide, etc.)  Search Alexa Fluor 700 primary antibodies
flow cytometry dot plot of human mononuclear cells using Zenon Alexa Fluor 700 Mouse IgG1 Labeling Kit
Flow cytometry analysis of human mononuclear cells using Zenon Alexa Fluor 700 Labeling Kit.
  An antibody complex was prepared using the Zenon Alexa Fluor 700 Mouse IgG1 Labeling Kit and mouse anti-human CD4. Human mononuclear cells were stained with the antibody complex and analyzed by flow cytometry using a 633 nm laser.
2 peak histogram showing cells stained with  anti-CD4 biotin followed by streptavidin Alexa Fluor 700 conjugate
Flow cytometry analysis of human mononuclear cells using Alexa Fluor 700 streptavidin. Human mononuclear cells were incubated with CD4, Mouse Anti-Human (Biotin), washed and stained with Streptavidin, Alexa Fluor 700 conjugate. Cells were washed and analyzed by flow cytometry using 633 nm excitation.
Additional resources
Flow cytometry protocols handbook
Protocols that fit your needs in flow cytometry ranging from sample preparation to numerous cell stimulation conditions, staining, immunophenotyping, and data analysis strategies.
Request the handbook
Cy™ is a trademark or registered trademark of GE Healthcare.
Not for resale. Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.
Brilliant Ultra Violet™ and Brilliant Violet™ are trademarks or registered trademarks of Becton, Dickinson and Company or its affiliates, and are used under license.
For Research Use Only. Not for use in diagnostic procedures.