Deliver consistent results with a furnace you can depend on to attain your daily goals. Thermo Scientific Box and Tube Furnaces come in a wide range of offerings to accommodate ordinary and technical tasks alike, such as: temperature ranges up to 1200°C, temperature control options to meet your application needs, and embedded or open heating elements designed to keep samples safe while maintaining reliable temperature uniformity.
Choosing between a box furnace or a tube furnace for your laboratory
Box Furnaces
  • Used for processing larger samples or easy access to samples
  • Small, medium and large box furnaces available
  • Ashing applications in every industry and research laboratory 
  • Attributes include variable density insulation, double shell cabinets, long-life heating elements and vertical, horizontal side swing or swing down doors
Thermolyne Furnace Large
Tube Furnaces
  • Used for processing small samples or samples in an inert atmosphere
  • Single and three-zoned designs for precise temperature control. Three zone control helps enable different temperatures in each zone.
  • Materials testing, pyrolysis, and crystal growing applications
  • Some models offer split-hinge design for ease of tube changing
Lindberg furnace small
BSC touchscreen display shows operational parameters
Discover a furnace for your application
View our quick application guide for choosing a furnace that is suitable for the workload you need. Choose from a variety of temperature control options, heating elements and furnace formats to meet your analytical requirements.
Lab Box Furnaces
Typically used for processing larger samples or to easily place and access samples. We offer a versatile selection of small, medium, and large box furnaces suitable for a variety of industrial and laboratory applications. Advanced engineering and specialized construction techniques include variable density insulation, double shell cabinets, long-life heating elements and side-swing doors (vertical or horizontal) or swing down doors.
Lindberg/Blue M LGO
1200°C Box Furnaces
Lindberg/Blue M Moldatherm Box furnaces
Thermolyne Largest Tabletop Muffle Furnaces
Thermolyne Benchtop Muffle Furnaces
Thermolyne Premium Large Muffle Furnaces
Thermolyne Small Benchtop Muffle Furnaces
Heat range
100 - 1200°C
100 - 1100°C
100 - 1093°C
100 - 1200°C
100 - 1200°C
100 - 1100°C
Available capacity
16.4 - 55.3 L
1.99 - 42.5 L
45 L
14 L
2 - 5.8 L
1.3 to 2.1 L
Heating elements
Embedded
Embedded
Embedded
Embedded
Open
Embedded
Lab Tube Furnaces
Tube furnaces are typically used for processing small samples or heating in an inert atmosphere. The single and three-zoned construction is designed to create precise temperature control. Three-zone control enables the user to select a different temperature in each zone (e.g. for gas applications or material experiments). Some models offer split-hinge design, which easily allows you to change the tube.
Lindberg/Blue M 1200°C Split-Hinge Tube Furnaces*
Lindberg/Blue M 1100°C Tube Furnaces
Lindberg/Blue M Mini-Mite Tube Furnaces
Heat range
100 - 1200°C
100 - 1100°C
100 - 1100°C
Available capacity
30.5, 61, or 91.4 cm /
12, 24, or 36 in.
61 or 91.4 cm /
24 or 36 in.
30.5 cm /
12 in.
Heating elements
Open
Embedded
Open
Furnace Accessories
Replacement Parts
*Additional replacement parts may be available. Contact your local representative or customer support to order the following: hearth tray for Thermolyne small benchtop muffle furnace model FB1300 (Cat. No. PH44X1); hearth tray for Thermolyne small benchtop muffle furnace model FB1400 (Cat. No. PH48X1); hearth tray for Thermolyne industrial benchtop muffle furnace (Cat. No. PHX2).
Support
Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services