Find an incubator that is right for you using the below tables. 
Choose between Heratherm Microbiological Incubators and Heratherm Refrigerated Incubators.
Microbiological incubators, also called "heat-only," or "standard" incubators, have heating elements and can provide incubation temperatures that are above ambient temperature only. If the laboratory has an ambient of about 22°C, they can only address incubation temperatures above about 27°C or even 30°C. Temperature ranges vary by model.
Refrigerated incubators, also called "cooling" incubators have cooling and heating and can provide a wider temperature range. They can offer temperatures close to ambient or even below ambient. They usually also cover the incubation temperature range above ambient, just as the microbiological or "heat-only" incubators do. 
Thermo Scientific Heratherm Microbiological Incubators
Heratherm Advanced Protocol Security Incubators
Ensure outstanding sample safety with multiple alarms and a 140℃ decontamination cycle.
Heratherm Advanced Protocol Incubators
Attain excellent heating consistency, well suited for a variety of drying applications.
Heratherm General Protocol Incubators
Benchtop and floor model versions available for routine usage.
Heratherm Compact Microbiological Incubators
Ideally suited for a personalized workspace or crowded lab.
Temperature Range
Ambient+5 to +105°C
Ambient+5 to +105°C
Ambient +5 to +75°C
+18 to + 40°C
Convection technology
Benchtop – Dual
Large capacity - Mechanical
Dual
Gravity
Peltier with Mechanical
Temperature performance
Best
Best
Good
Good 
Alarms
Automatic over-temperature
Automatic under temperature
Door alarm
Automatic over-temperature
Automatic over-temperature
Timer
Weekly/daily/real time
Weekly/daily/real time
on/off
Additional sample security features
Certified decontamination cycle*
Lockable doors
Connection for temperature sensor
– –
– –
Easy calibration routine for high temperature assuracy
Ordering information
View all models
View all models
View all models
View all models
*Verified by independent testing facility (IBFE 9/2010)
NOTE: All figures in tables are typical average values for series devices, based on factory standard following norm Din12880. Please contact us for certification information or IQ/OQ documents.
Thermo Scientific Heratherm Refrigerated Incubators
Heratherm Refrigerated Incubator IMP180 benchtop model   Heratherm Refrigerated Incubator, IMP400 floor model Thermo Scientific Refrigerated Incubators
Ideally suited for a personalized workspace or crowded lab.
Reach and maintain the precise ambient temperatures you need for reliable results while maintaining efficient energy usage.
Ideal for precise temperature control in applications such as biological sample preservation, product development, quality control, and microbial analysis
Temperature Range
+5 to +70°C
+5 to +70°C
+4 to +60°C
Capacity
Benchtop- IMP180
Floor model- IMP400
Benchtop  - 150L and 250L
Floor models- 420L and 720L
Convection technology
Peltier
Peltier
Compressor
Temperture performance
Best Best Good
Number of shelf positions
19
39
19 for Benchtop models
39 for Floor models
Energy consumption at 20°C
80 Wh/h
150 Wh/h
80 Wh/h for Benchtop models
150 Wh/h for Floor models
Ordering information
View all models
View all models
View all models
*Verified by independent testing facility (IBFE 9/2010)
NOTE: All figures in tables are typical average values for series devices, based on factory standard following norm Din12880. Please contact us for certification information or IQ/OQ documents.
For Laboratory Use.
1x1 image pixel for data collection