Achieve excellent specificity, sensitivity, and reproducibility in with TaqMan Assays. TaqMan gene expression assays represent the largest collection of predesigned gene expression assays in the industry, with over 2.8 million assays available across more than 30 species and numerous microbes. These assays are designed to detect virtually any gene transcript, providing comprehensive coverage to identify the maximun number of transcript variants. By eliminating the need to optimize SYBR primers, TaqMan qPCR chemistry allows for faster and more reliable results. Furthermore, these assays come in a range of flexible formats, including single tubes, 96-well plates, 384-well plates, 384-well array cards, and OpenArray plates, offering exceptional versatility to suit various experimental needs. With TaqMan gene expression assays, scientists can streamline their gene expression studies and obtain high-quality data with ease.
Search and order TaqMan gene expression assays
Video: How to find & order the right TaqMan predesigned assay using the assay search tool above
TaqMan Assays & Arrays for oncology biomarkers
Learn more
TaqMan gene expression assay formats
TaqMan gene expression assays come in several types to suit diverse research needs, including predesigned assays that cover over 2.8 million targets across more than 30 species, enabling broad applicability and convenience. Custom assays allow researchers to tailor the design to specific target sequences, offering precision and flexibility for unique experimental requirements. Flexible and specialty assays offer additional customization options, including the ability to configure assays in various formats such as 96-well plates and 384-well microfluidic cards, catering to both standard and high-throughput applications.
Ready-to-go format. Predesigned TaqMan Gene Expression Assays require no optimization or bioinformatics expertise to design qPCR assays.
Search: Predesigned TaqMan gene expression assays
Available for human, mouse, and rat in various formats. Use a custom configurator to select and design a combination of assays.
Explore: Flexible gene expression assays
TaqMan gene expression assay configurations
TaqMan gene expression assays are available in several configurations to accommodate different research needs. Configuration offerings include single-tube assays for individual experiments, 96-well plates for medium-throughput applications, and 384-well microfluidic cards for high-throughput screening. Additionally, the Applied Biosystems OpenArray platform offers a flexible and scalable solution for large-scale gene expression studies.
More TaqMan gene expression resources
Comprehensive resources and information on gene expression analysis using real-time PCR, including protocols, application notes, and troubleshooting tips to support researchers in optimizing their experiments.
Learn more: TaqMan Gene Expression Learning Center
Detailed information on using real-time PCR for gene expression analysis, highlighting its applications, benefits, and technical considerations for accurate and efficient experimentation.
Learn more: Gene expression applications
For Research Use Only. Not for use in diagnostic procedures.
Stylesheet for Classic Wide Template adjustments
Hero banner dark gradient overlay