We offer a wide variety of staining and drying accessories to help you process your protein gel electrophoresis gels.
StainEase® Staining Trays 
StainEase® Staining Trays
The StainEase® Staining Tray is a convenient way to evenly stain minigels and prevent gels from breaking. Easily fits up to 4 mini- or midi-gels.
DryEase® Mini-Gel Drying System   
DryEase® Mini-Gel Drying System
Designed for even, crack-free drying of minigels, the DryEase® Mini-Gel Drying System is a simple-to-use kit that dries gels using passive evaporation. Gels dried with this system are suitable for fluorography, densitometry, autoradiography, or permanent storage.
Each DryEase® Mini-Gel Drying System contains the following components:
  • 2-DryEase® Mini-Gel Drying Frames (includes 2 frames + 8 clamps)
  • 1-DryEase® Mini-Gel Drying Base
  • 200-DryEase® Mini Cellophane precut sheets
  • 500 mL Gel-Dry™ Drying Solution
DryEase® Mini-Gel Drying System 
Large Gel Drying Kit
The Large Gel Drying Kit offers the same even, crack-free gel drying as available with the DryEase® Mini-Gel Drying System, but is designed to accommodate larger gels or 4 mini-gels. The system does not require a base.
Each Large Gel Drying Kit contains the following components:
  • 1-DryEase® Large Gel Drying Frame  (includes 8 clamps)
  • 100-DryEase® Large Cellophane precut sheets
  • 500 mL Gel-Dry™ Drying Solution
Gel staining and drying accessories ordering Information
Catalog # Name Size Price (USD) Qty
NI2400 StainEase™ Staining Trays Each
147.65
Online Exclusive
152.00
Save 4.35 (3%)
NC2380 DryEase™ Mini Cellophane Each
133.00
NI2300 DryEase™ Mini-Gel Drying Base Each
94.25
NI2380 DryEase™ Mini-Gel Drying Frame Each
147.65
Online Exclusive
152.00
Save 4.35 (3%)
NI2387 DryEase™ Mini-Gel Drying System Each
308.00
NI2100 Gel-Dry™ Clamp Each
46.40
NC2200 Large Cellophane Each
186.00
NI2200 Large Gel Drying Frame Each
240.00
NI2207 Large Gel Drying Kit Each
284.00
LC4025 Gel-Dry™ Drying Solution, 500 mL Each
65.65
Online Exclusive
69.25
Save 3.60 (5%)
LC40254 Gel-Dry™ Drying Solution, 8 x 500 mL Each
324.65
Online Exclusive
345.00
Save 20.35 (6%)
For Research Use Only. Not for use in diagnostic procedures.