Gel-Dry™ Drying Solution
Gel-Dry™ Drying Solution
Invitrogen™

Gel-Dry™ Drying Solution

Gel-Dry™ Drying Solution is one component of the DryEase™ Mini-Gel Drying System, which is a simple, easy-to-use kit for dryingRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric) Buffer
LC4025500 mL
LC402548 x 500 mL
Catalog number LC4025
Price (USD)
65.65
Online Exclusive
69.25
Save 3.60 (5%)
Each
-
Add to cart
Volume (Metric) Buffer:
500 mL
Recurring order eligible. Learn more »
Price (USD)
65.65
Online Exclusive
69.25
Save 3.60 (5%)
Each
Add to cart
Ask our AI about this Product
Gel-Dry™ Drying Solution is one component of the DryEase™ Mini-Gel Drying System, which is a simple, easy-to-use kit for drying gels evenly using passive evaporation. The kit allows gels to be assembled on a base between two open frames, so that both outer surfaces of the gels are exposed to air. Consequently, drying is even and cracking is minimized. The proprietary Gel-Dry™ Drying Solution modulates the rate of water release to ensure crack-free gels. With the DryEase™ System, gels are dried overnight with uniform tension on all points of the gel to eliminate cracking.

Applications: Gels dried with the DryEase™ System are suitable for fluorography, densitometry, autoradiography, or permanent gel storage.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
BufferDrying Solutions
Quantity500 mL
Shipping ConditionRoom Temperature
Volume (Metric) Buffer500 mL
For Use With (Equipment)DryEase™ Mini-Gel Drying System
Gel SizeMini
Product LineNovex
TypeDrying Solution
Unit SizeEach
Contents & Storage
Store at room temperature.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3197422Certificate of AnalysisJun 19, 2025LC4025
3123484Certificate of AnalysisApr 30, 2025LC4025
3079862Certificate of AnalysisJan 13, 2025LC4025
3005626Certificate of AnalysisSep 30, 2024LC4025
2990031Certificate of AnalysisAug 23, 2024LC40254, LC4025-4
5 results displayed, search above for a specific certificate

Safety Data Sheets

Frequently asked questions (FAQs)

The Gel-Dry Drying solution has a proprietary composition.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

Yes, it is compatible. You need to put the gel in the enhancer solution, wash it and then proceed with the gel drying solution. For detection of signal using 32P, the cellophane is fine. For 35S, the upper layer of cellophane may need to be removed (which is possible) depending on the enhancer being used. For detection of 14C, there is no need to remove the cellophane as it will pass through.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

Share catalog number, name or link

1x1 image pixel for data collection