Zymogram gels
Invitrogen Novex Zymogram Gels are excellent tools for detecting and characterizing proteases that utilize casein or gelatin as a substrate.
Interactive selection toolAvailable Novex Zymogram Gels
Migration chart
Novex Zymogram Gels are excellent tools for detecting and characterizing proteases that utilize gelatin as a substrate. Novex Zymogram Gels can be used to analyze a variety of enzymes, including matrix metalloproteinases, lipases, and other proteases. Novex Zymogram Gels are based on tris-glycine gel chemistry containing gelatin as the substrate.
How Novex Zymogram Gels work
Protease samples are denatured in SDS buffer under non-reducing conditions and without heating, and run on a Novex Zymogram Gel using tris-glycine SDS running buffer. After electrophoresis, the enzyme is renatured by incubating the gel in Invitrogen Novex Zymogram Renaturing Buffer, which contains a nonionic detergent. The gels are then equilibrated in Invitrogen Novex Zymogram Developing Buffer to add divalent metal cations required for enzymatic activity, and then stained and destained. Regions of protease activity appear as clear bands against a dark blue background where the protease has digested the substrate.
Available gel sizeMini: 8 cm x 8 cm (1 mm thick) 
Storage conditions2–8°C
Shelf life2 months
Gel chemistry Tris-glycine–containing gelatin or casein
Running bufferTris-glycine SDS running buffer
Available polyacrylamide concentrations10% (w/gelatin)
Separation range10–220 kDa
For use with (equipment) mini gelsMini Gel Tank or XCell SureLock Mini-Cell
Mode of separation Molecular weight
 Zymogram Gelatin Gel
Gel composition10% tris-glycine gel
Substrate0.1% gelatin
Sensitivity5 x 10-6 units of collagenase
Post-staining requiredYes
Separation range20–120 kDa
Catalog # Name Size Price (USD) Qty
ZY00100BOX Novex™ 10% Zymogram Plus (Gelatin) Protein Gels, 1.0 mm, 10-well, 10 gels (1 box) 10 gels (1 box)
226.65
Online Exclusive
233.00
Save 6.35 (3%)
ZY00102BOX Novex™ 10% Zymogram Plus (Gelatin) Protein Gels, 1.0 mm, 12-well, 10 gels (1 box) 10 gels (1 box)
226.65
Online Exclusive
233.00
Save 6.35 (3%)
ZY00105BOX Novex™ 10% Zymogram Plus (Gelatin) Protein Gels, 1.0 mm, 15-well, 10 gels (1 box) 10 gels (1 box)
226.65
Online Exclusive
233.00
Save 6.35 (3%)
Catalog # Name Size Price (USD) Qty
LC2675 Novex™ Tris-Glycine SDS Running Buffer (10X), 500 mL Each
38.65
Online Exclusive
39.91
Save 1.26 (3%)
A25977 Mini Gel Tank Each
735.65
Online Exclusive
760.00
Save 24.35 (3%)
LC2676 Novex™ Tris-Glycine SDS Sample Buffer (2X) Each
29.02
LC26754 Novex™ Tris-Glycine SDS Running Buffer (10X), 4 x 1 L Each
150.65
Online Exclusive
155.00
Save 4.35 (3%)
B1000 Mini Blot Module Each
480.00
LC2670 Novex™ Zymogram Renaturing Buffer (10X) Each
60.00
NW2000 Mini Gel Tank and Blot Module Set Each
1,314.65
Online Exclusive
1,368.00
Save 53.35 (4%)
LC2671 Novex™ Zymogram Developing Buffer (10X) Each
71.65
Online Exclusive
73.00
Save 1.35 (2%)
Novex Zymogram Gels are excellent tools for detecting and characterizing proteases that utilize gelatin as a substrate. Novex Zymogram Gels can be used to analyze a variety of enzymes, including matrix metalloproteinases, lipases, and other proteases. Novex Zymogram Gels are based on tris-glycine gel chemistry containing gelatin as the substrate.
How Novex Zymogram Gels work
Protease samples are denatured in SDS buffer under non-reducing conditions and without heating, and run on a Novex Zymogram Gel using tris-glycine SDS running buffer. After electrophoresis, the enzyme is renatured by incubating the gel in Invitrogen Novex Zymogram Renaturing Buffer, which contains a nonionic detergent. The gels are then equilibrated in Invitrogen Novex Zymogram Developing Buffer to add divalent metal cations required for enzymatic activity, and then stained and destained. Regions of protease activity appear as clear bands against a dark blue background where the protease has digested the substrate.
Available gel sizeMini: 8 cm x 8 cm (1 mm thick) 
Storage conditions2–8°C
Shelf life2 months
Gel chemistry Tris-glycine–containing gelatin or casein
Running bufferTris-glycine SDS running buffer
Available polyacrylamide concentrations10% (w/gelatin)
Separation range10–220 kDa
For use with (equipment) mini gelsMini Gel Tank or XCell SureLock Mini-Cell
Mode of separation Molecular weight
 Zymogram Gelatin Gel
Gel composition10% tris-glycine gel
Substrate0.1% gelatin
Sensitivity5 x 10-6 units of collagenase
Post-staining requiredYes
Separation range20–120 kDa
Catalog # Name Size Price (USD) Qty
ZY00100BOX Novex™ 10% Zymogram Plus (Gelatin) Protein Gels, 1.0 mm, 10-well, 10 gels (1 box) 10 gels (1 box)
226.65
Online Exclusive
233.00
Save 6.35 (3%)
ZY00102BOX Novex™ 10% Zymogram Plus (Gelatin) Protein Gels, 1.0 mm, 12-well, 10 gels (1 box) 10 gels (1 box)
226.65
Online Exclusive
233.00
Save 6.35 (3%)
ZY00105BOX Novex™ 10% Zymogram Plus (Gelatin) Protein Gels, 1.0 mm, 15-well, 10 gels (1 box) 10 gels (1 box)
226.65
Online Exclusive
233.00
Save 6.35 (3%)
Catalog # Name Size Price (USD) Qty
LC2675 Novex™ Tris-Glycine SDS Running Buffer (10X), 500 mL Each
38.65
Online Exclusive
39.91
Save 1.26 (3%)
A25977 Mini Gel Tank Each
735.65
Online Exclusive
760.00
Save 24.35 (3%)
LC2676 Novex™ Tris-Glycine SDS Sample Buffer (2X) Each
29.02
LC26754 Novex™ Tris-Glycine SDS Running Buffer (10X), 4 x 1 L Each
150.65
Online Exclusive
155.00
Save 4.35 (3%)
B1000 Mini Blot Module Each
480.00
LC2670 Novex™ Zymogram Renaturing Buffer (10X) Each
60.00
NW2000 Mini Gel Tank and Blot Module Set Each
1,314.65
Online Exclusive
1,368.00
Save 53.35 (4%)
LC2671 Novex™ Zymogram Developing Buffer (10X) Each
71.65
Online Exclusive
73.00
Save 1.35 (2%)
For Research Use Only. Not for use in diagnostic procedures.