The most commonly used solvents or solvent blends include LC-MS grade water and acetonitrile, with ion pairing agents such as trifluoroacetic acid (TFA), formic acid (FA) or heptafluorobuteric acid (HFBA). These solvents are available as single component formulations or convenient blends. Solvents are manufactured in an ISO9001 facility and are provided with a Certificate of Analysis. Solvents and blends have been validated using Thermo Scientific™ Mass Spectrometers.
  • High purity—processed, filtered and sealed to help extend LC-MS column life
  • More choices—solvents and solvent blends to meet a variety of applications/needs
  • Stable—store for more than one year at room temperature
  • Validated—products evaluated with over 20 specifications measured and fully tested using Thermo Scientific Mass™ Spectrometers
Choose the right LC-MS solvent for your application
 Trifluoroacetic Acid (TFA)Formic Acid (FA)0.1% TFA in Acetonitrile, LC-MS Grade0.1%TFA in Water, LC-MS Grade0.1% FA in Acetonitrile, LC-MS Grade0.1% FA in Water, LC-MS Grade
StructureChemical structure of trifluoroacetic acid (TFA)Chemical structure of formic acid (FA)Chemical structure of trifluoroacetic acid (TFA)Chemical structure of trifluoroacetic acid (TFA)Chemical structure of formic acid (FA)Chemical structure of formic acid (FA)
FormulationSolventSolventSolvent BlendSolvent BlendSolvent BlendSolvent Blend
FormatsLiquid (in ampules or bottles)
Liquid (in ampules or bottles)
Liquid (bottles)Liquid (bottles)Liquid (bottles)Liquid (bottles)
Specifications measured>107-20>20>20>20>20
Order nowOrder nowOrder nowOrder nowOrder nowOrder now
Featured data
Specifications of Thermo Scientific Pierce 0.1% Trifluoroacetic Acid (v/v) in Acetonitrile.
Specifications of Thermo Scientific Pierce 0.1% Trifluoroacetic Acid (v/v) in Acetonitrile.
Related mass spec solvents and reagents
Katalognr. Produkt Größe Preis (EUR) Menge
53104 Pierce™ Heptafluorobutyric Acid (HFBA), HPLC Grade 10 x 1 mL
200,00
28903 Pierce™ Trifluoroacetic Acid (TFA), Sequencing grade, 100 g Each
220,65
Exklusiv online
258,00
Erparnis 37,35 (14%)
28902 Pierce™ Trifluoroacetic Acid (TFA), Sequencing grade, 10 x 1 g Each
143,65
Exklusiv online
167,00
Erparnis 23,35 (14%)
For Research Use Only. Not for use in diagnostic procedures.