Search Thermo Fisher Scientific
The Thermo Scientific Pierce Standards are designed for a variety of applications, including sensitivity assessment, determination of digestion efficiency, chromatography assessment and/or as controls for sample analysis. Standards range from single component formulations to complex human-based digests. Each standard is provided with a Certificate of Analysis.
![]() | ![]() | ![]() | ![]() | ![]() | |
Pierce BSA Digest Standard | Pierce 6-Protein Mixture Standard | Pierce Yeast Digest Standard | Pierce HeLa Protein Digest Standard | Pierce TMT11plex Yeast Digest Standard | |
---|---|---|---|---|---|
Components (abbreviated) | BSA tryptic peptides | Lysozyme, BSA, Cytochrome C, Alcohol dehydrogenase, B-galactosidase, Apotransferrin (multi-species) | Yeast lysate peptide digest | HeLa lysate peptides | Yeast lysate peptide digest |
Format | Lyophilized | Lyophilized | Lyophilized | Lyophilized | Lyophilized |
Primary application | Protein sample control | Chromatography assessment | Monitoring LC/MS system performance | Complex sample control | Complex sample for TMT-labeling |
Complexity of standard | ++ | +++ | +++ | ++++ | +++ |
Recommended storage | -20°C | -20°C | -20°C | -20°C | -20°C |
Order | Order | Order | Order | Order |
![]() | |
Pierce Reserpine Standard for LC-MS | |
---|---|
Components (abbreviated) | Reserpine molecule |
Format | Liquid |
Primary application | Sensitivity assessment |
Complexity of standard | + |
Recommended storage | 4°C |
Order |
![]() | ![]() | ![]() | |
Pierce Peptide Retention Time Calibration Mixture | Pierce LC-MS/MS System Suitability Standard (7 x 5 Mix) | Pierce HeLa Digest/PRTC Standard | |
---|---|---|---|
Components (abbreviated) | 15 heavy tryptic peptides (yeast) | 7 yeast tryptic peptides (5 variations of light or heavy amino acids) | Mixture of 10 µg of HeLa protein digest with 5 pmol PRTC peptides |
Format | Frozen liquid | Frozen liquid | Lyophilized |
Primary application | Chromatography assessment | Targeted MS Assay control | Monitoring LC/MS system performance |
Complexity of standard | ++ | ++ | ++++ |
Recommended storage | -80°C | -80°C | -20°C |
Order | Order | Order |
![]() | ![]() | |
Pierce Intact Protein Standard Mix | Pierce Aquaporin Z Membrane Protein Standard | |
---|---|---|
Components (abbreviated) | IGF-I LR3, Thioredoxin, Protein G, Carbonic Anhydrase II, Protein AG (chimeric), Exo Klenow | Purified Aquaporin Z protein |
Format | Lyophilized | Liquid |
Primary application | Top down method development | Native mass spectrometry |
Complexity of standard | +++ | + |
Recommended storage | -20°C | -80°C |
Order | Order |
![]() | ![]() | ![]() | ![]() | ![]() | |
Pierce BSA Digest Standard | Pierce 6-Protein Mixture Standard | Pierce Yeast Digest Standard | Pierce HeLa Protein Digest Standard | Pierce TMT11plex Yeast Digest Standard | |
---|---|---|---|---|---|
Components (abbreviated) | BSA tryptic peptides | Lysozyme, BSA, Cytochrome C, Alcohol dehydrogenase, B-galactosidase, Apotransferrin (multi-species) | Yeast lysate peptide digest | HeLa lysate peptides | Yeast lysate peptide digest |
Format | Lyophilized | Lyophilized | Lyophilized | Lyophilized | Lyophilized |
Primary application | Protein sample control | Chromatography assessment | Monitoring LC/MS system performance | Complex sample control | Complex sample for TMT-labeling |
Complexity of standard | ++ | +++ | +++ | ++++ | +++ |
Recommended storage | -20°C | -20°C | -20°C | -20°C | -20°C |
Order | Order | Order | Order | Order |
![]() | |
Pierce Reserpine Standard for LC-MS | |
---|---|
Components (abbreviated) | Reserpine molecule |
Format | Liquid |
Primary application | Sensitivity assessment |
Complexity of standard | + |
Recommended storage | 4°C |
Order |
![]() | ![]() | ![]() | |
Pierce Peptide Retention Time Calibration Mixture | Pierce LC-MS/MS System Suitability Standard (7 x 5 Mix) | Pierce HeLa Digest/PRTC Standard | |
---|---|---|---|
Components (abbreviated) | 15 heavy tryptic peptides (yeast) | 7 yeast tryptic peptides (5 variations of light or heavy amino acids) | Mixture of 10 µg of HeLa protein digest with 5 pmol PRTC peptides |
Format | Frozen liquid | Frozen liquid | Lyophilized |
Primary application | Chromatography assessment | Targeted MS Assay control | Monitoring LC/MS system performance |
Complexity of standard | ++ | ++ | ++++ |
Recommended storage | -80°C | -80°C | -20°C |
Order | Order | Order |
![]() | ![]() | |
Pierce Intact Protein Standard Mix | Pierce Aquaporin Z Membrane Protein Standard | |
---|---|---|
Components (abbreviated) | IGF-I LR3, Thioredoxin, Protein G, Carbonic Anhydrase II, Protein AG (chimeric), Exo Klenow | Purified Aquaporin Z protein |
Format | Lyophilized | Liquid |
Primary application | Top down method development | Native mass spectrometry |
Complexity of standard | +++ | + |
Recommended storage | -20°C | -80°C |
Order | Order |
The TMT11plex Yeast Digest Standard can be used to assess instrument readiness and protein quantification accuracy. The TMT11plex Yeast Digest Standard was used to assess MS instrument readiness on the precision of quantification of a housekeeping protein, ENO2. Protein abundance was scaled to the parental lines in Proteome Discover 2.2 software, with expected ratios of 1:1. A total of 500 ng of the Pierce TMT11plex Yeast Digest Standard was measured using a 50 min gradient on an UltiMate 3000 RSLCnano system with a synchronous precursor selection (SPS)-MS3 (SPS-MS3) method of the Orbitrap Fusion Lumos Tribrid Mass Spectrometer.
Thermo Scientific Pierce HeLa Digest Standard base peak chromatogram. Chromatogram of 200 ng Pierce HeLa Protein Digest Standard separated using a Thermo Scientific Acclaim PepMap 100 3 μm x 75 μm x 15 cm column (Product # 160321) with a 2-35% gradient (A: 0.1% FA in water, B: 0.1% FA in 100% acetonitrile) at 300 nL/min for 120 minutes and detected on a Thermo Scientific LTQ Orbitrap XL Mass Spectrometer.
Example selected reaction monitoring (SRM) of 500fg reserpine on a Thermo Scientific Vantage triple quadrupole mass spectrometer. Chromatogram of three 5 μL loop injections of 100 fg/μL Pierce Reserpine on a Hypersil GOLD aQ 2.1x20 mm Javelin column with isocratic 300 μL/min flow of 70% methanol: 30% water: 0.05% formic acid (upper panel).The SRM transitions monitored were 609.3-195.1 and 609.3-448.2 m/z with a 0.2 FWHM Q1 peak width (lower panel).
The TMT11plex Yeast Digest Standard can be used to assess instrument readiness and protein quantification accuracy. The TMT11plex Yeast Digest Standard was used to assess MS instrument readiness on the precision of quantification of a housekeeping protein, ENO2. Protein abundance was scaled to the parental lines in Proteome Discover 2.2 software, with expected ratios of 1:1. A total of 500 ng of the Pierce TMT11plex Yeast Digest Standard was measured using a 50 min gradient on an UltiMate 3000 RSLCnano system with a synchronous precursor selection (SPS)-MS3 (SPS-MS3) method of the Orbitrap Fusion Lumos Tribrid Mass Spectrometer.
Thermo Scientific Pierce HeLa Digest Standard base peak chromatogram. Chromatogram of 200 ng Pierce HeLa Protein Digest Standard separated using a Thermo Scientific Acclaim PepMap 100 3 μm x 75 μm x 15 cm column (Product # 160321) with a 2-35% gradient (A: 0.1% FA in water, B: 0.1% FA in 100% acetonitrile) at 300 nL/min for 120 minutes and detected on a Thermo Scientific LTQ Orbitrap XL Mass Spectrometer.
Example selected reaction monitoring (SRM) of 500fg reserpine on a Thermo Scientific Vantage triple quadrupole mass spectrometer. Chromatogram of three 5 μL loop injections of 100 fg/μL Pierce Reserpine on a Hypersil GOLD aQ 2.1x20 mm Javelin column with isocratic 300 μL/min flow of 70% methanol: 30% water: 0.05% formic acid (upper panel).The SRM transitions monitored were 609.3-195.1 and 609.3-448.2 m/z with a 0.2 FWHM Q1 peak width (lower panel).
For Research Use Only. Not for use in diagnostic procedures.