Search Thermo Fisher Scientific
当社では、様々なThermo Scientific™ 活性担体およびアクセサリをタンパク質、抗体、他の分子の固定用途でご用意しております。これらレジンは別々に、または便利なキットでご用意しています。
![]() | ![]() | ![]() | ![]() | ![]() | ![]() | |
---|---|---|---|---|---|---|
Pierce® NHS-Activated Agarose | AminoLink Coupling Resin | AminoLink Plus Coupling Resin | SulfoLink Coupling Resin | GlycoLink Coupling Resin | CarboxyLink Coupling | |
Target | -NH2 | -NH2 | -NH2 | -SH | -CHO | -COOH |
Support | 6% agarose (slurry or dry powder) | 4% agarose | 4% agarose, highly crosslinked | 6% agarose | Ultralink (polyacrylamide/ azlactone copolymer) | 4% agarose |
Coupling capacity | >25 mg/mL resin | 1–20 mg/mL resin | 20 mg/mL resin | 20 mg/mL resin | 1–10 mg/mL resin | 5 mg/mL resin |
Coupling time | 30 min | 4 hr | 4 hr | 2–3.5 hr | 30 min | 4 hr |
Recommended for | Proteins | Proteins, antibodies | Proteins, antibodies | Proteins, peptides, antibodies | Glycoproteins, polyclonal antibodies | Unmodified peptides |
Advantages | High coupling efficiency and capacity | High coupling efficiency | High coupling efficiency | Allows for gentle elution | Correctly orients antibodies during coupling | Flexible and gentle coupling conditions |
Disadvantages | Narrow pH range (7–9) | May be coupled at antigen-binding site | May be coupled at antigen-binding site | Protein must be reduced first | Not recommended for monoclonal antibodies | Not recommended for antibodies |
26196, 26197 (dry resin), 26200 (slurry) | 20381 (10 mL), 20382 (50 mL) | 20501 (10 mL), 20505 (50 mL) | 20401 (10 mL), 20402 (50 mL), 20404 (250 mL) | 53149 (10 mL) | 20266 (25 mL) |
pH 7.2でのカップリング手順を用いて固定した際、AminoLink Plus Resin 1 mL 上の様々なタンパク質に対する固定効率
タンパク質 | Protein (mg) reacted | Protein (mg) coupled | Coupling efficiency (%) |
---|---|---|---|
Protein G | 4.6 | 4.0 | 83 |
Mouse IgG | 4.7 | 4.5 | 96 |
Rat IgG | 4.7 | 4.4 | 93 |
GAM*-IgG | 0.9 | 0.8 | 84 |
Human IgG | 4.8 | 4.6 | 97 |
Human IgM | 0.9 | 0.8 | 93 |
*GAM-IgG is goat anti-mouse IgG, a secondary antibody.*GAM-IgGはヤギ抗マウスIgG、二次抗体です。
ディスポーザブルカラム、バッファ、カップリング試薬が入った便利なキット同様、追加で活性担体やアクセサリがございます。活性担体には、 磁気ビーズの形でご提供できるものもあります。
For Research Use Only. Not for use in diagnostic procedures.