Lipofectamine 3000 Transfection Reagent product photo
Lipofectamine 3000 Transfection Reagent delivers a high efficiency for hard-to-transfect cell lines. You can achieve over 70% transfection efficiency in a cost-effective way. Explore the benefits of Lipofectamine 3000 reagent:
  • Superior transfection efficiency—10-fold high efficiency into the broadest spectrum of difficult-to-transfect cells
  • Gentle with low toxicity—for improved cell viability
  • Great value—just pennies per reaction for best-in-class transfection results
Request a sample  Protocol library
 Download the DNA delivery flyer
Ordering information for Lipofectamine 3000 Reagent
Catalog # Name Size Price (USD) Qty
L3000001 Lipofectamine™ 3000 Transfection Reagent, 0.1 mL Each
85.65
Online Exclusive
89.75
Save 4.10 (5%)
L3000008 Lipofectamine™ 3000 Transfection Reagent, 0.75 mL Each
537.65
Online Exclusive
570.00
Save 32.35 (6%)
L3000015 Lipofectamine™ 3000 Transfection Reagent, 1.5 mL Each
836.65
Online Exclusive
894.00
Save 57.35 (6%)
L3000075 Lipofectamine™ 3000 Transfection Reagent, 5 x 1.5 mL Each
4,025.65
Online Exclusive
4,340.00
Save 314.35 (7%)
L3000150 Lipofectamine™ 3000 Transfection Reagent, 15 mL, 15 mL Each
7,760.00
Superior transfection performance across a range of hard-to-transfect cell line
When performing transfections in hard-to-transfect cell lines, Lipofectamine 3000 reagent can be an ideal choice. Lipofectamine 3000 reagent leverages our most advanced lipid nanoparticle technology to enable exceptional transfection performance and reproducible results. It delivers exceptional transfection efficiency with improved cell viability for the widest range of hard-to-transfect and common cell types including HEK 293, HeLa, LNCaP, HepG2, and A549 cell lines (Figure 1).
Fluorescent micrographs showing GFP transfection in cells transfected using Lipofectamine 3000 reagent, Lipofectamine 2000 reagent, and FuGene HD. Cells transfected with Lipofectamine 3000 reagent show the greatest GFP activity.
Figure 1. Lipofectamine 3000 reagent outperforms Invitrogen Lipofectamine 2000 and FuGENE HD reagents. Each reagent was used to transfect HEK 293, HeLa, LNCaP, HepG2, and A549 cell lines in a 96-well format, and GFP expression was analyzed 48 hours post-transfection. Lipofectamine 3000 reagent provided higher GFP transfection efficiency than Lipofectamine 2000 and FuGENE HD reagents for all five cell lines.
Figure 2. The Lipofectamine 3000 transfection protocol was developed to be easy to use, while still ensuring optimum performance and reliability in a wide panel of cell lines.
 Download the Lipofectamine 3000 reagent protocol
A gentle reagent with low transfection toxicity
All four steps in the transfection process were optimized and our most advanced lipid nanoparticle technologies were combined. This superior transfection performance allows you to reduce the reagent dose, thus helping lower toxicity risk when transfecting your cell line.
Graph comparing transfection efficiencies at increasing doses for each transfection reagent, Lipofectamine 2000, Lipofectamine 3000, and Lipofectamine LTX.
Figure 3. Each reagent was used to transfect HeLa cells in a 96-well format at the indicated doses with an emerald green fluorescent protein (GFP)-expressing vector. Analysis was performed 48 hours post transfection utilizing flow cytometry to determine percent transfection efficiency and the intensity of GFP expression. Lipofectamine 3000 reagent delivered higher efficiency and protein expression than both Lipofectamine 2000 reagent and Invitrogen Lipofectamine LTX Reagent.
A gentle reagent for CHO cell transfection
Chinese hamster ovary (CHO) is one of the most widely used non-human mammalian cell lines. They are easy to culture and are commonly used in transfection and large-scale production of therapeutic proteins. Lipofectamine 3000 reagent is a specially formulated gentle transfection reagent that is optimized for plasmid delivery into CHO cells with high efficiency and low cell toxicity. The reagent allows for transfection to be performed both in presence or absence of serum. The easy protocol, exceptional plasmid DNA delivery, and the ability to effectively generate stable cell lines makes Lipofectamine 3000 reagent, the perfect reagent for CHO cell transfection experiments.
Video Player is loading.
Current Time 0:00
Duration 2:58
Loaded: 0%
Stream Type LIVE
Remaining Time 2:58
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Increase transfection efficiency and reproducibility with Lipofectamine 3000 reagent
    Lipofectamine 3000 reviews by researchers
    Lipofectamine 3000 reviews by researchers
    See full list
    Comparison between Lipofectamine 3000 and Lipofectamine 2000 reagents
    Using the most biologically relevant cell lines helps provide the most relevant answers to your research questions. The exceptional cell spectrum and high transfection efficiency of Lipofectamine 3000 reagent enables excellent flexibility to use your cell line of choice in your studies.
    Compare Lipofectamine 3000 reagent and Lipofectamine 2000 reagent in a variety of cell lines using the comparison charts below. Find Lipofectamine 3000 reagent transfection efficiency in each cell line as well as fold protein expression improvement when using Lipofectamine 3000 over Lipofectamine 2000 reagent.
    Table 1. Lipofectamine 3000 reagent yields higher transfection efficiencies than Lipofectamine 2000 reagent when tested in a variety of cell lines
    Cell typeLipofectamine 3000 reagent transfection efficiencyFold protein expression improvement, Lipofectamine 3000 vs 2000 reagentCell type origin
    NIH/3T3Lipofectamine 3000 reagent transfection efficiency4Mouse embryonic fibroblast cell line
    4T1Lipofectamine 3000 reagent transfection efficiency2Mouse breast tumor cell line
    A431Lipofectamine 3000 reagent transfection efficiency2Human epidermoid carcinoma cell line
    A549Lipofectamine 3000 reagent transfection efficiency3Human lung cancer cell line
    ACHNLipofectamine 3000 reagent transfection efficiency2Human renal cancer cell line
    bEnd.3Lipofectamine 3000 reagent transfection efficiency9Mouse brain endothelioma cell line
    BJLipofectamine 3000 reagent transfection efficiency3Human skin fibroblast cell line
    BT-549Lipofectamine 3000 reagent transfection efficiency4Human breast cancer cell line
    C2C12Lipofectamine 3000 reagent transfection efficiency3Mouse myoblast cell line
    C6Lipofectamine 3000 reagent transfection efficiency5Rat glioma cell line
    Caco-2Lipofectamine 3000 reagent transfection efficiency2Human colon carcinoma cell line
    Caki-1Lipofectamine 3000 reagent transfection efficiency4Human kidney clear cell carcinoma cell line
    CHO-K1Lipofectamine 3000 reagent transfection efficiency1Chinese hamster ovary cell line
    CHO-SLipofectamine 3000 reagent transfection efficiency1Chinese hamster ovary cell line
    COLO 205Lipofectamine 3000 reagent transfection efficiency4Human colon cancer cell line
    COS-7Lipofectamine 3000 reagent transfection efficiency4Monkey fibroblast-like cell line
    Transfection efficiency (%): <30%  Transfection efficiency   30–50% Transfection efficiency   51–79% Transfection efficiency   >80% Transfection efficiency
    Cell typeLipofectamine 3000 reagent transfection efficiencyFold protein expression improvement, Lipofectamine 3000 vs 2000 reagentCell type origin
    DU 145Lipofectamine 3000 reagent transfection efficiency2Human prostate cancer cell line
    H460Lipofectamine 3000 reagent transfection efficiency3Human lung cancer cell line
    H9c2Lipofectamine 3000 reagent transfection efficiency3Rat cardiomyocyte cell line
    HCC1937Lipofectamine 3000 reagent transfection efficiency5Human breast cancer cell line
    HCT 116Lipofectamine 3000 reagent transfection efficiency1Human colon carcinoma cell line
    HEK-293Lipofectamine 3000 reagent transfection efficiency2Human embryonic kidney cell line
    HeLaLipofectamine 3000 reagent transfection efficiency3Human cervical cancer cell line
    Hep 3BLipofectamine 3000 reagent transfection efficiency2Human liver cancer cell line
    Hepa 1-6Lipofectamine 3000 reagent transfection efficiency1Mouse liver cancer cell line
    Hep G2Lipofectamine 3000 reagent transfection efficiency9Human liver cancer cell line
    Hs 578TLipofectamine 3000 reagent transfection efficiency3Human breast cancer cell line
    HT-1080Lipofectamine 3000 reagent transfection efficiency1Human fibrosarcoma cell line
    HT-29Lipofectamine 3000 reagent transfection efficiency1Human colon cancer cell line
    Huh-7Lipofectamine 3000 reagent transfection efficiency4Human hepatoma cell line
    JurkatLipofectamine 3000 reagent transfection efficiency1Human T lymphocyte cell line
    Transfection efficiency (%): <30%  Transfection efficiency   30–50% Transfection efficiency   51–79% Transfection efficiency   >80% Transfection efficiency
    Cell type (cont.)
    Lipofectamine 3000 reagent transfection efficiencyFold protein expression improvement, Lipofectamine 3000 vs 2000 reagentCell type origin
    K-562Lipofectamine 3000 reagent transfection efficiency1Human myelogenous leukemia cell line
    L6Lipofectamine 3000 reagent transfection efficiency8Rat skeletal muscle cell line
    L929Lipofectamine 3000 reagent transfection efficiency2Mouse fibrosarcoma cell line
    LNCaPLipofectamine 3000 reagent transfection efficiency6Human prostate cancer cell line
    MCF 10ALipofectamine 3000 reagent transfection efficiency5Human mammary epithelial cell line
    MCF7Lipofectamine 3000 reagent transfection efficiency2Human breast cancer cell line
    MDA-MB-231Lipofectamine 3000 reagent transfection efficiency3Human breast cancer cell line
    MDA-MB-453Lipofectamine 3000 reagent transfection efficiency1Human breast cancer cell line
    MDA-MB-468Lipofectamine 3000 reagent transfection efficiency9Human breast cancer cell line
    MDCKLipofectamine 3000 reagent transfection efficiency1Canine kidney cell line
    Neuro-2aLipofectamine 3000 reagent transfection efficiency1Mouse neuroblastoma cell line
    NCI-H23Lipofectamine 3000 reagent transfection efficiency2Human lung adenocarcinoma cell line
    NCI-H460Lipofectamine 3000 reagent transfection efficiency17Human lung cancer cell line
    P19Lipofectamine 3000 reagent transfection efficiency1Mouse embryonal carcinoma cell line
    PANC-1Lipofectamine 3000 reagent transfection efficiency3Human epithelioid carcinoma cell line
    PC-12Lipofectamine 3000 reagent transfection efficiency2Rat pheochromocytoma cell line
    Transfection efficiency (%): <30%  Transfection efficiency   30–50% Transfection efficiency   51–79% Transfection efficiency   >80% Transfection efficiency
    Cell type (cont.)
    Lipofectamine 3000 reagent transfection efficiencyFold protein expression improvement, Lipofectamine 3000 vs 2000 reagentCell type origin
    RAW 264.7Lipofectamine 3000 reagent transfection efficiency4Mouse macrophage cell line
    RBLLipofectamine 3000 reagent transfection efficiency2Rat basophil leukemia cells
    RDLipofectamine 3000 reagent transfection efficiency4Human rhabdomyosarcoma cell line
    Saos-2Lipofectamine 3000 reagent transfection efficiency4Human osteosarcoma cell line
    SH-SY5YLipofectamine 3000 reagent transfection efficiency1Human neuroblastoma cell line
    SK-BR-3Lipofectamine 3000 reagent transfection efficiency4Human breast cancer cell line
    SK-MEL-28Lipofectamine 3000 reagent transfection efficiency2Human melanoma cell line
    SK-N-SHLipofectamine 3000 reagent transfection efficiency6Human neuroblastoma cell line
    SK-OV-3Lipofectamine 3000 reagent transfection efficiency3Human ovarian carcinoma cell line
    SW480Lipofectamine 3000 reagent transfection efficiency2Human colon cancer cell line
    SW620Lipofectamine 3000 reagent transfection efficiency5Human colon cancer cell line
    T98GLipofectamine 3000 reagent transfection efficiency4Human glioblastoma cell line
    U2OSLipofectamine 3000 reagent transfection efficiency3Human osteosarcoma cell line
    U-937Lipofectamine 3000 reagent transfection efficiency2Human histiocytic leukemia cell line
    VeroLipofectamine 3000 reagent transfection efficiency1Monkey kidney cell line
    Transfection efficiency (%): <30%  Transfection efficiency   30–50% Transfection efficiency   51–79% Transfection efficiency   >80% Transfection efficiency
    Cell typeLipofectamine 3000 reagent transfection efficiencyFold protein expression improvement, Lipofectamine 3000 vs 2000 reagentCell type origin
    NIH/3T3Lipofectamine 3000 reagent transfection efficiency4Mouse embryonic fibroblast cell line
    4T1Lipofectamine 3000 reagent transfection efficiency2Mouse breast tumor cell line
    A431Lipofectamine 3000 reagent transfection efficiency2Human epidermoid carcinoma cell line
    A549Lipofectamine 3000 reagent transfection efficiency3Human lung cancer cell line
    ACHNLipofectamine 3000 reagent transfection efficiency2Human renal cancer cell line
    bEnd.3Lipofectamine 3000 reagent transfection efficiency9Mouse brain endothelioma cell line
    BJLipofectamine 3000 reagent transfection efficiency3Human skin fibroblast cell line
    BT-549Lipofectamine 3000 reagent transfection efficiency4Human breast cancer cell line
    C2C12Lipofectamine 3000 reagent transfection efficiency3Mouse myoblast cell line
    C6Lipofectamine 3000 reagent transfection efficiency5Rat glioma cell line
    Caco-2Lipofectamine 3000 reagent transfection efficiency2Human colon carcinoma cell line
    Caki-1Lipofectamine 3000 reagent transfection efficiency4Human kidney clear cell carcinoma cell line
    CHO-K1Lipofectamine 3000 reagent transfection efficiency1Chinese hamster ovary cell line
    CHO-SLipofectamine 3000 reagent transfection efficiency1Chinese hamster ovary cell line
    COLO 205Lipofectamine 3000 reagent transfection efficiency4Human colon cancer cell line
    COS-7Lipofectamine 3000 reagent transfection efficiency4Monkey fibroblast-like cell line
    Transfection efficiency (%): <30%  Transfection efficiency   30–50% Transfection efficiency   51–79% Transfection efficiency   >80% Transfection efficiency
    Cell typeLipofectamine 3000 reagent transfection efficiencyFold protein expression improvement, Lipofectamine 3000 vs 2000 reagentCell type origin
    DU 145Lipofectamine 3000 reagent transfection efficiency2Human prostate cancer cell line
    H460Lipofectamine 3000 reagent transfection efficiency3Human lung cancer cell line
    H9c2Lipofectamine 3000 reagent transfection efficiency3Rat cardiomyocyte cell line
    HCC1937Lipofectamine 3000 reagent transfection efficiency5Human breast cancer cell line
    HCT 116Lipofectamine 3000 reagent transfection efficiency1Human colon carcinoma cell line
    HEK-293Lipofectamine 3000 reagent transfection efficiency2Human embryonic kidney cell line
    HeLaLipofectamine 3000 reagent transfection efficiency3Human cervical cancer cell line
    Hep 3BLipofectamine 3000 reagent transfection efficiency2Human liver cancer cell line
    Hepa 1-6Lipofectamine 3000 reagent transfection efficiency1Mouse liver cancer cell line
    Hep G2Lipofectamine 3000 reagent transfection efficiency9Human liver cancer cell line
    Hs 578TLipofectamine 3000 reagent transfection efficiency3Human breast cancer cell line
    HT-1080Lipofectamine 3000 reagent transfection efficiency1Human fibrosarcoma cell line
    HT-29Lipofectamine 3000 reagent transfection efficiency1Human colon cancer cell line
    Huh-7Lipofectamine 3000 reagent transfection efficiency4Human hepatoma cell line
    JurkatLipofectamine 3000 reagent transfection efficiency1Human T lymphocyte cell line
    Transfection efficiency (%): <30%  Transfection efficiency   30–50% Transfection efficiency   51–79% Transfection efficiency   >80% Transfection efficiency
    Cell type (cont.)
    Lipofectamine 3000 reagent transfection efficiencyFold protein expression improvement, Lipofectamine 3000 vs 2000 reagentCell type origin
    K-562Lipofectamine 3000 reagent transfection efficiency1Human myelogenous leukemia cell line
    L6Lipofectamine 3000 reagent transfection efficiency8Rat skeletal muscle cell line
    L929Lipofectamine 3000 reagent transfection efficiency2Mouse fibrosarcoma cell line
    LNCaPLipofectamine 3000 reagent transfection efficiency6Human prostate cancer cell line
    MCF 10ALipofectamine 3000 reagent transfection efficiency5Human mammary epithelial cell line
    MCF7Lipofectamine 3000 reagent transfection efficiency2Human breast cancer cell line
    MDA-MB-231Lipofectamine 3000 reagent transfection efficiency3Human breast cancer cell line
    MDA-MB-453Lipofectamine 3000 reagent transfection efficiency1Human breast cancer cell line
    MDA-MB-468Lipofectamine 3000 reagent transfection efficiency9Human breast cancer cell line
    MDCKLipofectamine 3000 reagent transfection efficiency1Canine kidney cell line
    Neuro-2aLipofectamine 3000 reagent transfection efficiency1Mouse neuroblastoma cell line
    NCI-H23Lipofectamine 3000 reagent transfection efficiency2Human lung adenocarcinoma cell line
    NCI-H460Lipofectamine 3000 reagent transfection efficiency17Human lung cancer cell line
    P19Lipofectamine 3000 reagent transfection efficiency1Mouse embryonal carcinoma cell line
    PANC-1Lipofectamine 3000 reagent transfection efficiency3Human epithelioid carcinoma cell line
    PC-12Lipofectamine 3000 reagent transfection efficiency2Rat pheochromocytoma cell line
    Transfection efficiency (%): <30%  Transfection efficiency   30–50% Transfection efficiency   51–79% Transfection efficiency   >80% Transfection efficiency
    Cell type (cont.)
    Lipofectamine 3000 reagent transfection efficiencyFold protein expression improvement, Lipofectamine 3000 vs 2000 reagentCell type origin
    RAW 264.7Lipofectamine 3000 reagent transfection efficiency4Mouse macrophage cell line
    RBLLipofectamine 3000 reagent transfection efficiency2Rat basophil leukemia cells
    RDLipofectamine 3000 reagent transfection efficiency4Human rhabdomyosarcoma cell line
    Saos-2Lipofectamine 3000 reagent transfection efficiency4Human osteosarcoma cell line
    SH-SY5YLipofectamine 3000 reagent transfection efficiency1Human neuroblastoma cell line
    SK-BR-3Lipofectamine 3000 reagent transfection efficiency4Human breast cancer cell line
    SK-MEL-28Lipofectamine 3000 reagent transfection efficiency2Human melanoma cell line
    SK-N-SHLipofectamine 3000 reagent transfection efficiency6Human neuroblastoma cell line
    SK-OV-3Lipofectamine 3000 reagent transfection efficiency3Human ovarian carcinoma cell line
    SW480Lipofectamine 3000 reagent transfection efficiency2Human colon cancer cell line
    SW620Lipofectamine 3000 reagent transfection efficiency5Human colon cancer cell line
    T98GLipofectamine 3000 reagent transfection efficiency4Human glioblastoma cell line
    U2OSLipofectamine 3000 reagent transfection efficiency3Human osteosarcoma cell line
    U-937Lipofectamine 3000 reagent transfection efficiency2Human histiocytic leukemia cell line
    VeroLipofectamine 3000 reagent transfection efficiency1Monkey kidney cell line
    Transfection efficiency (%): <30%  Transfection efficiency   30–50% Transfection efficiency   51–79% Transfection efficiency   >80% Transfection efficiency
    Enhance your cancer research
    For exceptional performance, utilize Lipofectamine 3000 to enhance transfection in your cancer research experiments. This advanced and versatile reagent works well for transfection in breast, lung, prostate, and liver cancer cells. There are step-by-step protocols to help ensure high transfection efficiency with low toxicity when a specific cancer cell line is coupled with Lipofectamine 3000 transfection reagent.
    Lipofectamine 3000 reagent demonstrates significantly higher transfection performance compared to leading transfection reagents, such as Lipofectamine 2000 reagent, in a panel of cancer cells derived from various tissues. While only 25% of the selected cancer cell line panel would be considered easy to transfect (>50% transfection efficiency) with Lipofectamine 2000 reagent, 60% of the panel is easy to transfect with Lipofectamine 3000 reagent.
    Graph demonstrating improved transfection efficiencies for Lipofectamine 3000 compared to Lipofectamine 2000 across 17 distinct cell lines.
    Figure 4. Lipofectamine 2000 reagent and Lipofectamine 3000 reagent were used to transfect 17 cell lines with a GFP-expressing plasmid in a 24-well plate format, using 0.5 µg plasmid/well and the recommended protocols for each reagent. GFP expression was analyzed 48 hours post-transfection. Each condition was tested in triplicate, and the data points show the mean transfection efficiency + SD.
    Transfect stem cells or generate induced pluripotent stem cells (iPSCs) with Lipofectamine 3000 reagent
    The exceptional transfection efficiency of Lipofectamine 3000 reagent in conjunction with Invitrogen Epi5 Episomal iPSC Reprogramming Kit enables highly efficient reprogramming of somatic cells without the need for electroporation.
    Figure 5. Transfection of stem cells. (A) H9 ESCs or (B) iPSCs were transfected using Lipofectamine 3000 reagent. Cells were visualized by fluorescence microscopy and processed using flow cytometry to determine transfection efficiency.
    Panel of fluorescent and brightfield micrographs demonstrating transfection efficiency of GFP in H9 ESCs(image A) and iPSCs (image B).
    Figure 6. Reprogramming efficiency of Lipofectamine 3000 reagent compared to electroporation. BJ fibroblasts as well as neonatal (HDFn) and adult (HDFa) human dermal fibroblasts were reprogrammed to iPSCs by transfection of Epi5 vectors using either Lipofectamine 3000 reagent or the Invitrogen Neon Transfection System. Colonies were (A) visualized by brightfield microscopy and (B) stained for alkaline phosphatase.
    Lipofectamine 3000 reagent helps improve genome editing outcomes
    Lipofectamine 3000 reagent helps break through the boundaries of delivery and facilitates new technologies, such as genome engineering, in more biologically relevant systems. With this reagent, TALENs and CRISPR sequences target the AAVS1 locus in HepG2 and U2OS cells, and show improved transfection efficiency, mean fluorescence intensity, and genomic cleavage. These advancements in delivery help minimize painstaking downstream workflows, enable easier stem cell manipulation, and enhance site-specific insertion of transgenes into the cellular genome.
     Download Lipofectamine 3000 application note
    Figure 7. Cleavage efficiency of GeneArt TALs and CRISPRs. The TALs and CRISPRs targeted the AAVS1 locus in (A) U2OS and (B) HepG2 cell lines. Cleavage was assayed using a Invitrogen GeneArt Genomic Cleavage Detection Kit.
    Two panels of gel images and corresponding bar graphs demonstrating cleavage and gene editing efficiency when comparing Lipofectamine 3000 to Lipofectamine 2000 for delivery of gene editing tools like TALEN and CRISPR sequences.
    Figure 8. Transfection efficiency and protein expression using a CRISPR vector. The vector contained an OFP reporter gene and was transfected with Lipofectamine 2000 or Lipofectamine 3000 reagent into (A) U2OS and (B) HepG2 cell lines. Bar graphs show reporter gene expression; images show fluorescence of corresponding cells expressing OFP.
    leaf icon Sustainable packaging
    Packaging must protect the product inside—but that doesn’t have to mean sacrificing sustainability. Lipofectamine products are shipped at ambient temperature, diverting 24,000 ft3 of Expanded Polystyrene (EPS) coolers from landfills annually. We reduce the carbon footprint from packaging and shipping these products by 80 tons (CO2 equivalents).
    Learn more about sustainable solutions
     Download the green fact sheet
    Resources
    Our Lipofectamine reagent–specific protocols have been optimized for efficiency, viability, and reproducibility across a broad range of cell types. Search this library to find protocols and citations curated to fit your experimental needs. Filter by cell type, payload, and product of choice to easily unlock your answers.
    Explore protocols and citations
    For Research Use Only. Not for use in diagnostic procedures.
    Stylesheet for Classic Wide Template adjustments