decorative
Relative fluorescent quantitation (or quantitative fluorescence PCR (QF-PCR)) is a technique used in a variety of fragment analysis applications that requires accurate peak height comparisons across multiple samples. Applications that utilize this technique include screening for loss of heterozygosity (LOH) using microsatellites or SNPs, aneuploidy assays, and detecting large chromosomal deletions.
Electropherogram of a microsatellite marker in DNA
Figure 1. Detecting Loss of Heterozygosity (LOH). Electropherogram of a microsatellite marker in DNA from a healthy (top panel) and tumor sample (bottom panel) run on a Life Technologies 3130 Series Genetic Analyzer. The reduced peak height in the tumor sample indicates potential LOH in the sample.  Click image to enlarge
Step-by-Step Guide to Relative Fluorescent Quantification
DNA Purification kit
DNA extraction is a critical first step in the experimental workflow of DNA Sequencing and Fragment analysis. The overall quality, accuracy and length of the DNA sequence read can be significantly affected by characteristics of the sample itself, and the method chosen for nucleic acid extraction. Ideal methods will vary depending on the source or tissue type, how it was obtained from its source, and how the sample was handled or stored prior to extraction.
Recommended Products: DNA Isolation
PCR Kit
Amplify microsatellite loci of interest by PCR using fluorescent labeled primer
Recommended Products: Design Primer and Perform PCR
Fragment Analysis sizing graph image.
Add aliquot of PCR product to size standard and Hi-Di Formamide mix; heat denature and proceed with electrophoresis.
Recommended Products: Prepare Sample for Analysis
Sizing Information
To learn more about our GeneScan Size standards used in sizing experiments please see our table below.
When DNA fragments are labeled with:Choose a size standards labeled with:Recommended Size StandardPossible ApplicationsOther Kits and Products
dR110, dR6G, dTAMRA™, dROX (Dye Set DS-02 - Filter E5)LIZ®
SNaPshot®
Microsatellite, LOH, ISSR, Chimerism, ISSR, RFLP, T-RFLP, MSMSA
5-FAM™, HEX™, NED™ (Dye Set DS-30 - Filter D)ROX™GeneScan™ 500 ROX™ Size StandardCustom Fragment AnalysisCustom Labeled Primer
6-FAM™, VIC®, NED™ (Dye Set DS-31 - Filter D**)ROX™GeneScan™ 500 ROX™ Size Standard
Custom Fragment Analysis
Custom Labeled Primer
5-FAM™, JOE, NED™ (Dye Set DS-32 - Filter F)ROX™GeneScan™ 500 ROX™ Size Standard
Microsatellite (Forensic)
AFLP
6-FAM™, VIC®, NED™, PET® (Dye Set DS-33 - Filter G5)LIZ®GeneScan™ 600 LIZ Size Standard v2.0
Microsatellite (Forensic)
Custom Fragment Analysis
Capillary Electrophoresis
During capillary electrophoresis, the products of the PCR are injected electrokinetically into capillaries filled with polymer. High voltage is applied so that the fluorescent DNA fragments are separated by size and are detected by a laser/camera system.
Which Electrophoresis Instrument (Genetic Analyzer) Is Right for You?
 Instrument
 3730xl
3730xL
3730
3730
3500xl
3500xL
3500
3500
3130xl
3130xL
3130
3130
310
310
Number of Capillaries96482481641
Compatible Applications:
(S) Supported; (A) AB Demonstrated; (C) Customer Demonstrated; (N) Not Supported
Relative Fluorescent Quantitation
- Loss of Heterozygosity (LOH)SSSSSSS
Analyze the microsatellite loci size and height.
Learn more
Which Data Analysis Software Is Right for You?
DNA Purification kit
DNA extraction is a critical first step in the experimental workflow of DNA Sequencing and Fragment analysis. The overall quality, accuracy and length of the DNA sequence read can be significantly affected by characteristics of the sample itself, and the method chosen for nucleic acid extraction. Ideal methods will vary depending on the source or tissue type, how it was obtained from its source, and how the sample was handled or stored prior to extraction.
Recommended Products: DNA Isolation
PCR Kit
Amplify microsatellite loci of interest by PCR using fluorescent labeled primer
Recommended Products: Design Primer and Perform PCR
Fragment Analysis sizing graph image.
Add aliquot of PCR product to size standard and Hi-Di Formamide mix; heat denature and proceed with electrophoresis.
Recommended Products: Prepare Sample for Analysis
Sizing Information
To learn more about our GeneScan Size standards used in sizing experiments please see our table below.
When DNA fragments are labeled with:Choose a size standards labeled with:Recommended Size StandardPossible ApplicationsOther Kits and Products
dR110, dR6G, dTAMRA™, dROX (Dye Set DS-02 - Filter E5)LIZ®
SNaPshot®
Microsatellite, LOH, ISSR, Chimerism, ISSR, RFLP, T-RFLP, MSMSA
5-FAM™, HEX™, NED™ (Dye Set DS-30 - Filter D)ROX™GeneScan™ 500 ROX™ Size StandardCustom Fragment AnalysisCustom Labeled Primer
6-FAM™, VIC®, NED™ (Dye Set DS-31 - Filter D**)ROX™GeneScan™ 500 ROX™ Size Standard
Custom Fragment Analysis
Custom Labeled Primer
5-FAM™, JOE, NED™ (Dye Set DS-32 - Filter F)ROX™GeneScan™ 500 ROX™ Size Standard
Microsatellite (Forensic)
AFLP
6-FAM™, VIC®, NED™, PET® (Dye Set DS-33 - Filter G5)LIZ®GeneScan™ 600 LIZ Size Standard v2.0
Microsatellite (Forensic)
Custom Fragment Analysis
Capillary Electrophoresis
During capillary electrophoresis, the products of the PCR are injected electrokinetically into capillaries filled with polymer. High voltage is applied so that the fluorescent DNA fragments are separated by size and are detected by a laser/camera system.
Which Electrophoresis Instrument (Genetic Analyzer) Is Right for You?
 Instrument
 3730xl
3730xL
3730
3730
3500xl
3500xL
3500
3500
3130xl
3130xL
3130
3130
310
310
Number of Capillaries96482481641
Compatible Applications:
(S) Supported; (A) AB Demonstrated; (C) Customer Demonstrated; (N) Not Supported
Relative Fluorescent Quantitation
- Loss of Heterozygosity (LOH)SSSSSSS
Analyze the microsatellite loci size and height.
Learn more
Which Data Analysis Software Is Right for You?
Step-by-Step Guide to Relative Fluorescent Quantification
DNA Purification kit
DNA extraction is a critical first step in the experimental workflow of DNA Sequencing and Fragment analysis. The overall quality, accuracy and length of the DNA sequence read can be significantly affected by characteristics of the sample itself, and the method chosen for nucleic acid extraction. Ideal methods will vary depending on the source or tissue type, how it was obtained from its source, and how the sample was handled or stored prior to extraction.
Recommended Products: DNA Isolation
PCR Kit
Amplify microsatellite loci of interest by PCR using fluorescent labeled primer
Recommended Products: Design Primer and Perform PCR
Fragment Analysis sizing graph image.
Add aliquot of PCR product to size standard and Hi-Di Formamide mix; heat denature and proceed with electrophoresis.
Recommended Products: Prepare Sample for Analysis
Sizing Information
To learn more about our GeneScan Size standards used in sizing experiments please see our table below.
When DNA fragments are labeled with:Choose a size standards labeled with:Recommended Size StandardPossible ApplicationsOther Kits and Products
dR110, dR6G, dTAMRA™, dROX (Dye Set DS-02 - Filter E5)LIZ®
SNaPshot®
Microsatellite, LOH, ISSR, Chimerism, ISSR, RFLP, T-RFLP, MSMSA
5-FAM™, HEX™, NED™ (Dye Set DS-30 - Filter D)ROX™GeneScan™ 500 ROX™ Size StandardCustom Fragment AnalysisCustom Labeled Primer
6-FAM™, VIC®, NED™ (Dye Set DS-31 - Filter D**)ROX™GeneScan™ 500 ROX™ Size Standard
Custom Fragment Analysis
Custom Labeled Primer
5-FAM™, JOE, NED™ (Dye Set DS-32 - Filter F)ROX™GeneScan™ 500 ROX™ Size Standard
Microsatellite (Forensic)
AFLP
6-FAM™, VIC®, NED™, PET® (Dye Set DS-33 - Filter G5)LIZ®GeneScan™ 600 LIZ Size Standard v2.0
Microsatellite (Forensic)
Custom Fragment Analysis
Capillary Electrophoresis
During capillary electrophoresis, the products of the PCR are injected electrokinetically into capillaries filled with polymer. High voltage is applied so that the fluorescent DNA fragments are separated by size and are detected by a laser/camera system.
Which Electrophoresis Instrument (Genetic Analyzer) Is Right for You?
 Instrument
 3730xl
3730xL
3730
3730
3500xl
3500xL
3500
3500
3130xl
3130xL
3130
3130
310
310
Number of Capillaries96482481641
Compatible Applications:
(S) Supported; (A) AB Demonstrated; (C) Customer Demonstrated; (N) Not Supported
Relative Fluorescent Quantitation
- Loss of Heterozygosity (LOH)SSSSSSS
Analyze the microsatellite loci size and height.
Learn more
Which Data Analysis Software Is Right for You?
DNA Purification kit
DNA extraction is a critical first step in the experimental workflow of DNA Sequencing and Fragment analysis. The overall quality, accuracy and length of the DNA sequence read can be significantly affected by characteristics of the sample itself, and the method chosen for nucleic acid extraction. Ideal methods will vary depending on the source or tissue type, how it was obtained from its source, and how the sample was handled or stored prior to extraction.
Recommended Products: DNA Isolation
PCR Kit
Amplify microsatellite loci of interest by PCR using fluorescent labeled primer
Recommended Products: Design Primer and Perform PCR
Fragment Analysis sizing graph image.
Add aliquot of PCR product to size standard and Hi-Di Formamide mix; heat denature and proceed with electrophoresis.
Recommended Products: Prepare Sample for Analysis
Sizing Information
To learn more about our GeneScan Size standards used in sizing experiments please see our table below.
When DNA fragments are labeled with:Choose a size standards labeled with:Recommended Size StandardPossible ApplicationsOther Kits and Products
dR110, dR6G, dTAMRA™, dROX (Dye Set DS-02 - Filter E5)LIZ®
SNaPshot®
Microsatellite, LOH, ISSR, Chimerism, ISSR, RFLP, T-RFLP, MSMSA
5-FAM™, HEX™, NED™ (Dye Set DS-30 - Filter D)ROX™GeneScan™ 500 ROX™ Size StandardCustom Fragment AnalysisCustom Labeled Primer
6-FAM™, VIC®, NED™ (Dye Set DS-31 - Filter D**)ROX™GeneScan™ 500 ROX™ Size Standard
Custom Fragment Analysis
Custom Labeled Primer
5-FAM™, JOE, NED™ (Dye Set DS-32 - Filter F)ROX™GeneScan™ 500 ROX™ Size Standard
Microsatellite (Forensic)
AFLP
6-FAM™, VIC®, NED™, PET® (Dye Set DS-33 - Filter G5)LIZ®GeneScan™ 600 LIZ Size Standard v2.0
Microsatellite (Forensic)
Custom Fragment Analysis
Capillary Electrophoresis
During capillary electrophoresis, the products of the PCR are injected electrokinetically into capillaries filled with polymer. High voltage is applied so that the fluorescent DNA fragments are separated by size and are detected by a laser/camera system.
Which Electrophoresis Instrument (Genetic Analyzer) Is Right for You?
 Instrument
 3730xl
3730xL
3730
3730
3500xl
3500xL
3500
3500
3130xl
3130xL
3130
3130
310
310
Number of Capillaries96482481641
Compatible Applications:
(S) Supported; (A) AB Demonstrated; (C) Customer Demonstrated; (N) Not Supported
Relative Fluorescent Quantitation
- Loss of Heterozygosity (LOH)SSSSSSS
Analyze the microsatellite loci size and height.
Learn more
Which Data Analysis Software Is Right for You?
Highlighted products for Fluorescent Quantification:
3500 Series Genetic Analyzers
GeneMapper® Software
Verti Thermal Cycler
Visistat reference component
For Research Use Only. Not for use in diagnostic procedures.