showcase.par.77081.image.200.150.1.sscp
Single-stranded conformation polymorphism (SSCP) analysis is a widely used screening method that allows you to identify different genomic variants in a large number of samples and in a broad range of organisms, from microorganisms to humans.
SSCP analysis detects sequence variations (single-point mutations and other small-scale changes) through electrophoretic mobility differences. DNA that contains a sequence mutation (even a single base pair change) has a measurable mobility difference compared to wild type DNA when subjected to nondenaturing (or partially denaturing) conditions.
Step-by-Step Guide to Single-Strand Polymorphism
DNA Purification kit
DNA extraction is a critical first step in the experimental workflow of DNA Sequencing and Fragment analysis. The overall quality, accuracy and length of the DNA sequence read can be significantly affected by characteristics of the sample itself, and the method chosen for nucleic acid extraction. Ideal methods will vary depending on the source or tissue type, how it was obtained from its source, and how the sample was handled or stored prior to extraction.
Recommended Products: DNA Isolation
PCR Kit
Amplify region of interest by PCR using fluorescent labeled primer.
Recommended Products: Perform PCR
Fragment Analysis sizing graph image.
Add aliquot of PCR product to size standard and Hi-Di Formamide mix; heat denature and proceed with electrophoresis.
Recommended Products: Prepare Sample for Analysis
Sizing Information
To learn more about our GeneScan Size standards used in sizing experiments please see our table below.
When DNA fragments are labeled with:Choose a size standards labeled with:Recommended Size StandardPossible ApplicationsOther Kits and Products
dR110, dR6G, dTAMRA™, dROX (Dye Set DS-02 - Filter E5)LIZ®
SNaPshot®
Microsatellite, LOH, ISSR, Chimerism, ISSR, RFLP, T-RFLP, MSMSA
5-FAM™, HEX™, NED™ (Dye Set DS-30 - Filter D)ROX™GeneScan™ 500 ROX™ Size StandardCustom Fragment AnalysisCustom Labeled Primer
6-FAM™, VIC®, NED™ (Dye Set DS-31 - Filter D**)ROX™GeneScan™ 500 ROX™ Size Standard
Custom Fragment Analysis
Custom Labeled Primer
5-FAM™, JOE, NED™ (Dye Set DS-32 - Filter F)ROX™GeneScan™ 500 ROX™ Size Standard
Microsatellite (Forensic)
6-FAM™, VIC®, NED™, PET® (Dye Set DS-33 - Filter G5)LIZ®GeneScan™ 600 LIZ Size Standard v2.0
Microsatellite (Forensic)
Custom Fragment Analysis
Capillary Electrophoresis
During capillary electrophoresis, the products of the PCR are injected electrokinetically into capillaries filled with polymer. High voltage is applied so that the fluorescent DNA fragments are separated by size and are detected by a laser/camera system.
Which Polymer & Electrophoresis Instrument (Genetic Analyzer) Is Right for You?
Polymer:
  Instrument
 3730xl
3730xL
3730
3730
3500xl
3500xL
3500
3500
3130xl
3130xL
3130
3130
310
310
Number of Capillaries96482481641
Compatible Applications:
(S) Supported; (A) AB Demonstrated; (C) Customer Demonstrated; (N) Not Supported
Conformation Analysis
- SSCPNNNNAAS
Analyze the SSCP profile.
Which Data Analysis Software Is Right for You?
DNA Purification kit
DNA extraction is a critical first step in the experimental workflow of DNA Sequencing and Fragment analysis. The overall quality, accuracy and length of the DNA sequence read can be significantly affected by characteristics of the sample itself, and the method chosen for nucleic acid extraction. Ideal methods will vary depending on the source or tissue type, how it was obtained from its source, and how the sample was handled or stored prior to extraction.
Recommended Products: DNA Isolation
PCR Kit
Amplify region of interest by PCR using fluorescent labeled primer.
Recommended Products: Perform PCR
Fragment Analysis sizing graph image.
Add aliquot of PCR product to size standard and Hi-Di Formamide mix; heat denature and proceed with electrophoresis.
Recommended Products: Prepare Sample for Analysis
Sizing Information
To learn more about our GeneScan Size standards used in sizing experiments please see our table below.
When DNA fragments are labeled with:Choose a size standards labeled with:Recommended Size StandardPossible ApplicationsOther Kits and Products
dR110, dR6G, dTAMRA™, dROX (Dye Set DS-02 - Filter E5)LIZ®
SNaPshot®
Microsatellite, LOH, ISSR, Chimerism, ISSR, RFLP, T-RFLP, MSMSA
5-FAM™, HEX™, NED™ (Dye Set DS-30 - Filter D)ROX™GeneScan™ 500 ROX™ Size StandardCustom Fragment AnalysisCustom Labeled Primer
6-FAM™, VIC®, NED™ (Dye Set DS-31 - Filter D**)ROX™GeneScan™ 500 ROX™ Size Standard
Custom Fragment Analysis
Custom Labeled Primer
5-FAM™, JOE, NED™ (Dye Set DS-32 - Filter F)ROX™GeneScan™ 500 ROX™ Size Standard
Microsatellite (Forensic)
6-FAM™, VIC®, NED™, PET® (Dye Set DS-33 - Filter G5)LIZ®GeneScan™ 600 LIZ Size Standard v2.0
Microsatellite (Forensic)
Custom Fragment Analysis
Capillary Electrophoresis
During capillary electrophoresis, the products of the PCR are injected electrokinetically into capillaries filled with polymer. High voltage is applied so that the fluorescent DNA fragments are separated by size and are detected by a laser/camera system.
Which Polymer & Electrophoresis Instrument (Genetic Analyzer) Is Right for You?
Polymer:
  Instrument
 3730xl
3730xL
3730
3730
3500xl
3500xL
3500
3500
3130xl
3130xL
3130
3130
310
310
Number of Capillaries96482481641
Compatible Applications:
(S) Supported; (A) AB Demonstrated; (C) Customer Demonstrated; (N) Not Supported
Conformation Analysis
- SSCPNNNNAAS
Analyze the SSCP profile.
Which Data Analysis Software Is Right for You?
Step-by-Step Guide to Single-Strand Polymorphism
DNA Purification kit
DNA extraction is a critical first step in the experimental workflow of DNA Sequencing and Fragment analysis. The overall quality, accuracy and length of the DNA sequence read can be significantly affected by characteristics of the sample itself, and the method chosen for nucleic acid extraction. Ideal methods will vary depending on the source or tissue type, how it was obtained from its source, and how the sample was handled or stored prior to extraction.
Recommended Products: DNA Isolation
PCR Kit
Amplify region of interest by PCR using fluorescent labeled primer.
Recommended Products: Perform PCR
Fragment Analysis sizing graph image.
Add aliquot of PCR product to size standard and Hi-Di Formamide mix; heat denature and proceed with electrophoresis.
Recommended Products: Prepare Sample for Analysis
Sizing Information
To learn more about our GeneScan Size standards used in sizing experiments please see our table below.
When DNA fragments are labeled with:Choose a size standards labeled with:Recommended Size StandardPossible ApplicationsOther Kits and Products
dR110, dR6G, dTAMRA™, dROX (Dye Set DS-02 - Filter E5)LIZ®
SNaPshot®
Microsatellite, LOH, ISSR, Chimerism, ISSR, RFLP, T-RFLP, MSMSA
5-FAM™, HEX™, NED™ (Dye Set DS-30 - Filter D)ROX™GeneScan™ 500 ROX™ Size StandardCustom Fragment AnalysisCustom Labeled Primer
6-FAM™, VIC®, NED™ (Dye Set DS-31 - Filter D**)ROX™GeneScan™ 500 ROX™ Size Standard
Custom Fragment Analysis
Custom Labeled Primer
5-FAM™, JOE, NED™ (Dye Set DS-32 - Filter F)ROX™GeneScan™ 500 ROX™ Size Standard
Microsatellite (Forensic)
6-FAM™, VIC®, NED™, PET® (Dye Set DS-33 - Filter G5)LIZ®GeneScan™ 600 LIZ Size Standard v2.0
Microsatellite (Forensic)
Custom Fragment Analysis
Capillary Electrophoresis
During capillary electrophoresis, the products of the PCR are injected electrokinetically into capillaries filled with polymer. High voltage is applied so that the fluorescent DNA fragments are separated by size and are detected by a laser/camera system.
Which Polymer & Electrophoresis Instrument (Genetic Analyzer) Is Right for You?
Polymer:
  Instrument
 3730xl
3730xL
3730
3730
3500xl
3500xL
3500
3500
3130xl
3130xL
3130
3130
310
310
Number of Capillaries96482481641
Compatible Applications:
(S) Supported; (A) AB Demonstrated; (C) Customer Demonstrated; (N) Not Supported
Conformation Analysis
- SSCPNNNNAAS
Analyze the SSCP profile.
Which Data Analysis Software Is Right for You?
DNA Purification kit
DNA extraction is a critical first step in the experimental workflow of DNA Sequencing and Fragment analysis. The overall quality, accuracy and length of the DNA sequence read can be significantly affected by characteristics of the sample itself, and the method chosen for nucleic acid extraction. Ideal methods will vary depending on the source or tissue type, how it was obtained from its source, and how the sample was handled or stored prior to extraction.
Recommended Products: DNA Isolation
PCR Kit
Amplify region of interest by PCR using fluorescent labeled primer.
Recommended Products: Perform PCR
Fragment Analysis sizing graph image.
Add aliquot of PCR product to size standard and Hi-Di Formamide mix; heat denature and proceed with electrophoresis.
Recommended Products: Prepare Sample for Analysis
Sizing Information
To learn more about our GeneScan Size standards used in sizing experiments please see our table below.
When DNA fragments are labeled with:Choose a size standards labeled with:Recommended Size StandardPossible ApplicationsOther Kits and Products
dR110, dR6G, dTAMRA™, dROX (Dye Set DS-02 - Filter E5)LIZ®
SNaPshot®
Microsatellite, LOH, ISSR, Chimerism, ISSR, RFLP, T-RFLP, MSMSA
5-FAM™, HEX™, NED™ (Dye Set DS-30 - Filter D)ROX™GeneScan™ 500 ROX™ Size StandardCustom Fragment AnalysisCustom Labeled Primer
6-FAM™, VIC®, NED™ (Dye Set DS-31 - Filter D**)ROX™GeneScan™ 500 ROX™ Size Standard
Custom Fragment Analysis
Custom Labeled Primer
5-FAM™, JOE, NED™ (Dye Set DS-32 - Filter F)ROX™GeneScan™ 500 ROX™ Size Standard
Microsatellite (Forensic)
6-FAM™, VIC®, NED™, PET® (Dye Set DS-33 - Filter G5)LIZ®GeneScan™ 600 LIZ Size Standard v2.0
Microsatellite (Forensic)
Custom Fragment Analysis
Capillary Electrophoresis
During capillary electrophoresis, the products of the PCR are injected electrokinetically into capillaries filled with polymer. High voltage is applied so that the fluorescent DNA fragments are separated by size and are detected by a laser/camera system.
Which Polymer & Electrophoresis Instrument (Genetic Analyzer) Is Right for You?
Polymer:
  Instrument
 3730xl
3730xL
3730
3730
3500xl
3500xL
3500
3500
3130xl
3130xL
3130
3130
310
310
Number of Capillaries96482481641
Compatible Applications:
(S) Supported; (A) AB Demonstrated; (C) Customer Demonstrated; (N) Not Supported
Conformation Analysis
- SSCPNNNNAAS
Analyze the SSCP profile.
Which Data Analysis Software Is Right for You?
Highlighted products for SSCP Analysis :
GeneMapper® Software
Verti Thermal Cycler
Visistat reference component
For Research Use Only. Not for use in diagnostic procedures.