ion-torrent.par.18475
Make ends meet
Create libraries for all of your DNA sequencing needs with our range of solutions. Simple and fast kits are available for various DNA fragmentation methods, for automation of library preparation and for normalization of library prior to template preparation.
Ion DNA Fragment Library Kits
The Ion Xpress™ Plus Fragment Library Kit for enzymatic shearing
The Ion Xpress™ Plus Fragment Library kit provides the reagents required for creation of high-quality DNA fragment libraries including the Ion Shear™ reagent for enzymatic fragmentation. The kit offers: 
  • Industry-leading coverage uniformity and recovery rate for library construction
  • Rapid and flexible enzymatic library construction methodology
  • Optimized use with both the Ion PGM™ and the Ion Proton™ Systems
  • Fast workflow, with completion of library preparation in as little as 2 hours
  • No physical shearing device requirement, saving time and cost
Catalog # Name Size Price (USD) Qty
4471269 Ion Xpress™ Plus Fragment Library Kit Each
1,312.00
4474517 Ion Xpress™ Barcode Adapters 1-96 Kit Each
13,550.00
The Ion Xpress™ Plus Fragment Library Kit for enzymatic shearing
The Ion Xpress™ Plus Fragment Library kit provides the reagents required for creation of high-quality DNA fragment libraries including the Ion Shear™ reagent for enzymatic fragmentation. The kit offers: 
  • Industry-leading coverage uniformity and recovery rate for library construction
  • Rapid and flexible enzymatic library construction methodology
  • Optimized use with both the Ion PGM™ and the Ion Proton™ Systems
  • Fast workflow, with completion of library preparation in as little as 2 hours
  • No physical shearing device requirement, saving time and cost
Catalog # Name Size Price (USD) Qty
4471269 Ion Xpress™ Plus Fragment Library Kit Each
1,312.00
4474517 Ion Xpress™ Barcode Adapters 1-96 Kit Each
13,550.00
Ion Library Equalizer™ Kit for library normalization
Use the Ion Library Equalizer™ Kit to normalize your Ion DNA libraries without the need for dilution or quantification, or the need for any additional equipment.
The Ion Library Equalizer™ Kit offers: 
  • Compatibility with Ion DNA Fragment Library Kits and Ion AmpliSeq™ Target Selection Technology
  • A cost-effective solution with reduced hands-on time and pipetting steps
  • Flexibility to process a single library or many libraries with automation and barcode compatibility
Catalog # Name Size Price (USD) Qty
4482298 Ion Library Equalizer™ Kit Each
914.00
Life Technologies now offers transposon-based library prep options for your lab. Learn more about Thermo Scientific® MuSeek™ Library Preparation Kit
Third-party library preparation products
Life Technologies also offers a range of third-party library preparation products for you to choose from.
For Research Use Only. Not for use in diagnostic procedures.