48ab - Appl/Samples/Products/Resources/Contact
HRTEM
Transmission and scanning transmission electron microscopes (S/TEM) are invaluable tools for the characterization of nanostructures, providing a range of different imaging modes, as well as access to information on elemental composition and electronic structure with high sensitivity. As materials research progressively begins to focus more and more on optimizing material function and behavior at the nanoscale, accurate information at this resolution becomes increasingly essential. High-resolution TEM (HRTEM) and STEM (HRSTEM) deliver the most detailed structural information possible in order to fundamentally characterize samples down to their atomic organization. This is invaluable to nanomaterials research as minor structural inconsistencies and variations can result in substantial changes to the properties of the material. For example, the crystal structure and atomic spacing of the atoms in platinum nanoparticles can have a drastic impact on their catalytic behavior in hydrogen fuel cells.
HRTEM microscopy
Thermo Fisher Scientific offers hardware and software innovations for HRTEM and HRSTEM analysis of a broad range of samples, including beam-sensitive materials. In particular, image quality is often reduced by the influence of drift, vibrations, or other instabilities during acquisition. Drift corrected frame integration (DCFI) is an acquisition method within the Thermo Scientific Velox Software that overcomes this problem, producing images with high contrast and a high signal-to-noise ratio. The addition of Integrated Differential Phase Contrast (iDPC) Software enables the collection of easily interpretable high-resolution images with more reliable, simultaneous imaging of light and heavy elements, even at low-dose conditions.
 High-resolution transmission electron microscopes from Thermo Fisher Scientific
The Spectra S/TEM instruments take high-resolution imaging one step further with additional aberration correction via the S-CORR probe aberration corrector, making sub-Angstrom (<0.8 Å) S/TEM imaging regularly attainable. Finally, new Talos and Spectra S/TEM instruments feature the Panther STEM detection system, which includes optimized mechanical alignment and detector geometry for better multi-signal acquisition and mechanical alignment accuracy. It has a higher throughput and easier operation, with linear response of gain/offset and more flexibility in signal processing. Visualize more details with up to 16 segments and a new amplifier design with ultrahigh electron sensitivity for low-dose STEM.
With the combination of high-quality automated S/TEM instrumentation and leading software solutions, HRTEM and HRSTEM imaging are more accessible than ever, giving you the ability to gather unparalleled atomic-resolution information on your most challenging materials.
Gallium nitride imaged at atomic resolution.
Gallium nitride [212] imaged with HAADF (DCFI) STEM at 300 kV, showing 40.5 pm Ga-Ga dumbbell splitting and 39 pm resolution in the fast Fourier transform on a wide gap (S-TWIN) pole piece.
Resources
Applications
Fundamental Materials Research_R&amp;D_Thumb_274x180_144DPI
Fundamental Materials Research
Novel materials are investigated at increasingly smaller scales for maximum control of their physical and chemical properties. Electron microscopy provides researchers with key insight into a wide variety of material characteristics at the micro- to nano-scale.
Samples
Battery Research
Battery development is enabled by multi-scale analysis with microCT, SEM and TEM, Raman spectroscopy, XPS, and digital 3D visualization and analysis. Learn how this approach provides the structural and chemical information needed to build better batteries.
Learn more ›
Nanoparticles
Materials have fundamentally different properties at the nanoscale than at the macroscale. To study them, S/TEM instrumentation can be combined with energy dispersive X-ray spectroscopy to obtain nanometer, or even sub-nanometer, resolution data.
Learn more ›
Metals Research and Characterization
Effective production of metals requires precise control of inclusions and precipitates. Our automated tools can perform a variety of tasks critical for metal analysis including; nanoparticle counting, EDS chemical analysis and TEM sample preparation.
Learn more ›
Fibers and Filters
The diameter, morphology and density of synthetic fibers are key parameters that determine the lifetime and functionality of a filter. Scanning electron microscopy (SEM) is the ideal technique for quickly and easily investigating these features.
Learn more ›
Polymers Research
Polymer microstructure dictates the material’s bulk characteristics and performance. Electron microscopy enables comprehensive microscale analysis of polymer morphology and composition for R&D and quality control applications.
Learn more ›
Geological Research
Geoscience relies on consistent and accurate multi-scale observation of features within rock samples. SEM-EDS, combined with automation software, enables direct, large-scale analysis of texture and mineral composition for petrology and mineralogy research.
Learn more ›
Catalysis Research
Catalysts are critical for a majority of modern industrial processes. Their efficiency depends on the microscopic composition and morphology of the catalytic particles; EM with EDS is ideally suited for studying these properties.
Learn more ›
Products
Style Sheet for Instrument Cards Original
Iliad (S)TEM
  • Advanced integration of EELS and (S)TEM optics
  • Electrostatic beam blanker
  • High energy resolution electron source
Spectra 300
  • Highest-resolution structural and chemical information at the atomic level
  • Flexible high-tension range from 30-300 kV
  • Three lens condenser system
Spectra 200
  • High-resolution and contrast imaging for accelerating voltages from 30-200 kV
  • Symmetric S-TWIN/X-TWIN objective lens with wide-gap pole piece design of 5.4 mm
  • Sub-Angstrom STEM imaging resolution from 60 kV-200 kV
Talos F200X TEM
  • High-resolution, EDS cleanliness, and quality in 2D as well as 3D
  • X-FEG and X-CFEG available for the highest brightness and energy resolution
  • High accuracy and repeatable results with integrated Thermo Scientific Velox Software
Thermo Scientific Talos F200C transmission electron microscope (TEM)
Talos F200C TEM
  • High-contrast and high-quality TEM and STEM imaging
  • 4k x 4k Ceta CMOS camera options for large FOV and high read-out speeds
  • Large pole piece gap and multiple in situ options
Talos F200i TEM
  • Compact design with X-TWIN objective lens
  • Available with S-FEG, X-FEG, and X-CFEG
  • Flexible and fast EDS options for comprehensive elemental analysis
Talos F200S TEM
  • Intuitive and easy-to-use automation software
  • Available with Super-X EDS for rapid quantitative chemical analysis
  • High-throughput with simultaneous multi-signal acquisition
Thermo Scientific Talos L120C transmission electron microscope (TEM)
Talos L120C TEM
  • High versatility and stability
  • 4k x 4k Ceta CMOS camera for speed and large FOV
  • TEM magnification range of 25X to 650kX
  • EDS and STEM options for compositional analyses
Velox
  • An experiments panel on the left side of the processing window.
  • Live quantitative mapping
  • Interactive detector layout interface for reproducible experiment control and setup
Style Sheet for Komodo Tabs
Style Sheet to change H2 style to p with em-h2-header class
Contact us
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards
Electron microscopy services for
the materials science
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.