Construct highly representative cDNA libraries with the CloneMiner™ II cDNA Library Construction Kit
  • High primary titers
  • Large average insert sizes, typically >1.5 kb
  • Highest percentage of full-length genes
CloneMiner™ II cDNA Library Construction Kit
The innovative CloneMiner™ II library construction technology combines SuperScript® III Reverse Transcriptase with Gateway® technology to produce highly representative cDNA libraries without restriction enzyme cloning.  This enables the discovery of previously unobtainable, fully intact clones.  The kit also avoids the use of lengthy and inefficient ligation reactions, making library construction faster and offering more complete representation.
Ordering Information
Catalog # Name Size Price (USD) Qty
A11180 CloneMiner™ II cDNA Library Construction Kit Each
3,591.65
Online Exclusive
3,805.00
Save 213.35 (6%)
For Research Use Only. Not for use in diagnostic procedures.