Fura-2, Indo-1 and Related Derivatives
Fura-2 and Indo-1
Fura-2 and indo-1 are UV light–excitable, ratiometric Ca2+ indicators. Fura-2 has become the dye of choice for ratio-imaging microscopy (photo), in which it is more practical to change excitation wavelengths than emission wavelengths.ref Upon binding Ca2+, fura-2 exhibits an absorption shift that can be observed by scanning the excitation spectrum between 300 and 400 nm, while monitoring the emission at ~510 nm (Figure 19.2.1). In contrast, indo-1 is a preferred dye for flow cytometry, where it is more practical to use a single laser for excitation—usually the 351–364 nm spectral lines of the argon-ion laser—and monitor two emissions.ref The emission maximum of indo-1 shifts from ~475 nm in Ca2+-free medium to ~400 nm when the dye is saturated with Ca2+ (Figure 19.2.2).
Modern two-photon excitation imaging techniques used with fura-2 and indo-1 ref avoid the deleterious effects of conventional ultraviolet illumination on live specimens. Indo-1 may be less subject to compartmentalization than fura-2,ref whereas fura-2 is more resistant to photobleaching than indo-1.ref Both fura-2 and indo-1 exhibit Kd values that are close to typical basal Ca2+ levels in mammalian cells (~100 nM) and display high selectivity for Ca2+ binding relative to Mg2+.ref Nevertheless, Ca2+ binding is discernibly perturbed by physiological levels of Mg2+; the Kd for Ca2+ of fura-2 is ~135 nM in Mg2+-free Ca2+ buffers and ~224 nM in the presence of 1 mM Mg2+ref (measured at 37°C in 100mM KCl, 10 mM MOPS, pH 7.0). Fura-2 and indo-1 also exhibit high affinities for other divalent cations such as Zn2+ and Mn2+, a property that is discussed further in Fluorescent Indicators for Zn2+ and Other Metal Ions—Section 19.7.
The sodium and potassium salts of fura-2 (F1200; Figure 19.2.3) and potassium salt of indo-1 are cell-impermeant probes that can be delivered into cells by microinjection or using our Influx pinocytic cell-loading reagent (I14402, Chelators, Calibration Buffers, Ionophores and Cell-Loading Reagents—Section 19.8). Free acids of fura-2 and indo-1 can also be loaded into some plant cells at pH 4–5.ref In addition, these salts are useful as standards for calibrating Ca2+ measurements.
Unlike the salt forms, the acetoxymethyl (AM) esters of fura-2 and indo-1 can passively diffuse across cell membranes, enabling researchers to avoid the use of invasive loading techniques. Once inside the cell, these esters are cleaved by intracellular esterases to yield cell-impermeant fluorescent indicators (Loading and Calibration of Intracellular Ion Indicators—Note 19.1). We offer fura-2 AM and indo-1 AM in 1 mg vials (F1201, I1203) or specially packaged in 20 vials of 50 µg each (F1221, I1223); the special packaging is recommended when small quantities of the dyes are to be used over a long period of time. We also provide stock solutions of fura-2 AM and indo-1 AM in anhydrous DMSO at 1 mg/mL (~1 mM; F1225, I1226). Our standard analytical specifications for fura-2 AM require ≥95% purity by HPLC. We also offer a special packaged high-purity grade of fura-2 AM that is specified to have ≥98% purity by HPLC (as a set of 20 vials, each containing 50 µg; F14185). The 10,000 MW dextran conjugate of fura is described in Fluorescent Ca2+ Indicator Conjugates—Section 19.4.
calcium-uv-light.par.33272.image.275.253.1.s000288-spectra-fura-2-gif
Figure 19.2.1 Fluorescence excitation spectra of fura-2 (F1200) in solutions containing 0–39.8 µM free Ca2+.
calcium-uv-light.par.54520.image.275.255.1.s000298-spectra-indo-1-gif
Figure 19.2.2 Fluorescence emission spectra of indo-1 in solutions containing 0–39.8 µM free Ca2+.
calcium-uv-light.par.59084.image.275.363.1.s001106-indicators-calcium-affinitiis-gif
Figure 19.2.3 Fura indicators with varying Ca2+ affinities.
Fura-2 Calcium Imaging Calibration Kit
The Fura-2 Calcium Imaging Calibration Kit (F6774) is designed to facilitate rapid calibration and standardization of digital imaging microscopes.ref This kit provides 11 CaEGTA:EGTA buffer solutions with free Ca2+ concentrations from zero (10 mM EGTA) to 39 µM. Each solution also includes 50 µM fura-2, as well as 15 µm unstained polystyrene microspheres to act both as spacers that ensure uniform separation between the slide and the coverslip and as focusing aids. We also provide a twelfth buffer—identical to the 10 mM CaEGTA standard but lacking fura-2—that serves as a control for background fluorescence. Our Calcium Calibration Kits are described further in Chelators, Calibration Buffers, Ionophores and Cell-Loading Reagents—Section 19.8.
Bis-Fura-2: Brighter Signal with Lower Affinity for Ca2+
By linking two fura fluorophores with one BAPTA chelator, we have produced bis-fura-2, a Ca2+ indicator that exhibits approximately twice the absorptivity of fura-2. Bis-fura-2 has a Kd for Ca2+ of ~370 nM and ~525 nM in the absence and presence of 1 mM Mg2+, respectively (measured at ~22°C using our Calcium Calibration Buffer Kits). In other aspects, the quantum yield of bis-fura-2 and its spectral response to Ca2+ (Figure 19.2.4) are virtually identical to those of fura-2. Although the difference between the Kd of fura-2 and bis-fura-2 for Ca2+ is sall, the change in excitation ratio for bis-fura-2 in response to Ca2+ concentrations >500 nM is larger than that of fura-2 (Figure 19.2.1); this difference can improve the dynamic range for Ca2+ measurements in cells.ref Other potential advantages of bis-fura-2 include:
  • Higher fluorescence output per indicator, which may allow the use of lower dye concentrations ref
  • Lower affinity for Ca2+, which decreases the buffering of intracellular Ca2+ and produces a faster response to Ca2+ spikes ref
  • An additional negative charge, which may facilitate dye retention
The hexapotassium salt of bis-fura-2 has been used for loading by microinjection ref or by infusion from a patch pipette.ref
calcium-uv-light.par.28403.image.275.254.1.s001425-spectra-bis-fura2-gif
Figure 19.2.4 Fluorescence excitation spectra of bis-fura-2 in solutions containing 0–39.8 µM free Ca2+.
Quin-2
Quin-2 belongs to the first generation of Ca2+ indicators developed by Tsien.ref Quin-2 has lower absorptivity and quantum yield values than the fura-2, indo-1, fluo-3, fluo-4 and Calcium Green indicators and thus requires higher loading concentrations. The resulting high intracellular concentration of the indicator may buffer intracellular Ca2+ transients.ref Quin-2 AM has been used to intentionally deplete cytosolic free Ca2+ref and to ensure unidirectional Ca2+ influx.ref Measurement of cytosolic free Ca2+ with quin-2 has been thoroughly reviewed by Tsien and Pozzan.ref
Indicators with Intermediate Calcium-Binding Affinity
Fura-4F, Fura-5F and Fura-6F
Calcium concentrations above 1 µM produce almost complete binding saturation of fura-2 but very low fractional saturation of the low-affinity fura analog mag-fura-2 (M1290, see below). To bridge this gap in the Ca2+ measurement range of fura-type indicators, we developed three additional ratiometric Ca2+ indicators—fura-4F, fura-5F and fura-6F—as well as the membrane-permeant fura-4F AM ref (F14175); these ratiometric fura-type indicators may be available upon request through Custom Services. Attachment of a single electron-withdrawing fluorine substituent at different positions on the BAPTA chelator moiety of fura-2 (Figure 19.2.3) results in an increase of the Kd value to ~770 nM, ~400 nM and 5.3 µM for fura-4F, fura-5F and fura-6F, respectively (measured at 22°C at 100 mM KCl, 10 mM MOPS, pH 7.2). Except for the change in the Ca2+ concentration response range (Figure 19.2.5), the Ca2+-dependent spectral shifts produced by fura-4F, fura-5F and fura-6F are essentially identical to those of fura-2 (Figure 19.2.6) and the probes use the same optical filter sets.
calcium-uv-light.par.2278.image.275.242.1.s001309-fluorescence-excitation-ratio-gif
Figure 19.2.5 Fluorescence excitation ratio versus Ca2+ concentration curves for fura-2 (red), fura-5F (orange), fura-4F (green) and fura-6F (blue).
calcium-uv-light.par.21999.image.275.254.1.s001557-excitation-spectra-fura-4f-gif
Figure 19.2.6 Ca2+-dependent fluorescence excitation spectra of fura-4F.
Fura-FF
Fura-FF is a difluorinated derivative of fura-2 (Figure 19.2.3) with a Kd value of ~5.5 µM ref (measured at 22°C in 100 mM KCl, 10 mM MOPS, pH 7.2) and similar spectroscopic properties (Figure 19.2.7). Fura-FF has negligible Mg2+ sensitivity, making Ca2+ detection less susceptible to interference than with mag-fura-2.ref These properties have made fura-FF particularly useful for spatial and functional characterization of intracellular Ca2+ stores ref and for tracking Ca2+ oscillations driven by the inositol 1,4,5-triphosphate receptor.ref The low-affinity indicator fura-FF detected NMDA- and kainate-induced neuronal Ca2+ fluxes that were not detectable with the higher-affinity indicator fura-2.ref Fura-FF has also been used in combination with FluoZin-3 (Fluorescent Indicators for Zn2+ and Other Metal Ions—Section 19.7) for simultaneous detection of Ca2+ and Zn2+.ref Both the water-soluble potassium salt and membrane-permeant AM ester derivative of Fura-FF have been used for Ca2+ detection.
calcium-uv-light.par.23914.image.275.252.1.s001338-excitation-spectra-fura-ff-gif
Figure 19.2.7 Ca2+-dependent fluorescence excitation spectra of fura-FF.
Low-Affinity Calcium Indicators
BTC
The coumarin benzothiazole–based Ca2+ indicator BTC and its cell-permeant derivative BTC AM (B6791) were developed in collaboration with Haralambos Katerinopoulos of the University of Crete.ref This Ca2+ indicator exhibits a shift in excitation maximum from about 480 nm to 400 nm upon binding Ca2+ (Figure 19.2.8), permitting ratiometric measurements that are essentially independent of uneven dye loading, cell thickness, photobleaching and dye leakage.ref Its high selectivity and moderate affinity for Ca2+ (Kd ~7 µM)ref allows accurate quantitation of high intracellular Ca2+ levels that are underestimated by fura-2 measurements.ref When loaded into neurons as its AM ester, BTC exhibits little compartmentalization; however, prolonged excitation appears to cause conversion of the indicator to a calcium-insensitive form.ref
BTC has been employed in investigations of Ca2+-dependent exocytosis in pancreatic β-cells,ref CHO fibroblasts ref and phaeochromocytoma cells.ref Neuronal Ca2+ transients detected by the low-affinity Ca2+ indicators BTC and mag-fura-2 are significantly more rapid than those reported by the higher-affinity indicators fura-2 and Calcium Green-2.ref
Fluorescence excitation spectra of BTC 
Figure 19.2.8 Fluorescence excitation spectra of BTC in solutions containing 0–100 µM free Ca2+.
Mag-Fura-2 and Mag-Indo-1
Mag-fura-2 (also called furaptra) and mag-indo-1 were originally designed to report intracellular Mg2+ levels (Fluorescent Mg2+ Indicators—Section 19.6); however, these indicators actually have much higher affinity for Ca2+ than for Mg2+. Although Ca2+ binding by these indicators may complicate analysis when they are employed to measure intracellular Mg2+,ref their increased effective range and improved linearity for Ca2+ measurements has been exploited for measuring intracellular Ca2+ levels between 1 µM and 100 µM.ref
The spectral shifts of mag-fura-2 and mag-indo-1 are very similar to those of fura-2 and indo-1 but occur at higher Ca2+ concentrations. Because the off-rates for Ca2+ binding of these indicators are much faster than those of fura-2 and indo-1, these dyes have been used to monitor action potentials in skeletal muscle and nerve terminals with little or no kinetic delay ref (Figure 19.2.9). The spectral properties, kinetics and selectivity of several of our low-affinity Ca2+ indicators have been reviewed by Zhao,ref Hyrc ref and their co-workers.
The moderate Ca2+ affinity of mag-fura-2 and the tendency of its AM ester to accumulate in subcellular compartments have proven useful for in situ monitoring of inositol 1,4,5-triphosphate–sensitive Ca2+ stores.ref Mag-fura-2 has also been employed to follow Ca2+ transients in presynaptic nerve terminals,ref gastric epithelial cells ref and cultured myocytes.ref Mag-indo-1 has been used to detect gonadotropin-releasing hormone–induced Ca2+ oscillations in gonadotropes ref and to investigate the role of Ca2+/K+ exchange in intracellular Ca2+ storage and release processes.ref The cell-impermeant potassium salts and cell-permeant AM esters of mag-fura-2 (M1290, M1292) and mag-indo-1 have been used for Ca2+ detection.
Cerebellar-Granule-Cells 
Figure 19.2.9 Ca2+ transients evoked by trains of 1–4 action potentials in rat cerebellar granule cells detected by fura-2 (upper panel, F1200) and mag-fura-2 (lower panel, M1290). The stimulus pulses are 50 milliseconds apart (20 Hz); timing is indicated by the double-headed arrows. The amplitude of the transients detected by fura-2 decreases with each successive stimulus due to Ca2+ saturation. Mag-fura-2 avoids saturation due to its lower Ca2+ binding affinity (Kd for Ca2+ = 25 µM), recording transients of approximately equal amplitude from successive action potentials. Adapted with permission from Biophys J (1995) 68:2165.
Data Table
For a detailed explanation of column headings, see Definitions of Data Table Contents
 Low Ca2+High Ca2+ 
Cat. No.
MWStorageSolubleAbsECEmSolventAbsECEmSolventProductKdNotes
BTC844.03F,D,LpH >646429,000533H2O40120,000529H2O/Ca2+ 7.0 µM1, 2, 3
B6791
BTC AM
979.92F,D,LDMSO43339,000504MeOH    BTC  
bis-fura-21007.14F,D,LpH >636656,000511H2O33868,000504H2O/Ca2+ 370 nM1, 2, 4, 5
F1200
fura-2, pentapotassium salt
832.00F,D,LpH >636328,000512H2O33534,000505H2O/Ca2+ 145 nM1, 2, 4, 5
F1201
fura-2 AM
1001.86F,D,LDMSO37031,000476EtOAc    F1200
fura-2
F1221
fura-2 AM
1001.86F,D,LDMSO37031,000476EtOAc    F1200
fura-2
F1225
fura-2 AM
1001.86F,D,LDMSO37031,000476EtOAc    F1200
fura-2
 6
fura-2, pentasodium salt751.45F,D,LpH >636328,000512H2O33534,000505H2O/Ca2+ 145 nM1, 2, 4, 5
fura-4F835.96F,D,LpH >636621,000511H2O33623,000505H2O/Ca2+ 770 nM1, 2, 4
F14175
fura-4F AM
1005.82F,D,LDMSO37029,000475EtOAc    fura-4F  
fura-5F835.96F,D,LpH >636326,000512H2O33629,000506H2O/Ca2+ 400 nM1, 2, 4
fura-6F835.96F,D,LpH >636425,000512H2O33628,000505H2O/Ca2+ 5.3 µM1, 2, 3
fura-FF853.95F,D,LpH >636425,000510H2O33528,000506H2O/Ca2+ 5.5 µM1, 2, 3
fura-FF AM1023.82F,D,LDMSO37030,000476EtOAc    fura-FF  
F14185
fura-2 AM
1001.86F,D,LDMSO37031,000476EtOAc    F1200
fura-2
 7
indo-1840.06F,D,LpH >634633,000475H2O33033,000401H2O/Ca2+ 230 nM1, 2, 4, 5
I1203
indo-1 AM
1009.93F,D,LDMSO35639,000478MeOH    indo-1  
I1223
indo-1 AM
1009.93F,D,LDMSO35639,000478MeOH    indo-1  
I1226
indo-1 AM
1009.93F,D,LDMSO35639,000478MeOH    indo-1 6
M1290
mag-fura-2
586.68F,D,LpH >636922,000511H2O32926,000508H2O/Ca2+ 25 µM1, 2, 3
M1292
mag-fura-2 AM
722.57F,D,LDMSO36631,000475EtOAc    M1290
mag-fura-2
mag-indo-1594.74F,D,LpH >634938,000480H2O32835,000390H2O/Ca2+ 35 µM1, 2, 8, 9
mag-indo-1 AM730.63F,D,LDMSO35437,000472MeOH    mag-indo-1  
quin-2541.51D,LpH >63534000495H2O3333900495H2O/Ca2+ 60 nM1, 2, 10
  1. Dissociation constants are known to vary considerably depending on the temperature, pH, ionic strength, viscosity, protein binding, presence of other ions (especially polyvalent ions), instrument setup and other factors. It is strongly recommended that these values be verified under user-specific experimental conditions.
  2. Spectra measured in aqueous buffers containing 10 mM EGTA (H2O) or a >10-fold excess of free Ca2+ relative to the Kd (H2O/Ca2+).
  3. Dissociation constant determined by fluorescence measurements in 100 mM KCl, 10 mM MOPS pH 7.2, 0 to 1 mM free Ca2+ at 22°C.
  4. Dissociation constant determined by fluorescence measurements in 100 mM KCl, 10 mM MOPS, pH 7.2, 0 to 39 µM free Ca2+ at 22°C.
  5. Kd(Ca2+) for fura-2 and indo-1 from the original reference by Grynkiewicz, Poenie and Tsien ref are 224 nM and 250 nM, respectively, measured in 1 mM EGTA, 100 mM KCl, 1 mM free Mg2+, 10 mM MOPS, pH 7.0 at 37°C. For bis-fura-2, Kd(Ca2+) in presence of Mg2+ is 525 nM (determined in our laboratories using 100 mM KCl, 10 mM MOPS, pH 7.2, 1 mM Mg2+ at 22°C).
  6. This product is supplied as a ready-made solution in the solvent indicated under "Soluble."
  7. This product is specified to equal or exceed 98% analytical purity by HPLC.
  8. The emission spectrum of Ca2+-bound mag-indo-1 excited at 340 nm has approximately equal peak intensities at ~390 nm and ~480 nm.ref
  9. Dissociation constant determined in 100 mM KCl, 40 mM HEPES, pH 7.0 at 22°C.ref
  10. Kd(Ca2+) for quin-2 was measured in 120 mM KCl, 20 mM NaCl, pH 7.05 at 37°C. Under the same conditions with addition of 1 mM Mg2+, Kd = 115 nM.ref
Ordering Information
Catalog # Name Size Price (USD) Qty
B6791 BTC, AM, cell permeant Each
400.00
F1201 Fura-2, AM, cell permeant, 1mg Each
292.65
Online Exclusive
311.00
Save 18.35 (6%)
F1221 Fura-2, AM, cell permeant, 20 x 50μg Each
397.65
Online Exclusive
424.00
Save 26.35 (6%)
F1225 Fura-2, AM, cell permeant (1 mM Solution in Anhydrous DMSO) Each
345.65
Online Exclusive
369.00
Save 23.35 (6%)
F14185 Fura-2, AM, FluoroPure™ grade - Special Packaging Each
500.00
F6774 Fura-2 Calcium Imaging Calibration Kit (Zero to 10 mM CaEGTA, 50 μM Fura-2) Each
374.00
F1200 Fura-2, Pentapotassium Salt, cell impermeant Each
240.65
Online Exclusive
257.00
Save 16.35 (6%)
F14175 Fura-4F, AM, cell permeant - Special Packaging Each
550.00
I1203 Indo-1, AM, cell permeant Each
245.00
I1223 Indo-1, AM, cell permeant - Special Packaging Each
327.65
Online Exclusive
351.00
Save 23.35 (7%)
I1226 Indo-1, AM, cell permeant Each
324.00
M1292 Mag-Fura-2, AM, cell permeant, 20 x 50μg Each
772.00
M1290 Mag-Fura-2, Tetrapotassium Salt, cell impermeant Each
550.00
For Research Use Only. Not for use in diagnostic procedures.