Related Product Information
Introduction
Lipofectamine® LTX Reagent is a proprietary, animal-origin free formulation for the transfection of DNA into eukaryotic cells with low cytotoxicity. This reference provides a recommended procedure to transfect plasmid DNA into human A549 lung carcinoma cells (ATCC, Cat. No. CCL-185) using Lipofectamine® LTX Reagent (Cat. No. 15338-100).
Important Guidelines for Transfection
Follow these important guidelines when transfecting DNA into A549 cells using Lipofectamine® LTX Reagent:
- The addition of antibiotics to media during transfection may result in cell death, and has not been tested for A549 cells. If you wish to use antibiotics during transfection, test your conditions thoroughly.
- Maintain the same seeding conditions between experiments. Use low-passage cells; make sure that cells are healthy and greater than 90% viable before transfection.
- Transfection can be performed both in the presence or absence of serum. Test serum-free media for compatibility with Lipofectamine® LTX Reagent.
- Using PLUS™ Reagent (Cat. No. 11514-015) enhances transfection performance in A549 cells.
- We recommend Opti-MEM® I Reduced Serum Medium (Cat. No. 31985-062) to dilute the DNA and Lipofectamine® LTX Reagent before complexing.
- Visit www.lifetechnologies.com/transfection or contact Technical Service for other specialized transfection protocols (including cell-type specific advice on use of PLUS™ Reagent and antibiotics, and a protocol for vector-based RNAi).
- Lipofectamine® LTX Reagent performs well with vector-based RNAi experiments. For siRNA and Stealth™ RNAi transfections, we recommend Lipofectamine® RNAiMAX (Cat. No. 13778-075). Go to www.lifetechnologies.com/RNAi or contact Technical Service for more information.
Materials Needed
Have the following reagents on-hand before beginning:
- A549 cells maintained in RPMI with L-glutamine (Cat. No. 11875-085) supplemented with 10% fetal bovine serum (Cat. No. 26140-079), and 0.1 mM MEM Non-Essential Amino Acids Solution (cat. No. 11140-050). Grow cells at 37°C with 5% CO2.
- Plasmid DNA of interest (100 ng/μl or higher)
- Lipofectamine® LTX Reagent (store at +4°C until use), and PLUS™ Reagent (if desired; store at 4°C)
- Opti-MEM® I Reduced Serum Medium
- Appropriate tissue culture plates and supplies.
15338100,31985062,115141015,11875085,26140079,11140050
Ordering Information
15338100,31985062,11875085,26140079,11140050
Use this procedure to transfect plasmid DNA into A549 cells in a 24-well format (for other formats, see Scaling Up or Down Transfections, below). All amounts and volumes are given on a per well basis.
- The day before transfection, trypsinize and count the cells. Plate 4 x 104cells per well in 0.5 ml of complete growth medium. Cell density should be 50~80% confluent on the day of transfection.
- For each well of cells to be transfected, dilute 0.5 μg of DNA into 100 μl of Opti-MEM® I Reduced Serum Medium without serum.
- If using PLUS™ Reagent: Mix PLUS™ Reagent gently before use, then add 0.5 μl PLUS™ Reagent (a 1:1 ratio to DNA) directly to the diluted DNA. Mix gently and incubate for 5-15 minutes at room temperature.
- For each well of cells, dilute 1.5-2.5 μl of Lipofectamine® LTX into the above diluted DNA solution, mix gently and incubate for 25 minutes at room temperature to form DNA-Lipofectamine® LTX complexes.
- Remove growth medium from cells and replace with 0.5 ml of complete growth medium. Add 100 μl of the DNA-Lipofectamine® LTX complexes directly to each well containing cells and mix gently by rocking the plate back and forth.
- Complexes do not have to be removed following transfection. Incubate the cells at 37°C in a CO2 incubator for 18-24 hours post-transfection before assaying for transgene expression.
Scaling Up or Down Transfections
To transfect A549 cells in different tissue culture formats, vary the amounts of Lipofectamine® LTX Reagent, DNA, cells, medium and PLUS™ Reagent used in proportion to the relative surface area, as shown in the table (amounts given on a per well basis).
Culture vessel | Surface area per well1 | Volume plating medium | Cell per well | Volume dilution medium2 | DNA | Lipofectamine® LTX Reagent | PLUS™ Reagent |
---|
96-well | 0.3 cm2 | 100 μl | 8 x 103 | 20 μl | 100 ng | 0.3 - 0.5 μl | 0.1 μl |
48-well | 1 cm2 | 200 μl | 2 x 104 | 40 μl | 200 ng | 0.6 - 1.0 μl | 0.2 μl |
24-well | 2 cm2 | 500 μl | 4 x 104 | 100 μl | 500 ng | 1.5 - 2.5 μl | 0.5 μl |
12-well | 4 cm2 | 1 ml | 8 x 104 | 200 μl | 1 μg | 3.0 - 5.0 μl | 1.0 μl |
6-well | 10 cm2 | 2 ml | 2 x 105 | 500 μl | 2.5 μg | 7.5 - 12.5 μl | 2.5 μl |
1 Surface areas may vary depending on the manufacturer.
2 If the volume of Lipofectamine® LTX Reagent is too small to dispense accurately, and you cannot pool dilutions, predilute Lipofectamine® LTX Reagent 10-fold in Opti-MEM® I Reduced Serum Medium, and dispense a 10-fold higher amount (should be at least 1.0 μl per well). Discard any unused diluted Lipofectamine® LTX Reagent.
25-0928W Rev. 8 June 2006