Search Thermo Fisher Scientific
To visualize the RNase digestion products from an RNA, the RNA should be end-labeled either at its 5' end using a kinase reaction with [gamma-32P]ATP (KinaseMax™, Cat #1520), or at its 3' end using an RNA ligation reaction with [32P]pCp (T4 RNA Ligase, Cat #2140). See Protocol for 5' End Labeling RNA and Protocol for 3' End Labeling RNA for more information.
Protocol
Before use, the Inactivation/Precipitation Buffer should be completed by addition of 3.2 mL of 100% ethanol.
Protocol
Before use, the Inactivation/Precipitation Buffer should be completed by addition of 3.2 mL of 100% ethanol.
RNase | Cleavage Site | Ambion Cat # |
---|---|---|
A | 3' of ss C's and U's | 2274 |
V1 | base-paired nucleotides | 2275 |
T1 | 3' of ss G's | 2283 |