Protocols
Introduction
Overview The Invitrogen T-REx System is a tetracycline-regulated mammalian expression system that uses regulatory elements from the E. coli Tn10-encoded tetracycline (Tet) resistance operon (Hillen and Berens, 1994; Hillen et al., 1983). Tetracycline regulation in the T-REx System is based on the binding of tetracycline to the Tet repressor and derepression of the promoter controlling expression of the gene of interest (Yao et al., 1998). The major components of the T-REx System include:
  • An inducible expression plasmid for expression of your gene of interest under the control of the strong human cytomegalovirus immediate-early (CMV) promoter and two tetracycline operator 2 (TetO2) sites
  • A regulatory plasmid, pcDNA6/TR™, which encodes the Tet repressor (TetR) under the control of the human CMV promoter Tetracycline for inducing expression
  • A control expression plasmid containing the lacZ gene, which when cotransfected with pcDNA6/TR™, expresses β-galactosidase upon induction with tetracycline.
For specific information on the inducible expression vector and the corresponding positive control vector containing the lacZ gene, please refer to the manual for the inducible expression vector you are using.
Description of the T-REx System
In the T-REx System, expression of your gene of interest is repressed in the absence of tetracycline and induced in the presence of tetracycline (Yao et al., 1998). Unlike other tetracycline-regulated systems which use hybrid regulatory molecules and viral transactivation domains (Gossen and Bujard, 1992), the T-REx System uses only regulatory elements from the native Tet operon (Yao et al., 1998). Tetracycline-regulated gene expression in the T-REx System more closely resembles the regulation of the native bacterial tet operon (Hillen and Berens, 1994; Hillen et al., 1983) and avoids the potentially toxic effects of viral transactivation domains observed in some mammalian cell lines.
The major component of the T-REx System is the inducible expression plasmid. Expression of your gene of interest from the inducible expression vector is controlled by the strong CMV promoter (Andersson et al., 1989; Boshart et al., 1985; Nelson et al., 1987) into which 2 copies of the tet operator 2 (TetO2) sequence have been inserted in tandem. The TetO2 sequences consist of 2 copies of the 19 nucleotide sequence, 5´-TCCCTATCAGTGATAGAGA-3´ separated by a 2 base pair spacer (Hillen and Berens, 1994; Hillen et al., 1983). Each 19 nucleotide TetO2 sequence serves as the binding site for 2 molecules of the Tet repressor. For more information about the Tet operator sequences and the specific features of each inducible expression vector, please refer to the manual for the vector you are using.
The second major component of the T-REx System is the pcDNA6/TR regulatory vector which expresses high levels of the TetR gene (Postle et al., 1984) under the control of the human CMV promoter. Both T-REx vectors can be introduced into mammalian host cells by standard transfection methods.
Mechanism of repression
In the absence of tetracycline, the Tet repressor forms a homodimer that binds with extremely high affinity to each TetO2 sequence in the promoter of the inducible expression vector (Hillen and Berens, 1994). The 2 TetO2 sites in the promoter of the inducible expression vector serve as binding sites for 4 molecules (or 2 homodimers) of the Tet repressor. The affinity of the Tet repressor for the tet operator is KB = 2 x 1011 M-1 (as measured under physiological conditions), where KB is the binding constant (Hillen and Berens, 1994). Binding of the Tet repressor homodimers to the TetO2 sequences represses transcription of your gene of interest. Upon addition, tetracycline binds with high affinity to each Tet repressor homodimer in a 1:1 stoichiometry and causes a conformational change in the repressor that renders it unable to bind to the Tet operator. The association constant, KA, of tetracycline for the Tet repressor is 3 x 109 M-1 (Hillen and Berens, 1994). The Tet repressor: tetracycline complex then dissociates from the Tet operator and allows induction of transcription from the gene of interest. 
Experimental outline
The gene of interest is cloned into the multiple cloning site of the inducible expression vector, and the resulting construct cotransfected with the regulatory plasmid, pcDNA6/TR™ into mammalian cells. After transfection, cells are treated with tetracycline to derepress the hybrid CMV/TetO2 promoter in the inducible expression vector and induce transcription of your gene of interest.
The positive control vector containing the lacZ gene can be transiently cotransfected into mammalian cells with pcDNA6/TR™ to demonstrate that the system is working properly in your cell line. Stable cell lines expressing Tet repressor from pcDNA6/TR™ can be established to serve as hosts for inducible expression vector-based constructs. 
Ordering information
Important information
The T-REx System manual is supplied with the kits listed below. The Core System includes the inducible expression vector of choice, the regulatory vector, and primers for sequencing. The Complete System includes the Core System plus inducing and selection agents. See below for a detailed description of the contents of each T-REx Kit.
Shipping/storage
The T-REx Core System is shipped at room temperature. Store at -20°C.
The T-REx Complete System is shipped in 2 boxes. Store as described below:
  • Box 1 contains vectors, primers, and blasticidin and is shipped at room temperature. Upon receipt, remove the vectors and primers and store at -20°C. Blasticidin powder should be stored at +4°C.
  • Box 2 contains tetracycline and Zeocin and is shipped on blue ice. Store at +4°C. For long-term storage (> 6 months), store at -20°C. Store the tetracycline and Zeocin protected from exposure to light.  
Kit contents
Both the T-REx Complete and the T-REx Core Systems include the following regulatory vector and sequencing primers. Store at -20°C.
Reagent
Amount
Comments
pcDNA6/TR™
20 µg, lyophilized in TE, pH 8.0
Regulatory vector that expresses the tetracycline (Tet) repressor
CMV Forward Primer
(21-mer)
2 µg (306 pmoles), lyophilized in TE, pH 8.0
5´-CGCAAATGGGCGGTAGGCGTG-3´
BGH Reverse Primer
(18-mer)
2 µg (358 pmoles), lyophilized in TE, pH 8.0
5´-tagaaggcacagtcgagg-3´
Inducible expression vector
Each T-REx Complete and Core System also includes one of the following inducible expression vectors and a corresponding positive control vector containing the lacZ gene. Please refer to the vector manual for specific information pertaining to each inducible expression vector. Store at -20°C.
Reagent
Amount supplied
Comments
Tetracycline
5 g, powder
Inducing agent
Zeocin
1 g
Selection agent for inducible expression plasmid
Blasticidin
50 mg, powder
Selection agent for pcDNA6/TR™ plasmid
T-REx cell lines
For your convenience, Invitrogen has available three mammalian cells lines that stably express the Tet repressor. T-REx-293 cells and T-REx-HeLa cells express the Tet repressor from pcDNA6/TR™ and should be maintained in medium containing blasticidin. T-REx-U2OS cells express the Tet repressor from pCEP4/tetR as described in Yao et al., 1998, and should be maintained in medium containing hygromycin. Please note that the pCEP4/tetR plasmid in the T-REx-U2OS cells is episomally-maintained, but is stable under hygromycin selection. For more information, please go to www.thermofisher.com or call Technical Service.
Using pcDNA6/TR™
Introduction
The following section contains guidelines for maintaining and propagating the pcDNA6/TR regulatory vector.
General molecular biology techniques
For assistance with E. coli transformations, restriction enzyme analysis, purification of single-stranded DNA, DNA sequencing, and DNA biochemistry, please refer to Molecular Cloning: A Laboratory Manual (Sambrook et al., 1989) or Current Protocols in Molecular Biology (Ausubel et al., 1994).
E. coli strain
Many E. coli strains are suitable for the propagation of the pcDNA6/TR vector including TOP10F´ (Catalog no. C615-00), DH5αF´, and INVαF´ (Catalog no. C658?00). We recommend that you propagate the pcDNA6/TR vector in E. coli strains that are recombination deficient (recA) and endonuclease A deficient (endA).
For your convenience, TOP10F´ E. coli are available as chemically competent or electrocompetent cells from Invitrogen.
Item
Quantity
Catalog no.
One Shot TOP10F´ (chemically competent cells)
21 x 50 µl
C3030-03
Electrocomp TOP10F´ (electrocompetent cells)
5 x 80 µl
C665-55
Max Efficiency DH10B Chemically Competent Cells
5 x 0.2 ml
18297-010
Transformation method
You may use any method of choice for transformation. Chemical transformation is the most convenient for many researchers. Electroporation is the most efficient and the method of choice for large plasmids.
Maintenance of plasmids
The pcDNA6/TR™ vector contains the ampicillin and blasticidin resistance genes to allow selection of the plasmid using ampicillin or blasticidin. To propagate and maintain the pcDNA6/TR plasmid, we recommend using the following procedure:
  1. Resuspend the vector in 20 µl sterile water to prepare a 1 µg/µl stock solution. Store the stock solution at -20°C.
  2.  Use the stock solution to transform a recA, endA E. coli strain like TOP10F´, INVαF´, or equivalent.
  3.  Select transformants on LB agar plates containing 50 to 100 µg/ml ampicillin or 50 µg/ml blasticidin in Low Salt LB.
  4. Prepare a glycerol stock of each plasmid for long-term storage (see protocol below).
Selection in E. coli
To facilitate selection of blasticidin-resistant E. coli, the salt concentration of the medium must remain low (< 90 mM) and the pH must be 7.0. Prepare Low Salt LB broth and plates using the recipe.  Failure to lower the salt content of your LB medium will result in non-selection due to inhibition of the drug unless a higher concentration of blasticidin is used.
Preparing a glycerol stock
Once you have identified the correct clone, be sure to purify the colony and make a glycerol stock for long-term storage. It is also a good idea to keep a DNA stock of your plasmid at -20°C.
  1. Streak the original colony out on an LB plate containing 50 µg/ml ampicillin or 100 µg/ml blasticidin in Low Salt LB. Incubate the plate at 37°C overnight.
  2. Isolate a single colony and inoculate into 1-2 ml of LB containing 50 µg/ml ampicillin or 50 µg/ml blasticidin in Low Salt LB.
  3. Grow the culture to mid-log phase (OD600 = 0.5-0.7).
  4. Mix 0.85 ml of culture with 0.15 ml of sterile glycerol and transfer to a cryovial.
  5. Store at -80°C.
Plasmid preparation
Plasmid DNA for transfection into eukaryotic cells must be very clean and free from phenol and sodium chloride. Contaminants will kill the cells, and salt will interfere with lipids decreasing transfection efficiency. We recommend isolating DNA using the S.N.A.P. MiniPrep Kit (10-15 µg DNA, Catalog no. K1900-01), the S.N.A.P. MidiPrep Kit (10-200 µg DNA, Catalog no. K1910-01) or CsCl gradient centrifugation.
Transfection
Introduction
Once you have obtained clean plasmid preparations of your inducible expression plasmid and pcDNA6/TR™, you are ready to cotransfect the two plasmids into the mammalian cell line of choice. We recommend that you include the positive control vector and a mock transfection (negative control) to evaluate your results. For more information about the positive control vector, please refer to the manual for the inducible expression vector you are using.
Methods of transfection
For established cell lines (e.g. HeLa, COS-1), please consult original references or the supplier of your cell line for the optimal method of transfection. We recommend that you follow exactly the protocol for your cell line. Pay particular attention to medium requirements, when to pass the cells, and at what dilution to split the cells. Further information is provided in Current Protocols in Molecular Biology (Ausubel et al., 1994).
Methods for transfection include calcium phosphate (Chen and Okayama, 1987; Wigler et al., 1977), lipid-mediated (Felgner et al., 1989; Felgner and Ringold, 1989) and electroporation (Chu et al., 1987; Shigekawa and Dower, 1988). Invitrogen offers the Calcium Phosphate Transfection Kit for mammalian cell transfection and Lipofectamine 2000 Reagent to perform lipid-mediated transfection.
Tetracycline-reduced serum
When culturing cells in medium containing fetal bovine serum (FBS), please note that many lots of FBS contain tetracycline as FBS is generally isolated from cows that have been fed a diet containing tetracycline. If you culture your cells in medium containing FBS that is not reduced in tetracycline, you may observe low basal expression of your gene of interest in the absence of tetracycline. We have cultured our mammalian cells in medium containing FBS that may not be reduced in tetracycline, and have observed undetectable to very low basal expression of ß-galactosidase from the positive control vector in the absence of additional tetracycline. If your gene of interest produces a toxic protein, you may wish to culture your cells in tetracycline-reduced FBS.
For more information, please contact our Gibco FBS Sales Representatives to find the right FBS for your cell culture needs
Tetracycline
Tetracycline (MW = 444.4) is commonly used as a broad spectrum antibiotic and acts to inhibit translation by blocking polypeptide chain elongation in bacteria. In the T-REx System, tetracycline is used as an inducing agent to induce transcription of the gene of interest from the inducible expression vector. Tetracycline induces transcription by binding to the Tet repressor homodimer and causing the repressor to undergo a conformational change that renders it unable to bind to the Tet operator. The association constant of tetracycline to the Tet repressor is 3 x 109 M-1 (Takahashi et al., 1991). Please note that the concentrations of tetracycline used to induce gene expression in the T-REx System are generally not high enough to be toxic to mammalian cells.
  • Tetracycline is light sensitive. Store the powdered drug at +4°C in the dark. Prepare medium containing tetracycline immediately before use.
  • Tetracycline is toxic. Do not ingest or inhale the powder or solutions containing the drug.
  • Wear gloves, a laboratory coat, and safety glasses or goggles when handling tetracycline and tetracycline-containing solutions.
Preparation of tetracycline
To prepare a stock solution from the tetracycline salt supplied with the Complete Kit:
  1.     Weigh out 10 mg of tetracycline and transfer to a sterile 15 ml conical polypropylene tube.
  2.     Resuspend 10 mg of tetracycline in 10 ml of water to produce a 1 mg/ml stock solution that is yellow in color.
Note:   If you are using a different form of tetracycline (i.e. free base form), resuspend in 100% ethanol rather than water.
  3.     Store the stock solution at -20°C protected from exposure to light.
Because tetracycline-regulated expression in the T-REx System is based on a repression/derepression mechanism, the amount of Tet repressor that is expressed in the host cell line from pcDNA6/TR will determine the level of transcriptional repression of the Tet operator sequences in your inducible expression construct. Tet repressor levels should be sufficiently high to suitably repress basal level transcription. We have varied the ratio of pcDNA6/TR and inducible expression plasmid that we transiently cotransfect into mammalian cells to optimize repression and inducibility of the hybrid CMV/TetO2 promoter in the inducible expression plasmid. We recommend that you cotransfect your mammalian host cell line with a ratio of at least 6:1 (w/w) pcDNA6/TR:inducible expression plasmid DNA.
Cotransfection and induction with tetracycline
Guidelines are provided below to cotransfect your inducible expression construct (or the control plasmid) and pcDNA6/TR into your mammalian cell line and to induce expression of your protein of interest with tetracycline. Since every cell line is different and may require a different method of transfection, some empirical experimentation may be needed to determine the optimal conditions for inducible expression.
  • Use cells that are approximately 60% confluent for transfection.
  • Cotransfect the pcDNA6/TR plasmid and your inducible expression construct at a ratio of 6:1 (w:w) into the cell line of choice using your preferred method. Absolute amounts of plasmid will vary depending on the method of transfection and the cell line used.
  • After transfection, add fresh medium and allow the cells to recover for 24 hours before induction.
  • Remove medium and add fresh medium containing the appropriate concentration of tetracycline to the cells. In general, we recommend that you add tetracycline to a final concentration of 1 µg/ml (5 µl of a 1 mg/ml stock per 5 ml of medium) to the cells and incubate the cells for 24 hours at 37°C.
  • Harvest the cells and assay for expression of your gene. 
Optimization of expression
You may want to vary the concentration of tetracycline (0.1 to 1 µg/ml) and time of exposure to tetracycline (8 to 24 hours) to optimize or modulate expression for your cell line.
Other inducers
You may use doxycycline as an alternative inducing agent in the T-REx System. Doxycycline is similar to tetracycline in its mechanism of action, and exhibits similar dose response and induction characteristics as tetracycline in the T-REx System. Doxycycline has been shown to have a longer half-life than tetracycline (48 hours vs. 24 hours, respectively). Doxycycline may be obtained from Sigma-Aldrich (Catalog no. D9891).
Creation of stable cell lines
Introduction
Once you have established that your construct can be inducibly expressed, you may wish to establish a stable cell line that constitutively expresses the Tet repressor and inducibly expresses your gene of interest. We recommend that you first create a stable cell line that expresses only the Tet repressor, then use that cell line to create a second cell line that will express your gene of interest from the inducible expression plasmid. Alternatively, you can transfect with both plasmids (pcDNA6/TR and inducible expression vector) and dual-select with blasticidin and Zeocin to isolate a single stable cell line expressing both the Tet repressor and your gene of interest.
Three T-REx cell lines that stably express the Tet repressor are available from Invitrogen. If you wish to assay for tetracycline-inducible expression of your gene of interest in 293, HeLa, or U2OS cells, you may want to use one of the T-REx cell lines as the host to establish your double stable cell line. For more information, go to www.invitrogen.com or call Technical Service.
Reminder: When generating a stable cell line expressing the Tet repressor, you will want to select for clones that express the highest levels of Tet repressor to use as hosts for your inducible expression construct. Those clones that express the highest levels of Tet repressor should exhibit the most complete repression of basal transcription of your gene of interest.
Determination of antibiotic sensitivity
To successfully generate a stable cell line expressing the Tet repressor and your protein of interest, you need to determine the minimum concentration of each antibiotic (blasticidin and Zeocin) required to kill your untransfected host cell line. For each antibiotic, test a range of concentrations (see below) to ensure that you determine the minimum concentration necessary for your cell line. Use the protocol below to determine the minimal concentrations of Zeocin and blasticidin required to prevent growth of the parental cell line. Please refer to the Appendix, page 14 for instructions on how to prepare and store blasticidin. Instructions on how to prepare and store Zeocin can be found in the inducible expression vector manual.
  1. Plate or split a confluent plate so the cells will be approximately 25% confluent. For each antibiotic, prepare a set of 6-7 plates. Add the following concentrations of antibiotic to each plate in a set:
    • For blasticidin selection, test 0, 1, 3, 5, 7.5, and 10 µg/ml blasticidin
    • For Zeocin selection, test 0, 50, 125, 250, 500, 750, and 1000 µg/ml Zeocin
  2. Replenish the selective media every 3-4 days, and observe the percentage of surviving cells.
  3. Count the number of viable cells at regular intervals to determine the appropriate concentration of antibiotic that prevents growth within 1-2 weeks after addition of the antibiotic.
Effect of Zeocin on sensitive and resistant cells
Zeocin's method of killing is quite different from other antibiotics including blasticidin, neomycin, and hygromycin. Cells do not round up and detach from the plate. Sensitive cells may exhibit the following morphological changes upon exposure to Zeocin:
  • Vast increase in size (similar to the effects of cytomegalovirus infecting permissive cells)
  • Abnormal cell shape
  • Presence of large empty vesicles in the cytoplasm (breakdown of the endoplasmic reticulum and golgi apparatus, or other scaffolding proteins)
  • Breakdown of plasma and nuclear membrane (appearance of many holes in these membranes)
  • Eventually, these "cells" will completely break down and only "strings" of protein remain.
  • Zeocin-resistant cells should continue to divide at regular intervals to form distinct colonies. There should not be any distinct morphological changes in Zeocin-resistant cells when compared to cells not under selection with Zeocin. For more information about Zeocin and its mechanism of action, please refer to the inducible expression vector manual.
Possible sites for linearization of pcDNA6/TR 
To obtain stable transfectants, you may choose to linearize pcDNA6/TR before transfection. While linearizing the vector may not improve the efficiency of transfection, it increases the chances that the vector does not integrate in a way that disrupts the Tet repressor gene or other elements necessary for expression in mammalian cells. The table below lists unique sites that may be used to linearize your construct prior to transfection. Other restriction sites are possible. 
Enzyme
Restriction site (bp)
Location
Supplier
Bst1107 I
4470
Backbone
AGS *, Fermentas, Takara
Sap I
4733
Backbone
New England Biolabs
BspLU11 I
4849
Backbone
Boehringer-Mannheim
Eam1105 I
5739
Ampicillin gene
AGS *, Fermentas, Takara
Fsp I
5961
Ampicillin gene
Many
Selection of stable integrants
Once you have determined the appropriate antibiotic concentrations to use for selection, you can generate a stable cell line expressing pcDNA6/TR and your inducible expression construct.
  1. Transfect mammalian cells with pcDNA6/TR using the desired protocol. Remember to include a plate of untransfected cells as a negative control.
  2. 24 hours after transfection, wash the cells and add fresh medium to the cells.
  3. 48 hours after transfection, split the cells into fresh medium containing blasticidin at the pre-determined concentration required for your cell line. Split the cells such that the cells are no more than 25% confluent. If the cells are too dense, the antibiotic will not kill the cells. Antibiotics work best on actively dividing cells.
  4. Feed the cells with selective medium every 3-4 days until foci can be identified.
  5. Pick at least 20 foci and expand them to test for tetracycline-inducible gene expression by transiently transfecting with the positive control plasmid expressing beta-galactosidase. Screen for those clones that exhibit the lowest levels of basal transcription and the highest levels of β-galactosidase expression after addition of tetracycline.
  6. Once you have obtained a stable cell line expressing the Tet repressor, you can use this cell line to isolate a stable cell line expressing your gene of interest from the inducible expression plasmid. Repeat Steps 1 through 4, above, using your inducible expression vector construct and Zeocin to select foci. Remember to maintain your cells in medium containing blasticidin as well.
  7. Pick and expand at least 20 foci to test for tetracycline-regulated gene expression.
Dual selection of stable integrants
We recommend using the protocol above to generate double stable cell lines. However, if you wish to perform dual selection, you may cotransfect both pcDNA6/TR and your inducible expression construct into the cell line of choice. When cotransfecting your cell line, we recommend using a ratio of at least 6:1 (w/w) pcDNA6/TR DNA:inducible expression plasmid DNA to increase the chances that your double stable cell lines will express sufficient levels of Tet repressor. Following the steps listed above, add selective medium containing blasticidin and Zeocin at the concentrations determined previously. Screen at least 40 foci for tetracycline-regulated expression of your gene of interest using an appropriate assay for your protein (e.g. western blot, enzymatic assay). 
Once you have isolated stable cell lines expressing both pcDNA6/TR and your inducible expression construct and have tested for tetracycline-inducible expression of your gene of interest, we recommend that you perform a time course of tetracycline induction to optimize expression of your protein of interest (e.g. 0, 8, 16, 24, 32 hours, etc.). Use the appropriate concentration of tetracycline for your cell line as previously determined. We have observed as much as 120-fold induction of β-galactosidase in a double stable cell line (T-REx 293 cells transfected with the positive control plasmid containing the lacZ gene) after 24 hours of treatment with 1 µg/ml tetracycline. Please note that your induction levels may vary depending on the nature of your gene of interest and the particular clone that you choose.
pcDNA6/TR Vector
Map of pcDNA6/TR
pcDNA6/TR is a 6662 bp vector that expresses the Tet repressor under the control of the human CMV promoter. A figure summarizing the features of this vector can be downloaded by clicking here. The complete sequence for pcDNA6/TR is available for downloading from www.thermofisher.com or by contacting Technical Service.
TetR gene
The TetR gene used in pcDNA6/TR was originally isolated from the Tn10 transposon which confers resistance to tetracycline in E. coli and other enteric bacteria (Postle et al., 1984). The TetR gene from Tn10 encodes a class B Tet repressor and is often referred to as TetR(B) in the literature (Hillen and Berens, 1994).  
The TetR gene encodes a repressor protein of 207 amino acids with a calculated molecular weight of 23 kDa. For more information about the Tet repressor and its interaction with the Tet operator, please refer to the review by Hillen and Berens (1994).
Features of pcDNA6/TR
The table below describes the relevant features of pcDNA6/TR. The vector includes the rabbit β-globin intron II to enhance expression of the TetR gene. For a more detailed description of the TetR gene and the Tet repressor, please refer to the previous page. All features have been functionally tested.
FeatureBenefit
Human cytomegalovirus (CMV) immediate early promoter
Permits high-level expression of the TetR gene (Andersson et al., 1989; Boshart et al., 1985; Nelson et al., 1987)
Rabbit β-globin intron II (IVS)Enhances expression of the TetR gene (van Ooyen et al., 1979) in cultured cells
TetR geneEncodes the Tet repressor that binds to tet operator sequences to repress transcription of the gene of interest in the absence of tetracycline (Postle et al., 1984; Yao et al., 1998)
SV40 early polyadenylation signalPermits efficient transcription termination and polyadenylation of mRNA
f1 originAllows rescue of single-stranded DNA
SV40 early promoter and originAllows efficient, high-level expression of the blasticidin resistance gene in mammalian cells and episomal replication in cells expressing SV40 large T antigen 
EM7 promoterSynthetic prokaryotic promoter for expression of the blasticidin resistance gene in E. coli
Blasticidin (bsd) resistance geneAllows selection of stable transfectants in mammalian cells (Kimura et al., 1994)
SV40 early polyadenylation signalAllows efficient transcription termination and polyadenylation of mRNA
pUC originPermits high-copy number replication and growth in E. coli
bla promoterAllows expression of the ampicillin (bla) resistance gene
Ampicillin ( bla) resistance gene
(ß-lactamase)
Allows selection of transformants in E. coli
Blasticidin
Blasticidin S HCl is a nucleoside antibiotic isolated from Streptomyces griseochromogenes which inhibits protein synthesis in both prokaryotic and eukaryotic cells (Takeuchi et al., 1958; Yamaguchi et al., 1965). Resistance is conferred by expression of either one of two blasticidin S deaminase genes: bsd from Aspergillus terreus (Kimura et al., 1994) or bsr from Bacillus cereus (Izumi et al., 1991). These deaminases convert blasticidin S to a non-toxic deaminohydroxy derivative (Izumi et al., 1991).
Molecular weight, formula, and structure
The formula for blasticidin S is C17H26N8O5-HCl, and the molecular weight is 458.9. The diagram below shows the structure of blasticidin.
Handling Blasticidin
Always wear gloves, mask, goggles, and protective clothing (e.g. a laboratory coat) when handling blasticidin. Weigh out blasticidin and prepare solutions in a hood.
Preparing and storing stock solutions
Blasticidin is supplied in the T-REx Complete Kit, but may also be obtained separately from Invitrogen (Catalog no. R210-01) in 50 mg aliquots. Blasticidin is soluble in water. Sterile water is generally used to prepare stock solutions of 5 to 10 mg/ml.
  • Dissolve blasticidin in sterile water and filter-sterilize the solution.
  • Aliquot in small volumes suitable for one time use (see next to last point below) and freeze at -20°C for long-term storage or store at +4°C for short-term storage.
  • Aqueous stock solutions are stable for 1-2 weeks at +4°C and 6-8 weeks at -20°C.
  • pH of the aqueous solution should be 7.5 to prevent inactivation of blasticidin.
  • Do not subject stock solutions to freeze/thaw cycles (do not store in a frost-free freezer).
  • Upon thawing, use what you need and store the thawed stock solution at +4°C for up to 2 weeks.
  • Containing blasticidin may be stored at +4°C for up to 2 weeks.
References
  1. Abruzzese, R. V., Godin, D., Burcin, M., Mehta, V., French, M., Li, Y., O'Malley, B. W., and Nordstrom, J. L. (1999). Ligand-Dependent Regulation of Plasmid-Based Transgene Expression in Vivo. Human Gene Therapy 10, 1499-1507.
  2. Ausubel, F. M., Brent, R., Kingston, R. E., Moore, D. D., Seidman, J. G., Smith, J. A., and Struhl, K. (1994). Current Protocols in Molecular Biology (New York: Greene Publishing Associates and Wiley-Interscience).
  3. Baeuerle, P. A. (1991). The Inducible Transcription Activator NF-kappa B: Regulation by Distinct Protein Subunits. Biochim. Biophys. Acta 1072, 63-80.
  4. Baeuerle, P. A., and Baltimore, D. (1988). I Kappa B: A Specific Inhibitor of the NF-kappa B Transcription Factor. Science 242, 540-546.
  5. Baron, M., Reynes, J. P., Stassi, D., and Tiraby, G. (1992). A Selectable Bifunctional b-Galactosidase: Phleomycin-resistance Fusion Protein as a Potential Marker for Eukaryotic Cells. Gene 114, 239-243.
  6. Baulieu, E. E. (1989). Contragestion and Other Clinical Applications of RU486, an Antiprogesterone at the Receptor. Science 245, 1351-1357.
  7. Burcin, M. M., Schiedner, G., Kochanek, S., Tsai, S. Y., and O'Malley, B. W. (1999). Adenovirus-Mediated Regulable Target Gene Expression in vivo. Proc. Natl. Acad. Sci. USA 96, 355-360.
  8. Calmels, T., Parriche, M., Burand, H., and Tiraby, G. (1991). High Efficiency Transformation of Tolypocladium geodes Conidiospores to Phleomycin Resistance. Curr. Genet. 20, 309-314.
  9. Carey, M., Kakidani, H., Leatherwood, J., Mostashari, F., and Ptashne, M. (1989). An Amino-Terminal Fragment of GAL4 Binds DNA as a Dimer. J. Mol. Biol. 209, 423-432.
  10. Chen, C., and Okayama, H. (1987). High-Efficiency Transformation of Mammalian Cells by Plasmid DNA. Mol. Cell. Biol. 7, 2745-2752.
  11. Chu, G., Hayakawa, H., and Berg, P. (1987). Electroporation for the Efficient Transfection of Mammalian Cells with DNA. Nucleic Acids Res. 15, 1311-1326.
  12. DeFranco, D. B. (1998) Subcellular and Subnuclear Trafficking of Steroid Receptors. In Molecular Biology of Steroid and Nuclear Hormone Receptors, L. P. Freedman, ed. (Boston, MA: Birkhauser), pp. 19-34.
  13. Deloukas, P., and Loon, A. P. V. (1993). Genomic Organization of the Gene Encoding the p65 Subunit of NF-kappa B: Multiple Variants of the p65 Protein May be Generated by Alternative Splicing. Hum. Mol. Genet. 2, 1895-1900.
  14. Drocourt, D., Calmels, T. P. G., Reynes, J. P., Baron, M., and Tiraby, G. (1990). Cassettes of the Streptoalloteichus hindustanus ble Gene for Transformation of Lower and Higher Eukaryotes to Phleomycin Resistance. Nucleic Acids Res. 18, 4009.
  15. Evans, R. M. (1988). The Steroid and Thyroid Hormone Receptor Superfamily. Science 240, 889-895.
  16. Felgner, P. L., Holm, M., and Chan, H. (1989). Cationic Liposome Mediated Transfection. Proc. West. Pharmacol. Soc. 32, 115-121.
  17. Felgner, P. L. a., and Ringold, G. M. (1989). Cationic Liposome-Mediated Transfection. Nature 337, 387-388.
  18. Gasc, J. M., Delahaye, F., and Baulieu, E. E. (1989). Compared Intracellular Localization of the Glucocorticosteroid and Progesterone Receptors: An Immunocytochemical Study. Exp. Cell Res. 181, 492-504.
  19. Giniger, E., Varnum, S. M., and Ptashne, M. (1985). Specific DNA Binding of GAL4, a Positive Regulatory Protein of Yeast. Cell 40, 767-774.
  20. Goodwin, E. C., and Rottman, F. M. (1992). The 3´-Flanking Sequence of the Bovine Growth Hormone Gene Contains Novel Elements Required for Efficient and Accurate Polyadenylation. J. Biol. Chem. 267, 16330-16334.
  21. Gritz, L., and Davies, J. (1983). Plasmid-Encoded Hygromycin-B Resistance: The Sequence of Hygromycin-B-Phosphotransferase Gene and its Expression in E. coli and S. cerevisiae. Gene 25, 179-188.
  22. Guiochon-Mantel, A., Loosfelt, H., Lescop, P., Sar, S., Atger, M., Perrot-Applanat, M., and Milgrom, E. (1989). Mechanisms of Nuclear Localization of the Progesterone Receptor: Evidence for Interaction between Monomers. Cell 57, 1147-1154.
  23. Kastner, P., Krust, A., Turcotte, B., Stropp, U., Tora, L., Gronemeyer, H., and Chambon, P. (1990). Two Distinct Estrogen-Regulated Promoters Generate Transcripts Encoding the Two Functionally Different Human Progesterone Receptor Forms A and B. EMBO J. 9, 1603-1614.
  24. Keegan, L., Gill, G., and Ptashne, M. (1986). Separation of DNA Binding from the Transcription-Activating Function of a Eukaryotic Regulatory Protein. Science 231, 699-704.
  25. Kozak, M. (1987). An Analysis of 5´-Noncoding Sequences from 699 Vertebrate Messenger RNAs. Nucleic Acids Res. 15, 8125-8148.
  26. Kozak, M. (1991). An Analysis of Vertebrate mRNA Sequences: Intimations of Translational Control. J. Cell Biology 115, 887-903.
  27. Kozak, M. (1990). Downstream Secondary Structure Facilitates Recognition of Initiator Codons by Eukaryotic Ribosomes. Proc. Natl. Acad. Sci. USA 87, 8301-8305.
  28. Laughon, A., and Gesteland, R. F. (1984). Primary Structure of the Saccharomyces cerevisiae GAL4 Gene. Mol. Cell. Biol. 4, 260-267.
  29. Lillie, J. W., and Green, M. R. (1989). Transcription Activation by the Adenovirus E1a Protein. Nature 338, 39-44.
  30. Lindner, P., Bauer, K., Krebber, A., Nieba, L., Kremmer, E., Krebber, C., Honegger, A., Klinger, B., Mocikat, R., and Pluckthun, A. (1997). Specific Detection of His-tagged Proteins With Recombinant Anti-His Tag scFv-Phosphatase or scFv-Phage Fusions. BioTechniques 22, 140-149.
  31. Marmorstein, R., Carey, M., Ptashne, M., and Harrison, S. C. (1992). DNA Recognition by GAL4: Structure of a Protein-DNA Complex. Nature 356, 408-414.
  32. McKnight, S. L. (1980). The Nucleotide Sequence and Transcript Map of the Herpes Simplex Virus Thymidine Kinase Gene. Nucleic Acids Res. 8, 5949-5964.
  33. Miller, J. H. (1972). Experiments in Molecular Genetics (Cold Spring Harbor, New York: Cold Spring Harbor Laboratory).
  34. Misrahi, M., Atger, M., d'Auriol, L., Loosfelt, H., Meriel, C., Fridlansky, F., Guiochon-Mantel, A., Galibert, F., and Milgrom, E. (1987). Complete Amino Acid Sequence of the Human Progesterone Receptor Deduced from Cloned cDNA. Biochem. Biophys. Res. Commun. 143, 740-748.
  35. Mulsant, P., Tiraby, G., Kallerhoff, J., and Perret, J. (1988). Phleomycin Resistance as a Dominant Selectable Marker in CHO Cells. Somat. Cell Mol. Genet. 14, 243-252.
  36. Nelson, M., and Silver, P. (1989). Context Affects Nuclear Protein Localization in Saccharomyces cerevisiae. Mol. Cell. Biol. 9, 384-389.
  37. Palmer, T. D., Hock, R. A., Osborne, W. R. A., and Miller, A. D. (1987). Efficient Retrovirus-Mediated Transfer and Expression of a Human Adenosine Deaminase Gene in Diploid Skin Fibroblasts from an Adenosine-Deficient Human. Proc. Natl. Acad. Sci. U.S.A. 84, 1055-1059.
  38. Perez, P., Tiraby, G., Kallerhoff, J., and Perret, J. (1989). Phleomycin Resistance as a Dominant Selectable Marker for Plant Cell Transformation. Plant Mol. Biol. 13, 365-373.
  39. Perrot-Applanat, M., Logeat, F., Groyer-Picard, M. T., and Milgrom, E. (1985). Immunocytochemical Study of Mammalian Progesterone Receptor Using Monoclonal Antibodies. Endocrinol. 116, 1473-1484.
  40. Philibert, D., Moguilewsky, M., Mary, I., Lecaque, D., Tournemine, C., Secchi, J., and Deraedt, R. (1985). In The Antiprogestin Steroid RU486 and Human Fertility Control, E. E. Baulieu and S. J. Segal, eds. (New York: Plenum), pp. 49-68.
  41. Ruben, S. M., Dillon, P. J., Schreck, R., Henkel, T., Chen, C. H., Maher, M., Baeuerle, P. A., and Rosen, C. A. (1991). Isolation of a Rel-related Human cDNA that Potentially Encodes the 65-kD Subunit of NF-kappa B. Science 251, 1490-1493.
  42. Sambrook, J., Fritsch, E. F., and Maniatis, T. (1989). Molecular Cloning: A Laboratory Manual, Second Edition (Plainview, New York: Cold Spring Harbor Laboratory Press).
  43. Schmitz, M. L., and Baeuerle, P. A. (1991). The p65 Subunit is Responsible for the Strong Transcription Activating Potential of NF-kappa B. EMBO J. 10, 3805-3817.
  44. Shigekawa, K., and Dower, W. J. (1988). Electroporation of Eukaryotes and Prokaryotes: A General Approach to the Introduction of Macromolecules into Cells. BioTechniques 6, 742-751.
  45. Simons, S. S., Jr. (1998) Structure and Function of the Steroid and Nuclear Receptor Ligand Binding Domain. In Molecular Biology of Steroid and Nuclear Hormone Receptors, L. P. Freedman, ed. (Boston, MA: Birkhauser), pp. 35-104.
  46. Southern, J. A., Young, D. F., Heaney, F., Baumgartner, W., and Randall, R. E. (1991). Identification of an Epitope on the P and V Proteins of Simian Virus 5 That Distinguishes Between Two Isolates with Different Biological Characteristics. J. Gen. Virol. 72, 1551-1557.
  47. Truss, M., and Beato, M. (1993). Steroid Hormone Receptors: Interaction with Deoxyribonucleic Acid and Transcription Factors. Endocr. Rev. 14, 459-479.
  48. Vegeto, E., Allan, G. F., Schrader, W. T., Tsai, M. J., McDonnell, D. P., and O'Malley, B. W. (1992). The Mechanism of RU486 Antagonism is Dependent on the Conformation of the Carboxy-Terminal Tail of the Human Progesterone Receptor. Cell 69, 703-713.
  49. Wang, Y., B.W. O'Malley, J., Tsai, S. Y., and O'Malley, B. W. (1994). A Regulatory System for Use in Gene Transfer. Proc. Natl. Acad. Sci. USA 91, 8180-8184.
  50. Wang, Y., Xu, J., Pierson, T., O'Malley, B. W., and Tsai, S. Y. (1997). Positive and Negative Regulation of Gene Expression in Eukaryotic Cells with an Inducible Transcriptional Regulator. Gene Therapy 4, 432-441.
  51. Wigler, M., Silverstein, S., Lee, L.-S., Pellicer, A., Cheng, Y.-C., and Axel, R. (1977). Transfer of Purified Herpes Virus Thymidine Kinase Gene to Cultured Mouse Cells. Cell 11, 223-232.
  52. Ylikomi, T., Bocquel, M. T., Berry, M., Gronemeyer, H., and Chambon, P. (1992). Cooperation of Proto-Signals for Nuclear Accumulation of Estrogen and Progesterone Receptors. EMBO J. 11, 3681-3694.
LT054