protocol
Introduction
The BenchMark™ Pre-Stained Protein Ladder is designed for use in SDSPAGE using Tris-Glycine gels for estimation of protein molecular weight, and evaluation of western transfer efficiency. Covalently coupled dyes allows easy visualization of the ladder during, and after electrophoresis.
Features of the BenchMark™ Pre-Stained Protein Ladder:
  • Consists of 10 pre-stained protein bands in the range of 6–180 kDa (proteins are covalently coupled to a blue or pink dye)
  • Includes a protein coupled to a pink dye (fourth band from the top) to ensure proper identification of each protein band
  • Supplied in a ready-to-use format
Note:
Coupling of the chromophores to the proteins affects the apparent molecular weight of proteins in SDS-PAGE relative to unstained standards. Use pre-stained protein ladders to determine the approximate molecular weight of proteins. For accurate molecular weight estimation of proteins, use unstained protein ladders.
Specifications
Contents: 2 × 250 μl of BenchMark™ Pre-Stained Protein Ladder
Storage Buffer: 50 mM Tris-HCl, pH 6.8; 5 mM EDTA; 10 mM DTT;
1% (w/v) SDS; 10% (w/v) glycerol
Storage: Store at –20°C
Stability: 1 year at –20°C
Ordering Information
Catalog # Name Size Price (USD) Qty
10748010 BenchMark™ Pre-stained Protein Ladder Each
263.65
Online Exclusive
280.00
Save 16.35 (6%)
Protocol
Directions
The BenchMark™ Pre-Stained Protein Ladder is supplied in a ready-to-use format.
Do not heat or add reducing agent to the ladder.
  1. Thaw the ladder at room temperature.
  2. Vortex gently to ensure the solution is homogenous.
  3. Load the ladder into the well of an SDS gel using the loading volumes listed below to obtain the best results:
GelElectrophoresisBlotting
Mini-Gel10 μl5 μl
Standard Gel20 μl10 μl
MAN0000876             27-Jul-2009