Introduction
The Invitrogen Novex SeeBlue Plus2 Pre-Stained Standard allows easy visualization of protein molecular weight ranges during electrophoresis and evaluation of western transfer efficiency. The important features of the standard are listed below:
  • Consists of 10 pre-stained protein bands (8 blue and 2 contrasting colors) in the range of 4-250 kDa
  • Suitable for Tris-Glycine, Tricine, and Invitrogen NuPAGE Novex Gels
Ordering Information
Catalog # Name Size Price (USD) Qty
LC5925 SeeBlue™ Plus2 Pre-stained Protein Standard Each
248.00
Protocol
The SeeBlue Plus2 Pre-Stained Standard is supplied ready-to-use. There is no need to heat or add reducing agent.
Use loading volumes listed below to obtain the best results:
GelElectrophoresisBlotting
Mini-Gel (1.0 mm thick)10 µl3 µl
Mini-Gel (1.5 mm thick)20 µl5 µl
Standard Gel (1.0 mm thick)20 µl7 µl
Standard Gel (1.5 mm thick)30 µl10 µl
Example
The apparent molecular weights of the protein bands in SeeBlue Plus2 Pre-Stained Standard in several buffer systems is shown below. The protein bands have different mobilities in various SDS-PAGE buffer systems.
Specifications
Contents:  500 µl of SeeBlue Plus2 Pre-Stained Standard
Loading Buffer:  Tris-HCl, Formamide, SDS, Phenol Red
Storage:  Store at 4°C
Stability:  4 months at 4°C
LT059