Technical Resources
10569 - DMEM, high glucose, GlutaMAX(TM), pyruvate
ComponentsMolecular WeightConcentration (mg/L)mM
Amino Acids
Glycine75.030.00.4
L-Alanyl-Glutamine217.0862.03.9723501
L-Arginine hydrochloride211.084.00.39810428
L-Cystine 2HCl313.063.00.20127796
L-Histidine hydrochloride-H2O210.042.00.2
L-Isoleucine131.0105.00.8015267
L-Leucine131.0105.00.8015267
L-Lysine hydrochloride183.0146.00.7978142
L-Methionine149.030.00.20134228
L-Phenylalanine165.066.00.4
L-Serine105.042.00.4
L-Threonine119.095.00.79831934
L-Tryptophan204.016.00.078431375
L-Tyrosine disodium salt dihydrate261.0104.00.39846742
L-Valine117.094.00.8034188
Vitamins
Choline chloride140.04.00.028571429
D-Calcium pantothenate477.04.00.008385744
Folic Acid441.04.00.009070295
Niacinamide122.04.00.032786883
Pyridoxine hydrochloride206.04.00.019417476
Riboflavin376.00.40.0010638298
Thiamine hydrochloride337.04.00.011869436
i-Inositol180.07.20.04
Inorganic Salts
Calcium Chloride (CaCl2) (anhyd.)111.0200.01.8018018
Ferric Nitrate (Fe(NO3)3"9H2O)404.00.12.4752476E-4
Magnesium Sulfate (MgSO4) (anhyd.)120.097.670.8139166
Potassium Chloride (KCl)75.0400.05.3333335
Sodium Bicarbonate (NaHCO3)84.03700.044.04762
Sodium Chloride (NaCl)58.06400.0110.344826
Sodium Phosphate monobasic (NaH2PO4-H2O)138.0125.00.9057971
Other Components
D-Glucose (Dextrose)180.04500.025.0
Phenol Red376.415.00.039851222
Sodium Pyruvate110.0110.01.0
Reference:
1. Dulbecco, R. and Freeman, G. (1959) Virology 8:396. 
Related Products:
10566 - DMEM, high glucose, GlutaMAX(TM)
Cat No: 10566016 , 10566024 , 10566032
10564 - DMEM, high glucose, GlutaMAX(TM), HEPES
Cat No: 10564011 , 10564029 , 10564037 , 10564045
31966 - DMEM, high glucose, GlutaMAX(TM), pyruvate
Cat No: 31966021 , 31966047
32430 - DMEM, high glucose, GlutaMAX(TM), HEPES
Cat No: 32430027 , 32430100
61965 - DMEM, high glucose, GlutaMAX(TM)
Cat No: 61965026 , 61965059 , 61965240