Mouse fibroblasts. MitoTracker Red CMXRos, BODIPY FL phallacidin and DAPI.
Four-panel composite image of mouse fibroblasts that were incubated with MitoTracker Red CMXRos (M7512), and then formaldehyde-fixed, acetone-permeabilized and stained with the F-actin–specific probe, BODIPY FL phallacidin (B607) and with DAPI (D1306, D3571, D21490). Images were obtained by taking single and multiple exposures through bandpass optical filter sets appropriate for fluorescein, the Texas Red dye and DAPI.
Four-panel composite image of mouse fibroblasts that were incubated with MitoTracker Red CMXRos (M7512), and then formaldehyde-fixed, acetone-permeabilized and stained with the F-actin–specific probe, BODIPY FL phallacidin (B607) and with DAPI (D1306, D3571, D21490). Images were obtained by taking single and multiple exposures through bandpass optical filter sets appropriate for fluorescein, the Texas Red dye and DAPI.
Related Products
Catalog # Name Size Price (USD) Qty
D1306 DAPI (4',6-diamidino-2-phenylindole, dihydrochloride), 10 mg, 10 mg Each
152.65
Online Exclusive
156.00
Save 3.35 (2%)
D21490 DAPI, FluoroPure™ grade, 10 mg, 10 mg Each
225.65
Online Exclusive
231.00
Save 5.35 (2%)
D3571 DAPI (4',6-diamidino-2-phenylindole, dilactate), 10 mg, 10 mg Each
151.65
Online Exclusive
156.00
Save 4.35 (3%)
M7512 MitoTracker™ Red CMXRos - Special Packaging, 20 x 50 μg, 20 x 50 μg Each
506.65
Online Exclusive
522.00
Save 15.35 (3%)
Related Images