Search Thermo Fisher Scientific
Thermo Scientific Nunc cell culture plastics offer a solution for every step of your workflow. Whether you need cell culture-treated plates for imaging or low-attachment dishes for your 3D work, easily find your cell cultureware by searching by surface, format, or other application needs.
Explore Nunc cultureware products Find Nunc equivalent Request sample/quote
To optimize results for different cell culture types and their subsequent cellular analysis, a range of cell culture surfaces, spanning a variety of formats, are available.
We’ve tested the most used cell lines on Nunc surface-treated cultureware side-by-side with leading competitors. With this data, you can research how specific cell lines thrive on various surface treatments. New cell data is being added continuously.
Choose from round, flat-bottom, or conical tubes with a variety of sizes and features—including recyclable racks and ease-of-use designs for enhanced performance.
Choose cell culture dishes in a wide selection of formats, materials, and surface modifications. To promote cell health and reproducibility, each dish is designed and produced with highest quality standards.
Choose tissue culture flasks from a variety of sizes, with culture areas ranging from 25 cm2 to 500 cm2 as well as a selection of surface-treated offerings to address your specific cell type and application.
Thermo Scientific Nunc cell culture plates and well plates are available in more than 100 combinations of format and surface, designed to help promote healthy cells and reproducibility. Lot-to-lot cell line testing helps to ensure consistency.
These inserts’ porous membranes and unique structures allow your cell’s attached surface to be simultaneously exposed to growth media and your experiment’s variable condition(s).
Cell culture imaging products offer comprehensive solutions to meet your needs in cell imaging applications, including fluorescence microscopy, phase-contrast microscopy, confocal microscopy, live-cell imaging, DIC, and FISH.
Healthy adherent cell growth requires quality treated plastics. Nunclon Delta cultureware features a more hydrophilic polystyrene surface modification, further promoting maximum adhesion for a broad range of cell types.
Learn how Gibco media and Nunc cultureware work better together
Plastics with Nunclon Delta surfaces endure rigorous cloning efficiency tests using cell lines chosen for their high toxin sensitivity. Every lot is tested and certified for cell growth and plating efficiency using L-929, HEL 299, V79-4, and primary chick embryo cells. In all but one cell line, this growth is assessed by the cultureware surface coverage percentage.
For V79-4 lines, cell growth is evaluated by the number of colonies formed per cultureware. For Nunclon Delta lot certification, cultureware must demonstrate:
Get to know the facts about your cell culture-treated plastics by using our Nunclon Surface cell line data. We've tested the most-used cell lines on Nunclon Delta surfaces alongside leading competitors, validating Nunclon Delta surface's place in your cell culture hood.
Compatible with 40+ cell lines, Thermo Scientific Nunc dishes, flasks and plates with Nunclon Supra surface show improved yield and morphology for primary cells and MSCs while enabling xeno-free or low-serum research applications.
Try the Nunclon Supra surface today: Dishes | Plates | Flasks | 96-well plates
Find out more about the Nunclon Supra surface today at thermofisher.com/supra
Collagen I, ordinarily found in the extracellular matrix (ECM), is rat-tail-tendon sourced, whereas poly-d-lysine (PDL) is fully a synthetic alternative. These ECM-coated surfaces mimic the body’s growth environment, making them ideal for finicky cells that have difficulties growing on regular tissue culture surfaces.
Explore PDL/collagen I multidishes
Explore PDL/collagen I EasYFlask products
The Lab-Tek II CC2 surface treatments mimics poly-D-lysine and aids in fastidious cell attachment. For cells such as neurons, these chamber slides provide positively charged growth surfaces with enhanced stability.
Nunc UpCell surfaces enable the harvesting of cells in single-cell suspension or as contiguous cell sheets via temperature reduction, eliminating the need for enzymatic cell dissociates such as trypsin. In addition, this surface can enable the creation of 3D tissue models without foreign-to-the-body scaffolding materials.
Explore Nunc UpCell surfaces for non-enzymatic cell dissociation
This 3D-cell-culture-optimized surface nearly eliminates cultureware cell attachment, enabling 3D spheroid and organoid cell-to-cell aggregation through naturally secreted extracellular matrices (ECM). This Thermo Scientific Nunclon Sphera culture system is designed for spheroid culture (e.g., 3D tumor spheroids) and organoid culture (e.g., brain organoids) and is available in a wide selection of formats, including plates, dishes, and flasks.
Healthy adherent cell growth requires quality treated plastics. Nunclon Delta cultureware features a more hydrophilic polystyrene surface modification, further promoting maximum adhesion for a broad range of cell types.
Learn how Gibco media and Nunc cultureware work better together
Plastics with Nunclon Delta surfaces endure rigorous cloning efficiency tests using cell lines chosen for their high toxin sensitivity. Every lot is tested and certified for cell growth and plating efficiency using L-929, HEL 299, V79-4, and primary chick embryo cells. In all but one cell line, this growth is assessed by the cultureware surface coverage percentage.
For V79-4 lines, cell growth is evaluated by the number of colonies formed per cultureware. For Nunclon Delta lot certification, cultureware must demonstrate:
Get to know the facts about your cell culture-treated plastics by using our Nunclon Surface cell line data. We've tested the most-used cell lines on Nunclon Delta surfaces alongside leading competitors, validating Nunclon Delta surface's place in your cell culture hood.
Compatible with 40+ cell lines, Thermo Scientific Nunc dishes, flasks and plates with Nunclon Supra surface show improved yield and morphology for primary cells and MSCs while enabling xeno-free or low-serum research applications.
Try the Nunclon Supra surface today: Dishes | Plates | Flasks | 96-well plates
Find out more about the Nunclon Supra surface today at thermofisher.com/supra
Collagen I, ordinarily found in the extracellular matrix (ECM), is rat-tail-tendon sourced, whereas poly-d-lysine (PDL) is fully a synthetic alternative. These ECM-coated surfaces mimic the body’s growth environment, making them ideal for finicky cells that have difficulties growing on regular tissue culture surfaces.
Explore PDL/collagen I multidishes
Explore PDL/collagen I EasYFlask products
The Lab-Tek II CC2 surface treatments mimics poly-D-lysine and aids in fastidious cell attachment. For cells such as neurons, these chamber slides provide positively charged growth surfaces with enhanced stability.
Nunc UpCell surfaces enable the harvesting of cells in single-cell suspension or as contiguous cell sheets via temperature reduction, eliminating the need for enzymatic cell dissociates such as trypsin. In addition, this surface can enable the creation of 3D tissue models without foreign-to-the-body scaffolding materials.
Explore Nunc UpCell surfaces for non-enzymatic cell dissociation
This 3D-cell-culture-optimized surface nearly eliminates cultureware cell attachment, enabling 3D spheroid and organoid cell-to-cell aggregation through naturally secreted extracellular matrices (ECM). This Thermo Scientific Nunclon Sphera culture system is designed for spheroid culture (e.g., 3D tumor spheroids) and organoid culture (e.g., brain organoids) and is available in a wide selection of formats, including plates, dishes, and flasks.
Quality involves all aspects of our organization from product development, choice of raw material, through production to customer service. Our cell culture plastics are manufactured per established quality standards and current Good Manufacturing Practices (cGMP) to help ensure product performance. Our quality system is routinely audited to ensure that we consistently maintain the highest standards.
Our registered ISO-certified and state-of-the-art facilities are strategically located around the world to support our customers.
For Research Use Only. Not for use in diagnostic procedures.