1x1 image pixel for data collection
Nunc™ Microwell 96-Well Microplate with Nunclon™ Supra Surface
Save 51% on plasticware essentials and stock your lab to culture with confidence. View promotion details >
Nunc™ Microwell 96-Well Microplate with Nunclon™ Supra Surface
Thermo Scientific™
Nunc™ Microwell 96-Well Microplate with Nunclon™ Supra Surface
Experience optimal performance with Thermo Scientific Nunc plates with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells.
Promo ImagePromotion
Promo code:RCELL51
Stock your lab with plasticware essentials so that you can culture with confidence
From now until Dec 31, get 51% off various cell culture plasticware like dishes, flasks, plates and tubes that help achieve reproducible results when you spend $1,000Learn More
Have Questions?
Catalog NumberQuantity
167013Promo Image
Catalog number 167013
Price (USD)
472.00
Case of 50
Add to cart
Price (USD)
472.00
Case of 50
Add to cart
Ask our AI about this Product
Experience optimal performance with Thermo Scientific Nunc plates with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.
Thermo Scientific Nunc vessels with Nunclon Supra surface treatment are designed for culturing fastidious adherent cells. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.
The Nunclon Supra surface supports serum-free and coating-free culture of human mesenchymal stromal cells (MSCs), retaining their morphology and promoting high cell viability. Nunclon Supra cultureware is a great xeno-free solution for cell therapy research applications.
Features of Nunc Microwell 96-Well Microplates with Nunclon Supra Surface include:
• Consistent lot-to-lot performance
• Sterile with SAL 10-6
• Non-cytotoxic
• Non-pyrogenic
• Biocompatible per USP Class VI for raw materials
• Shelf life: 5 years
For Research Use Only.
Specifications
DisposableSingle-use
LidWith Lid
No. per Case50 Microplates
No. per Pack1 Microplate
ColorClear
MaterialPolystyrene
No. of Wells96
Product LineNunc
SterilitySterile
Surface TreatmentNunclon Supra
Well ShapeFlat Bottom (F-well)
Unit SizeCase of 50
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Customers who viewed this item also viewed
Documents & Downloads
Certificates
Lot #Certificate TypeDateCatalog Number(s)
60772880Certificate of ComplianceMay 19, 2025167013
60866436Certificate of ComplianceApr 21, 2025167013
2 results displayed, search above for a specific certificate
Share catalog number, name or link