Video Player is loading.
Current Time 0:00
Duration 2:32
Loaded: 6.47%
Stream Type LIVE
Remaining Time 2:32
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Tissue culture tubes
    Choose from round and flat-bottom tubes as well as conical tubes that offer a variety of sizes and features, including recyclable racks and ease-of-use designs for enhanced performance.
    Centrifuge tubes  Storage tubes Request sample/quote
    Cell culture centrifuge tubes
    Approximately 24 conical bottomed centrifuge tube in a blue rack matching their blue caps
    Available in a wide variety of sizes, colors, and formats, Nunc centrifuge tubes are designed to meet a range of needs. Navigate centrifuge offerings below by size.
      Tubes brochure
      Read now how our protein expression systems perform bioreactor tubes
    EZFlip conical centrifuge tubes
    A blue-gloved hand holding an EZFlip tube demonstrating its ability to be opened one handed
    Nunc 15 mL EZFlip conical centrifuge tubes feature:
    • Clarified polypropylene composition
    • Nonpyrogenic and non-cytotoxic guarantee
    • Integrated leakproof cap
    • Writing area for sample identification
    Nunc 50 mL EZFlip conical centrifuge tubes feature:
    • Clarified polypropylene composition
    • Nonpyrogenic and non-cytotoxic guarantee
    • Integrated leakproof cap
    • Writing area for sample identification
    Cat. No.DescriptionSurfaceTube shapeWith rackSterilePack sizeCase 
    362694Nunc 15 mL EZFlip Conical Centrifuge TubesUntreatedConicalNoNo50500Order
    362694NSNunc 15 mL EZFlip Conical Centrifuge Tubes (Non-sterile)UntreatedConicalNoYes50500Order
    362695Nunc 15 mL EZFlip Conical Centrifuge TubesUntreatedConicalYesYes50500Order
    362695NSNunc 15 mL EZFlip Conical Centrifuge Tubes (Non-sterile)UntreatedConicalYesNo50500Order
    362696NSNunc 50 mL EZFlip Conical Centrifuge Tubes (Non-sterile)UntreatedConicalNoNo25500Order
    362697NSNunc 50 mL EZFlip Conical Centrifuge Tubes (Non-sterile)UntreatedConicalNoYes25500Order
    Return to top of tab
    Bioreactor conical centrifuge tubes
    Three bioreactor tubes, one laying on its side
    Nunc 50 mL bioreactor tubes feature:
    • Virgin-polypropylene-resin composition
    • Nonpyrogenic and non-cytotoxic guarantee
    • Leakproof guarantee
    • Four different sizes of ventilation (dial type) on the cap
    • 2 µm PTFE filter membrane
      Read now how our protein expression systems perform bioreactor tubes
    Cat. No.DescriptionSurfaceTube shapeWith rackSterilePack sizeCase 
    332260Nunc 50 mL Bioreactor Tube UntreatedConicalNoYes390Order
    Return to top of tab
    15 mL-500 mL conical centrifuge tubes
    5 conical bottomed centrifuge tube in a blue rack matching their blue caps
    Nunc 15 mL conical centrifuge tubes feature:
    • Virgin-polypropylene-resin composition
    • Nonpyrogenic and non-cytotoxic guarantee
    • Leakproof guarantee
    A white gloved hand on white background holding a conical bottomed 50mL tube
    Nunc 50 mL conical centrifuge tubes feature:
    • Virgin-polypropylene-resin composition
    • Nonpyrogenic and non-cytotoxic guarantee
    • Leakproof guarantee
    Media being pipetted into a single wide-mouthed 250mL conical centrifuge tube
    Nunc 250 mL conical centrifuge tubes feature:
    • Clarified polypropylene composition
    • Nonpyrogenic and non-cytotoxic guarantee
    • Leakproof guarantee
    • Wide-mouth design
    A single 250mL conical centrifuge tube with light blue cap
    250 mL conical centrifuge tubes feature:
    • Clarified polypropylene composition
    • Nonpyrogenic and non-cytotoxic guarantee
    • Leakproof guarantee
    A single 500mL conical centrifuge tube with light blue cap
    500 mL conical centrifuge tubes feature:
    • Clarified polypropylene composition
    • Nonpyrogenic and non-cytotoxic guarantee
    • DNase-/RNase-free
    • BSE-/TSE-free
    Cat. No.DescriptionSurfaceTube shapeWith rackSterilePack sizeCase 
    339650Nunc 15 mL Conical Sterile Polypropylene Centrifuge TubesUntreatedConicalNoYes50500Order
    339651Nunc 15 mL Conical Sterile Polypropylene Centrifuge TubesUntreatedConicalYesYes25500Order
    339652Nunc 50 mL Conical Sterile Polypropylene Centrifuge TubesUntreatedConicalNoYes25500Order
    339653Nunc 50 mL Conical Sterile Polypropylene Centrifuge TubesUntreatedConicalYesYes25300Order
    376814250 mL Conical Centrifuge TubeUntreatedConicalNoYesNA40Order
    332265250 mL Conical Centrifuge TubesUntreatedConicalNoYes648Order
    332270500 mL Conical Centrifuge TubeUntreatedConicalNoYesNA40Order
    Return to top of tab
    EZFlip conical centrifuge tubes
    A blue-gloved hand holding an EZFlip tube demonstrating its ability to be opened one handed
    Nunc 15 mL EZFlip conical centrifuge tubes feature:
    • Clarified polypropylene composition
    • Nonpyrogenic and non-cytotoxic guarantee
    • Integrated leakproof cap
    • Writing area for sample identification
    Nunc 50 mL EZFlip conical centrifuge tubes feature:
    • Clarified polypropylene composition
    • Nonpyrogenic and non-cytotoxic guarantee
    • Integrated leakproof cap
    • Writing area for sample identification
    Cat. No.DescriptionSurfaceTube shapeWith rackSterilePack sizeCase 
    362694Nunc 15 mL EZFlip Conical Centrifuge TubesUntreatedConicalNoNo50500Order
    362694NSNunc 15 mL EZFlip Conical Centrifuge Tubes (Non-sterile)UntreatedConicalNoYes50500Order
    362695Nunc 15 mL EZFlip Conical Centrifuge TubesUntreatedConicalYesYes50500Order
    362695NSNunc 15 mL EZFlip Conical Centrifuge Tubes (Non-sterile)UntreatedConicalYesNo50500Order
    362696NSNunc 50 mL EZFlip Conical Centrifuge Tubes (Non-sterile)UntreatedConicalNoNo25500Order
    362697NSNunc 50 mL EZFlip Conical Centrifuge Tubes (Non-sterile)UntreatedConicalNoYes25500Order
    Return to top of tab
    Bioreactor conical centrifuge tubes
    Three bioreactor tubes, one laying on its side
    Nunc 50 mL bioreactor tubes feature:
    • Virgin-polypropylene-resin composition
    • Nonpyrogenic and non-cytotoxic guarantee
    • Leakproof guarantee
    • Four different sizes of ventilation (dial type) on the cap
    • 2 µm PTFE filter membrane
      Read now how our protein expression systems perform bioreactor tubes
    Cat. No.DescriptionSurfaceTube shapeWith rackSterilePack sizeCase 
    332260Nunc 50 mL Bioreactor Tube UntreatedConicalNoYes390Order
    Return to top of tab
    15 mL-500 mL conical centrifuge tubes
    5 conical bottomed centrifuge tube in a blue rack matching their blue caps
    Nunc 15 mL conical centrifuge tubes feature:
    • Virgin-polypropylene-resin composition
    • Nonpyrogenic and non-cytotoxic guarantee
    • Leakproof guarantee
    A white gloved hand on white background holding a conical bottomed 50mL tube
    Nunc 50 mL conical centrifuge tubes feature:
    • Virgin-polypropylene-resin composition
    • Nonpyrogenic and non-cytotoxic guarantee
    • Leakproof guarantee
    Media being pipetted into a single wide-mouthed 250mL conical centrifuge tube
    Nunc 250 mL conical centrifuge tubes feature:
    • Clarified polypropylene composition
    • Nonpyrogenic and non-cytotoxic guarantee
    • Leakproof guarantee
    • Wide-mouth design
    A single 250mL conical centrifuge tube with light blue cap
    250 mL conical centrifuge tubes feature:
    • Clarified polypropylene composition
    • Nonpyrogenic and non-cytotoxic guarantee
    • Leakproof guarantee
    A single 500mL conical centrifuge tube with light blue cap
    500 mL conical centrifuge tubes feature:
    • Clarified polypropylene composition
    • Nonpyrogenic and non-cytotoxic guarantee
    • DNase-/RNase-free
    • BSE-/TSE-free
    Cat. No.DescriptionSurfaceTube shapeWith rackSterilePack sizeCase 
    339650Nunc 15 mL Conical Sterile Polypropylene Centrifuge TubesUntreatedConicalNoYes50500Order
    339651Nunc 15 mL Conical Sterile Polypropylene Centrifuge TubesUntreatedConicalYesYes25500Order
    339652Nunc 50 mL Conical Sterile Polypropylene Centrifuge TubesUntreatedConicalNoYes25500Order
    339653Nunc 50 mL Conical Sterile Polypropylene Centrifuge TubesUntreatedConicalYesYes25300Order
    376814250 mL Conical Centrifuge TubeUntreatedConicalNoYesNA40Order
    332265250 mL Conical Centrifuge TubesUntreatedConicalNoYes648Order
    332270500 mL Conical Centrifuge TubeUntreatedConicalNoYesNA40Order
    Return to top of tab
    Nunclon Delta culture tubes for fastidious cells
    Nunclon Delta tube surface for adherent cells
    The Nunclon Delta surface treatment for adherent cells, makes standard polystyrene surfaces more hydrophilic, facilitating in maximum adhesion for a range of cell types.
    With this culture tube surface treatment, you can perform adherent cell cultures in biotechnology, pharmaceutical, and research labs as well as with vaccine development and diagnostic kits.
    Explore cell performance with Nunclon Delta surfaces
    Cat. No.DescriptionSurfaceTube shapeCapSterilePack sizeCase 
    156758
    Nunc Cell Culture Tubes (3 mL)
    Nunclon Delta
    FlatScrew capYes75450Order
    145470
    Nunc Cell Culture Tubes (7 mL)
    Nunclon Delta
    RoundPush-onYes100600Order
    146183
    Nunc Cell Culture Tubes (7 mL)
    Nunclon Delta
    RoundScrew capYes100600Order
    Cell culture storage tubes and racks
    Nunc storage tubes for tissue culture
    Available in a wide variety of sizes, colors, and formats Nunc Storage Tubes provide storage tubes for all your needs.  Matrix 2D tube technologies securely store samples at low temperatures, including vapor phase liquid nitrogen. A variety of racks for your choice of storage tube are arranged below by size.
    Order Matrix ScrewTop tube caps, cap trays, and cap inserts
    500 µL storage tubes
    A storage rack holding a variety of 500 µL tubes with various colored caps
    Matrix 500 µL V-bottom storage tubes feature:
    • Virgin-polypropylene-resin composition
    • ScrewTop caps with soft silicone gaskets
    • Stringent 2D barcode quality control
    Order matrix storage tube racks
    Cat. No.DescriptionColorTube shapeWith rackSterilePack sizeCase 
    3743
    Matrix 500 μL ScrewTop Tubes
    Colorless
    V-bottomNoYesNA480Order
    3744
    Matrix 500 μL ScrewTop Tubes
    Colorless
    V-bottomYesYes96/rack480Order
    3743RED
    Matrix 500 μL ScrewTop Tubes
    Red
    V-bottomNoYesNA480Order
    3744RED
    Matrix 500 μL ScrewTop Tubes
    Red
    V-bottomYesYes96/rack480Order
    3743YEL
    Matrix 500 μL ScrewTop Tubes
    Yellow
    V-bottomNoYesNA480Order
    3744YEL
    Matrix 500 μL ScrewTop Tubes
    Yellow
    V-bottomYesYes96/rack480Order
    3743GRE
    Matrix 500μL ScrewTop Tubes
    Green
    V-bottomNoYesNA480Order
    3743BLU
    Matrix 500 μL ScrewTop Tubes
    Blue
    V-bottomNoYesNA480Order
    3743PUR
    Matrix 500 μL ScrewTop Tubes
    Purple
    V-bottomNoYesNA480Order
    3743WHI
    Matrix 500 μL ScrewTop Tubes
    White
    V-bottomNoYesNA480Order
    0.75 mL storage tubes
    A storage rack filled with 0.75 mL tubes
    Matrix 0.75 mL blank and alphanumeric storage tubes feature:
    • Virgin-polypropylene-resin composition
    • Alphanumeric codes on tube bottoms
    • A variety of cap options
    Order matrix storage tube racks
    Cat. No.DescriptionBarcodeWith latch rackSterilePack sizeCase 
    4170
    Matrix Blank and Alphanumeric Storage Tubes
    Blank
    NoNoNA1000Order
    4272
    Matrix Blank and Alphanumeric Storage Tubes
    Blank
    YesYes10/rack960Order
    4273
    Matrix Blank and Alphanumeric Storage Tubes
    Alphanumeric
    YesNo10/rack960Order
    1 mL storage tubes
    A storage rack filled with 1 mL tubes with various cap colors
    Matrix 1 mL V-bottom storage tubes feature:
    • Virgin-polypropylene-resin composition
    • ScrewTop caps with soft silicone gaskets
    • Stringent 2D barcode quality control
    Order matrix storage tube racks
    Cat. No.DescriptionColorTube shapeWith rackSterilePack sizeCase 
    3741
    Matrix 1.0 mL ScrewTop Tubes
    Colorless
    V-bottomYesYes96/rack480Order
    3740RED
    Matrix 1.0 mL ScrewTop Tubes
    Red
    V-bottomNoYesNA480Order
    3741RED
    Matrix 1.0 mL ScrewTop Tubes
    Red
    V-bottomYesYes96/rack480Order
    3740YEL
    Matrix 1.0 mL ScrewTop Tubes
    Yellow
    V-bottomNoYesNA480Order
    3741YEL
    Matrix 1.0 mL ScrewTop Tubes
    Yellow
    V-bottomYesYes96/rack480Order
    3740GRE
    Matrix 1.0 mL ScrewTop Tubes
    Green
    V-bottomNoYesNA480Order
    3740BLU
    Matrix 1.0 mL ScrewTop Tubes
    Blue
    V-bottomNoYesNA480Order
    3741BLU
    Matrix 1.0 mL ScrewTop Tubes
    Blue
    V-bottomYesYes96/rack480Order
    3740PUR
    Matrix 1.0 mL ScrewTop Tubes
    Purple
    V-bottomNoYesNA480Order
    3740WHI
    Matrix 1.0 mL ScrewTop Tubes
    White
    V-bottomNoYesNA480Order
    1.4 mL storage tubes
    A storage rack filled with 1.4 mL tubes
    Matrix 1.4 mL blank and alphanumeric storage tubes feature:
    • Virgin-polypropylene-resin composition
    • Alphanumeric codes on tube bottoms
    • A variety of cap options
    Order matrix storage tube racks
    Cat. No.DescriptionBarcodeWith latch rackSterilePack sizeCase 
    4253
    Matrix Blank and Alphanumeric Storage Tubes
    Alphanumeric
    NoNo10/carrier960Order
    4140
    Matrix Blank and Alphanumeric Storage Tubes
    Blank
    NoNoNA1000Order
    4147
    Matrix Blank and Alphanumeric Storage Tubes
    Alphanumeric
    YesNo10/rack960Order
    4250
    Matrix Blank and Alphanumeric Storage Tubes
    Blank
    NoYes10/carrier960Order
    Return to top of tab
    12 mL storage culture tubes
    A single unfilled 12 mL storage tube on white background
    Matrix 12 mL storage tubes feature:
    • Laser-etched 2D barcode on tube bottom
    • Tested against leaks
    • Built-in error correction
    Order matrix storage tube racks
    Cat. No.DescriptionSurfaceTube shapeWith rackSterilePack sizeCase 
    3778
    Matrix 2D Barcoded 12 mL Screw-top Storage Tubes
    Untreated
    RoundNoYes4/2496Order
    3776
    Matrix 2D Barcoded 12 mL Screw-top Storage Tubes
    Untreated
    RoundYesNoNA4Order
    3777TS
    Matrix 2D Barcoded 12 mL Screw-top Storage Tubes
    Untreated
    RoundYesNoNAEachOrder
    3770
    Matrix 2D Barcoded 12 mL Screw-top Storage Tubes
    Untreated
    RoundNoNoNA100Order
    3774
    Matrix 2D Barcoded 12 mL Screw-top Storage Tubes
    Untreated
    RoundYesNoNAEachOrder
    3775
    Matrix 2D Barcoded 12 mL Screw-top Storage Tubes
    Untreated
    RoundYesYes4/2496Order
    500 µL storage tubes
    A storage rack holding a variety of 500 µL tubes with various colored caps
    Matrix 500 µL V-bottom storage tubes feature:
    • Virgin-polypropylene-resin composition
    • ScrewTop caps with soft silicone gaskets
    • Stringent 2D barcode quality control
    Order matrix storage tube racks
    Cat. No.DescriptionColorTube shapeWith rackSterilePack sizeCase 
    3743
    Matrix 500 μL ScrewTop Tubes
    Colorless
    V-bottomNoYesNA480Order
    3744
    Matrix 500 μL ScrewTop Tubes
    Colorless
    V-bottomYesYes96/rack480Order
    3743RED
    Matrix 500 μL ScrewTop Tubes
    Red
    V-bottomNoYesNA480Order
    3744RED
    Matrix 500 μL ScrewTop Tubes
    Red
    V-bottomYesYes96/rack480Order
    3743YEL
    Matrix 500 μL ScrewTop Tubes
    Yellow
    V-bottomNoYesNA480Order
    3744YEL
    Matrix 500 μL ScrewTop Tubes
    Yellow
    V-bottomYesYes96/rack480Order
    3743GRE
    Matrix 500μL ScrewTop Tubes
    Green
    V-bottomNoYesNA480Order
    3743BLU
    Matrix 500 μL ScrewTop Tubes
    Blue
    V-bottomNoYesNA480Order
    3743PUR
    Matrix 500 μL ScrewTop Tubes
    Purple
    V-bottomNoYesNA480Order
    3743WHI
    Matrix 500 μL ScrewTop Tubes
    White
    V-bottomNoYesNA480Order
    0.75 mL storage tubes
    A storage rack filled with 0.75 mL tubes
    Matrix 0.75 mL blank and alphanumeric storage tubes feature:
    • Virgin-polypropylene-resin composition
    • Alphanumeric codes on tube bottoms
    • A variety of cap options
    Order matrix storage tube racks
    Cat. No.DescriptionBarcodeWith latch rackSterilePack sizeCase 
    4170
    Matrix Blank and Alphanumeric Storage Tubes
    Blank
    NoNoNA1000Order
    4272
    Matrix Blank and Alphanumeric Storage Tubes
    Blank
    YesYes10/rack960Order
    4273
    Matrix Blank and Alphanumeric Storage Tubes
    Alphanumeric
    YesNo10/rack960Order
    1 mL storage tubes
    A storage rack filled with 1 mL tubes with various cap colors
    Matrix 1 mL V-bottom storage tubes feature:
    • Virgin-polypropylene-resin composition
    • ScrewTop caps with soft silicone gaskets
    • Stringent 2D barcode quality control
    Order matrix storage tube racks
    Cat. No.DescriptionColorTube shapeWith rackSterilePack sizeCase 
    3741
    Matrix 1.0 mL ScrewTop Tubes
    Colorless
    V-bottomYesYes96/rack480Order
    3740RED
    Matrix 1.0 mL ScrewTop Tubes
    Red
    V-bottomNoYesNA480Order
    3741RED
    Matrix 1.0 mL ScrewTop Tubes
    Red
    V-bottomYesYes96/rack480Order
    3740YEL
    Matrix 1.0 mL ScrewTop Tubes
    Yellow
    V-bottomNoYesNA480Order
    3741YEL
    Matrix 1.0 mL ScrewTop Tubes
    Yellow
    V-bottomYesYes96/rack480Order
    3740GRE
    Matrix 1.0 mL ScrewTop Tubes
    Green
    V-bottomNoYesNA480Order
    3740BLU
    Matrix 1.0 mL ScrewTop Tubes
    Blue
    V-bottomNoYesNA480Order
    3741BLU
    Matrix 1.0 mL ScrewTop Tubes
    Blue
    V-bottomYesYes96/rack480Order
    3740PUR
    Matrix 1.0 mL ScrewTop Tubes
    Purple
    V-bottomNoYesNA480Order
    3740WHI
    Matrix 1.0 mL ScrewTop Tubes
    White
    V-bottomNoYesNA480Order
    1.4 mL storage tubes
    A storage rack filled with 1.4 mL tubes
    Matrix 1.4 mL blank and alphanumeric storage tubes feature:
    • Virgin-polypropylene-resin composition
    • Alphanumeric codes on tube bottoms
    • A variety of cap options
    Order matrix storage tube racks
    Cat. No.DescriptionBarcodeWith latch rackSterilePack sizeCase 
    4253
    Matrix Blank and Alphanumeric Storage Tubes
    Alphanumeric
    NoNo10/carrier960Order
    4140
    Matrix Blank and Alphanumeric Storage Tubes
    Blank
    NoNoNA1000Order
    4147
    Matrix Blank and Alphanumeric Storage Tubes
    Alphanumeric
    YesNo10/rack960Order
    4250
    Matrix Blank and Alphanumeric Storage Tubes
    Blank
    NoYes10/carrier960Order
    Return to top of tab
    12 mL storage culture tubes
    A single unfilled 12 mL storage tube on white background
    Matrix 12 mL storage tubes feature:
    • Laser-etched 2D barcode on tube bottom
    • Tested against leaks
    • Built-in error correction
    Order matrix storage tube racks
    Cat. No.DescriptionSurfaceTube shapeWith rackSterilePack sizeCase 
    3778
    Matrix 2D Barcoded 12 mL Screw-top Storage Tubes
    Untreated
    RoundNoYes4/2496Order
    3776
    Matrix 2D Barcoded 12 mL Screw-top Storage Tubes
    Untreated
    RoundYesNoNA4Order
    3777TS
    Matrix 2D Barcoded 12 mL Screw-top Storage Tubes
    Untreated
    RoundYesNoNAEachOrder
    3770
    Matrix 2D Barcoded 12 mL Screw-top Storage Tubes
    Untreated
    RoundNoNoNA100Order
    3774
    Matrix 2D Barcoded 12 mL Screw-top Storage Tubes
    Untreated
    RoundYesNoNAEachOrder
    3775
    Matrix 2D Barcoded 12 mL Screw-top Storage Tubes
    Untreated
    RoundYesYes4/2496Order