How can pet allergen components help with allergy treatment?
Cat allergy
Cat allergen components provide an array of information on cat allergies and other food or respiratory diseases.1-3 Some of these components can indicate if someone is at a high risk of developing asthma or if they might have pork-cat syndrome, which is a type of allergy that arises from eating pork.2 Cat allergen components also help allergists optimize their immunotherapy so that they can administer the most relevant allergy shots.1-3
It's worth noting that the Fel d 1 cat allergen component, which is produced in a cat's salivary and sebaceous glands, can be transferred to a cat's fur and dander through licking and grooming. 5 Therefore, as a cat sheds, Fel d 1 becomes scattered around your home and on your clothes.
Dog allergy
Similar to cat allergen-components, dog allergen components can provide information to healthcare providers that help them decide on the best types of allergy shots to use during immunotherapy. Dog allergen components also help with determining how at risk someone may be for severe asthma, allowing them to understand how risky it may be to bring a dog into their home.1-3
Contrary to popular belief, hypoallergenic dogs aren't "allergen free."2 Many of these types of dogs, such as the poodle, simply shed less meaning that there is less animal dander floating about. All breeds produce the proteins that make up the allergens found in saliva, urine, and dander, and while these levels can vary, there is no such thing as a completely "allergen free" pup.2
However, the "Can f 5" dog-allergen component is only produced in male dogs, meaning that if you are only allergic to that specific component you may be able to tolerate a female dog and have fewer allergic reactions, which is why it's valuable to get tested with allergen components if you are considering adopting a dog.1-3